Detect, localize, identify, and measure radioactivity in any scenario
From routine monitoring and surveillance to emergency response situations, Thermo Fisher Scientific’s advanced, integrated radiation detection and measurement instruments mitigate radiological threats to keep you safe. Our product portfolio of radiation detectors, radiation monitoring devices, and radiation measurement instruments provide comprehensive, real-time monitoring, early warning, and complete dose and exposure information in the palm of your hand, in the workplace, and in your neighborhood.
Hazmat team scans for radiation at site of demolished building
Dosimetry instruments
From individual personnel monitoring to enterprise-wide dose management solutions, our range of dosimetry products and services can meet your current and future radiation protection requirements.
NetDose Pro
Customize seamlessly to your facilities’ needs, eliminate the need to collect and redistribute badges and simplify investigations with to-the-minute exposure information. The Thermo Scientific NetDose Pro Digital Dosimeter is a comprehensive solution for real-time personal dose monitoring, improving safety, and reducing cost.
RadHalo at LAX airport scanning arriving cars for radiation sources
EPD TruDose Electronic Personal Dosimeter (EPD)
The Thermo Scientific EPD TruDose Electronic Personal Dosimeter (EPD) monitors gamma, beta, and neutron radiation in any environment where workers need to monitor their exposure to radiation, with improved accuracy and simplified operation over previous generations of EPDs.
RadHalo at LAX airport scanning arriving cars for radiation sources
TLD readers, cards and materials
Thermo Scientific TLD readers, cards and detector material combine to create cost-effective systems to measure radiation dose in medical, nuclear, research, and health physics applications.
RadHalo at LAX airport scanning arriving cars for radiation sources
Handheld instruments
Protection goes wherever you go with our compact, portable radiation detection and identification instruments. We offer the pager sized Thermo Scientific RadEye line, featuring patented Natural Background Rejection (NBR) technology, to detect, locate, and identify radioactive nuclides such as nuclear weapons, dirty bombs, and orphaned or purposely masked sources. We also offer a wide range of alpha and beta probes well suited for applications in nuclear medical labs, industrial facilities, geological surveys, hospitals, and academia.
RadEye PRD4 and PRD-ER4 Personal Radiation Detectors
The Thermo Scientific RadEye PRD4 and PRD-ER4 Personal Radiation Detectors enable accurate dose rate measurements across the broader gamma spectrum, providing a greater assurance of personal safety. The neutron indication capability of the RadEye PRD4 provides an additional layer of detection critical to determining the level of radiation threat.
RadEye PRD4 scanning waste container for discarded radiation sources
RadEye SPRD and SPRD-ER Personal Radiation Detectors
The Thermo Scientific RadEye SPRD and SPRD-ER Personal Radiation Detectors are handheld radiation detectors which identify and categorize specific types of radiation or radioactive material and can be configured to support your missions or user skills. The RadEye SPRD and SPRD-ER offer a unique combination of detector sensitivity and intelligent alarming to help ensure detection in the most challenging applications while eliminating nuisance alarms.
RadEye SPRD Personal Radiation Detector identifies a natural source of radiation
RadEye SPRD-GN Spectroscopic Personal Radiation Detector
Get RIID capability in the palm of your hand. The Thermo Scientific RadEye SPRD-GN Personal Radiation Detector is the first pager-sized PRD that significantly exceeds the original ANSI N42-48 2008 standards for neutron alarms. With outstanding neutron performance and reliable gamma IDs, the RadEye SPRD-GN Personal Radiation Detector may be the only radiation detection tool you will need.
RadEye SPRD-GN Personal radiation detector
RadEye G Series Personal Dose Rate Meters
Continuously monitor your radiation exposure in any nuclear or radiological emergency. The Thermo Scientific RadEye G Series of personal radiation detectors provide first responders with quick, reliable gamma dose rate measurements. These lightweight, handheld Geiger counters detect and measure very low gamma energies; even the smallest change in radiation rates are displayed immediately. For hazmat teams investigating potentially explosive atmospheres, intrinsically safe RadEye G models meet the latest ATEX standards.
RadEye G Gamma Survey Meter
RadEye X Series
Be prepared for virtually any radiation contamination scenario. The Thermo Scientific RadEye X Series Survey Meters comprise three different types of compact, multi-purpose survey meters suited for a wide range of applications where personnel risk radiation exposure, whether they are emergency responders, fire fighters, nuclear power plant employees, or medical professionals. RadEye X Series Survey Meters can operate with virtually any manufacturer's Geiger-Mueller, scintillation, or proportional detectors to perform general count rate, surface contamination, and dose rate measurements.
RadEye GX Survey Meter and probes for detecting radiation
RadEyeB20 and B20-ER Multi-Purpose Survey Meters
First responders need to quickly identify mixed radioactive surface contamination in facility and field environments. Thermo Scientific RadEye B20 and B20-ER Multi-Purpose Survey Meters are simple, robust, reliable contamination and dose rate measurement tools for characterizing alpha, beta, gamma and X-ray radiation. RadEye B20 and B20-ER Multi-Purpose handheld Geiger counters provide an on-site solution for homeland security personnel, fire brigades, emergency response personnel, agencies involved in decontamination and decommissioning projects, and hospital and pharmaceutical industry employees.
RadEye B20-ER Survey Meter
FH40 G Multi-Purpose Digital Survey Meter
The Thermo Scientific FH 40 G Multi-Purpose Digital Survey Meter is a wide range digital Geiger counter suitable for nearly all measurement tasks arising in radiation protection through optional plug and play probes available for neutron measurement, alpha and beta contamination readings and even to detect artificial gamma radiation in variable natural radiation fields.
Thermo Scientific FH 40 Gwide range digital Geiger counter
RadEye SPRD scanning shipping container for radiation sources
Accurate results with natural background rejection
Naturally occurring radiation exists in varying levels from the earth and atmosphere as well as non-threatening artificial radiation exists from medical treatments or as sources in gauging instruments, which can obscure and confuse targeted detection efforts. 
Natural Background Rejection (NBR) is a technology used to eliminate fluctuating natural background levels without sacrificing sensitivity. This proprietary and patented technology quickly differentiates between natural and artificial radiation and strips it from the measurement, delivering you an instant and more accurate result of artificial radiation levels. Operators using instruments without NBR often set their alarm thresholds higher to eliminate the nuisances of false alarms or ignore alarms due to their frequency potentially missing out on hidden or shielded sources. Using an instrument with NBR allows you to keep your alarm threshold extremely low without the need to worry about false alarms from non-threatening sources.  When a RadEye the alarm goes off, you know it is time to take actionact.
Answer a few questions about your application and discover the right RadEye personal radiation detector for your job. 
Portable instruments
Our portable radiation detectors are designed to meet a diverse range of working environments. With key ease of use and advanced detections features, our portable radiation detection products ensure the safety and peace of mind of users. Backed by our decades of experience in radiation detection technology, Thermo Fisher Scientific continues to innovate and provide cutting-edge solutions for portable radiation detection.
RadHalo RDP and FM Spectroscopic Area Monitors
The Thermo Scientific RadHalo RDP (portable drop probe) and FM (fixed install) Spectroscopic Area Monitors detect and identify radiation on location or from miles away.
RadHalo at LAX airport scanning arriving cars for radiation sources
PackEye Radiation Detection Backpack
The Thermo Scientific PackEye Radiation Detection Backpack provides your protection team with a tool for effectively finding orphaned sources, radiological contamination, and maliciously introduced sources in a large area while maintaining a discreet form factor. 
RadHalo at LAX airport scanning arriving cars for radiation sources
RIIDEye Radiation Isotope Identifier
The Thermo Scientific RIIDEye X and RIIDEye M Handheld Radioisotope Identifiers, with patented Quadratic Compression Conversion (QCC) technology, are well suited to support users seeking quick and accurate identification of specific radiation sources.
RadHalo at LAX airport scanning arriving cars for radiation sources
Portal monitors
Thermo Scientific portal monitoring systems deliver highly sensitive, accurate detection of illicit nuclear materials inside packages or cargo at airports, seaports, borders, government buildings, food storage and handling facilities, transportation stations, couriers, and freight companies. Our portal monitors also help scrap metal facilities avoid costly and damaging contamination events from uncontrolled sources.
ASM IV Series Automatic Scrap Monitoring System
Radioactively contaminated scrap threatens both the workers who handle it and consumers of products made with recycled scrap metal. The Thermo Scientific ASM IV Series Automatic Scrap Monitoring System monitors vehicles entering the scrap yard, steel mill, or processes within the facility, and provides accurate, reliable detection of radiation sources commonly found in scrap metal. With high performance and a low false alarm rate, the system rejects radioactive materials before they can harm people, facilities or the environment.
Delivery of recycled material being scanned for radiation using ASM IV system
Safety Guard Series II System
Thermo Scientific Safety-Guard Series II System for of radiation detection systems offers optimized protection against illicit trafficking of nuclear materials. Offer The system offers significant gains in detection sensitivity for screening packages, people, or vehicles, for incident prevention and response.
train on tracks
Contamination monitors
Achieve the best radiation contamination detection limits possible with Thermo Scientific radiation contamination measurement systems. Employees who work in radiation-controlled areas of nuclear power plants and nuclear facilities can become contaminated with trace amounts of radiation on their bodies, clothes, and equipment. To prevent the spread of radioactive contamination beyond the controlled area and keep outside areas secure, we offer a range of personnel contamination monitors to survey the body, hands and feet, small and large articles, and laundry.
SAM12 Small Article and Tool Monitor
The Thermo Scientific SAM12 Small Articles Monitor is a well-proven, reliable, rugged instrument that monitors articles for gamma-emitting radionuclides and easily achieves clearance limits for higher energy gamma emitters such as 137Cs and  60Co. This monitor allows dynamic discrimination between natural (NORM) and man-made radiation and offers a unique feature for  60Co monitoring.
Small articles are scanned for radiation using the SAM 12
IPCM12 Personal Contamination Monitor
The Thermo Scientific iPCM12 Installed Personnel Contamination Monitor identifies surface contamination on the body, hands, and feet using 21 detectors to provide excellent alpha and low energy beta detection. Detectors are split into four zones to minimize background and achieve high detection limits to get personnel cleared quickly.
iPCM12 monitors for radiation presence on the body
IPM96 Whole Body Contamination Monitor
Thoroughly scan for beta/gamma contamination on the entire body surface with the Thermo Scientific IPM96 Whole Body Contamination Monitor.  Through innovative features including new Thermo Scientific Thorough-Scan gasless scintillation detector technology, reduced count times, and the highest number of detectors available in the industry, this radiation contamination portal monitor increases the accuracy of body scans and most importantly, provides confidence that your radiation-controlled facility delivers on the promise of worker safety.
IPM96 scans an employee for radiation exposure prior to leaving facility
Environmental monitors
Keep your plant, the environment, and surrounding communities safe from nuclear radiation leaks with Thermo Scientific environmental monitors. We can help you achieve event-free operation with reliable, continuous environmental radiation monitoring and assessment equipment to meet the most demanding safety objectives within and outside of your facility. Our environmental monitors support and sustain nuclear safety by measuring radioactive material or noble gases released by nuclear power plants, and by detecting airborne alpha- and beta-emitting particulates in work areas, stacks, and ducts.
PM12 Personnel Gamma Portal Monitor
The Thermo Scientific PM12 Personnel Gamma Portal Monitor provides simplified but advanced monitoring capability for workers, articles and waste well below clearance levels.
Thermo Scientific™ PM12 Personnel Gamma Portal Monitor
For radiation measurement, identification, or monitoring only.
1x1 image pixel for data collection1x1 image pixel for data collection