Ready-to-use standards enable quick instrument calibration
by John C. Rogers, Ph.D.; Michael M. Rosenblatt, Ph.D.; Eugene J. Cichon, B.S.; Rosa Viner, Ph.D.*; Monica Noonan, M.S. - 06/01/11
Routine calibration of mass spectrometers is required for optimal performance. Our new calibration solutions for Thermo Scientific Mass Spectrometers are ready-to-use liquid formulations for quick and easy instrument calibration.
Calibration solutions for Thermo Scientific Ion Trap and Orbitrap Mass Spectrometers
Calibration of Thermo Scientific Ion Trap and Orbitrap Mass Spectrometers is required for proper instrument performance. These high-quality calibration solutions are for regulated environments and busy laboratories and avoid time-consuming reagent preparation. Reagents are provided in high-purity Teflon*-coated containers tested rigorously to provide the highest batch-to-batch reproducibility (Figures 1 and 2).
88323-v15-Fig1-VelosPos
Figure 1. Representative mass spectrum for the Thermo Scientific Pierce LTQ Velos ESI Positive Ion Calibration Solution. N-butylamine, caffeine, Met-Arg-Phe-Ala (MRFA) and Ultramark 1621 are used to calibrate Thermo Scientific LTQ Velos Ion Trap and Orbitrap Mass Spectrometers in the ESI positive ionization mode. The N-butylamine is included to extend to low mass calibration range.
88324-v15-Fig2-ESI-Neg
Figure 2. Representative mass spectrum for the negative ion calibration. Sodium dodecyl sulfate, sodium taurocholate and Ultramark 1621, comprise the calibration mixture used to calibrate Ion Trap and Orbitrap* Mass Spectrometers in the negative ionization mode.
Calibration solutions for Thermo Scientific Triple Quadrupole Mass Spectrometers
Thermo Scientific TSQ Mass Spectrometers are calibrated with multimers of tyrosine (Tyr1, Tyr3, Tyr6). All of the components of this mixture are prepared from the highest purity raw materials and are stored in inert Teflon-coated containers (Figure 3).
88325-v15-Fig3-TripleQuad
Figure 3. Representative mass spectrum for the triple-quadrupole MS calibration. Multimers of tyrosine are used to calibrate the TSQ series of Triple Quadrupole Mass Spectrometers.
LTQ ESI Positive Ion Calibration Solution
The Thermo Scientific Pierce LTQ ESI Positive Ion Calibration Solution is a mixture of highly purified ionizable molecules specifically designed for positive mode calibration of Thermo Scientific ion trap and orbitrap instruments.
Features of LTQ ESI Positive Ion Calibration Solution:
  • Strong peaks—mixture of caffeine, MRFA and Ultramark 1621 in a acetonitrile/methanol/acetic solution.
  • Ready-to-use—load the mass reference standard into a syringe and inject into the instrument
  • High-purity—mass spectrometry grade reagents in a non-leachable container
  • Stable—store at room temperature for more than one year
Learn more about Thermo Scientific Pierce LTQ ESI Positive Ion Calibration Solution
Related products
Catalog # Name Size Price (USD) Qty
88323 Pierce™ LTQ Velos ESI Positive Ion Calibration Solution Each
302.00
88324 Pierce™ Negative Ion Calibration Solution Each
267.65
Online Exclusive
302.00
Save 34.35 (11%)
88325 Pierce™ Triple Quadrupole Calibration Solution 10 mL
276.65
Online Exclusive
302.00
Save 25.35 (8%)
88320 Pierce™ Peptide Retention Time Calibration Mixture Each
108.65
Online Exclusive
124.00
Save 15.35 (12%)
88321 Pierce™ Peptide Retention Time Calibration Mixture Each
701.65
Online Exclusive
792.00
Save 90.35 (11%)
88326 Pierce™ Reserpine Standard for LC-MS Each
161.65
Online Exclusive
176.00
Save 14.35 (8%)
88328 Pierce™ HeLa Protein Digest Standard, 20 μg Each
141.65
Online Exclusive
169.00
Save 27.35 (16%)
88329 Pierce™ HeLa Protein Digest Standard, 5 x 20 μg Each
551.65
Online Exclusive
654.00
Save 102.35 (16%)
For Research Use Only. Not for use in diagnostic procedures.