Pierce™ LTQ ESI Positive Ion Calibration Solution
Pierce™ LTQ ESI Positive Ion Calibration Solution
Thermo Scientific™

Pierce™ LTQ ESI Positive Ion Calibration Solution

The Thermo Scientific Pierce LTQ ESI Positive Ion Calibration Solution is a mixture of highly purified ionizable molecules specifically designedRead more
Have Questions?
Catalog NumberQuantity
8832210 mL
Catalog number 88322
Price (USD)
256.70
Special offer
Online exclusive
Ends: 15-Aug-2025
302.00
Save 45.30 (15%)
Each
In stock
Add to cart
Quantity:
10 mL
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
256.70
Special offer
Online exclusive
Ends: 15-Aug-2025
302.00
Save 45.30 (15%)
Each
Add to cart
Ask our AI about this Product
The Thermo Scientific Pierce LTQ ESI Positive Ion Calibration Solution is a mixture of highly purified ionizable molecules specifically designed for positive mode calibration of Thermo Scientific ion trap and orbitrap instruments.

Features of LTQ ESI Positive Ion Calibration Solution:

Strong peaks—mixture of caffeine, MRFA and Ultramark 1621 in a solution of acetonitrile, methanol, and acetic acid
Ready-to-use—load the mass reference standard into a syringe and inject into the instrument
High-purity—mass spectrometry grade reagents in a non-leachable container

The Pierce LTQ ESI Positive Ion Calibration Solution is a ready-to-use liquid formulation ideal for quickly performing the routine calibration required to maintain the robust performance of Thermo Scientific Mass Spectrometers. The LTQ ESI Positive Ion Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.

The LTQ ESI Positive Ion Calibration Solution can be used to calibrate the LTQ series, the LTQ Orbitrap series, the LXQ, LCQ FLEET and the Exactive Mass Spectrometer instruments.

More Product Data
Calibration solutions for Thermo Scientific Mass Spectrometers
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypePeptides
For Use With (Equipment)Mass Spectrometer
Quantity10 mL
Workflow StepCalibration
Detection MethodMass Spectrometry
FormLiquid
Product LinePierce™
Product TypeCalibration Solution
Starting MaterialPeptides
Unit SizeEach
Contents & Storage
Store between 2°C and 8°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
AA407403Certificate of AnalysisJan 28, 202588322
ZI395573Certificate of AnalysisOct 16, 202488322
ZD388766Certificate of AnalysisMay 17, 202488322
YK388765Certificate of AnalysisJan 25, 202488322
YI353433Certificate of AnalysisJan 25, 202488322
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection