Objectively precise, incredibly easy
The EVOS M3000 Imaging System was designed to be the workhorse of any cell culture or research laboratory by democratizing the ability to produce reliable, stunning high-quality cell images and video. This system automatically and precisely measures cell confluency in real time, with no expertise required. This compact, versatile, easy-to-use system is capable of two-color fluorescence, color-imaging, brightfield, and phase contrast, making it your go-to manual microscope.
Features
In addition to the common features of all EVOS microscopes, the EVOS M3000 Imaging System offers:
Cell confluency in seconds
Built-in automated real-time image analysis for measuring cell confluency
Versatile
Capture images in brightfield, phase contrast, color imaging, and fluorescence
Flexible
Compatible with the full selection of EVOS objectives (1.25-60X) and light cubes
No training required
Simple user interface with touchscreen display
Space-saving design
Our smallest EVOS footprint fits easily on the lab bench or in a cell culture hood
Connectivity
Network-capable instrument facilitates seamless data transfer, storage and collaboration enhancing productivity and data management
EVOS real-time confluency tool
The patent-pending EVOS real-time confluency tool can typically measure confluency in less than 1 second without requiring any image capture, making it excellent for eliminating user bias in routine cell culture. The output from this proprietary bioapplication correlates well with confluency measurements from other software, e.g., Cell Profiler, and is standard on all EVOS M3000 instruments. 
Cell imaging data showing induced pluripotent stem cells imaged over time with the EVOS M3000 showing cell confluency
Induced pluripotent stem cells imaged over time with the EVOS M3000 instrument. Human fibroblast-derived induced pluripotent stem cells were cultured on a vitronectin-coated 6-well plate in Gibco Essential 8 Flex Medium for 3 days. Cells were imaged with the EVOS M3000 instrument under phase microscopy with and without the automatically generated confluency mask and measurement.
Decorative image
How accurately can you measure confluency in stem cell culture images?
Put your skills to the test and learn about tools for more consistent and accurate cell confluency measurement in our latest blog.
Accessories
EVOS objectives
Visit EVOS Cell Imaging System Accessories to learn more about the complete objectives available for EVOS Imaging Systems.
EVOS light cubes
EVOS LED-based light cubes provide excellent illumination uniformity with maximal signal-to-background ratios and minimal bleed-through. EVOS light cubes are user-changeable and automatically recognized for plug-and play across the spectrum and across EVOS imaging systems. Light cubes are available individually or as convenient light cube starter kits.
*This is not a complete list of compatible dyes. 
The Fluorescence SpectraViewer tool is designed to help optimize light cube and dye compatibility for your EVOS or Countess imaging systems. 
EVOS M3000 mechanical stage
The EVOS M3000 Mechanical Stage is an optional addition to the universal, fixed stage provided with the EVOS M3000 Imaging System. This mechanical stage has X-Y axis controls and a vessel holder framework that accommodates our broad range of vessel holders. When using the mechanical stage, vessel holders and large vessels can be held securely in place by a stage clip during operation.
Video Player is loading.
Current Time 0:00
Duration 7:24
Loaded: 0.00%
Stream Type LIVE
Remaining Time 7:24
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Follow step-by-step instructions on how to install the EVOS M3000 Imaging System.
    Video Player is loading.
    Current Time 0:00
    Duration 2:52
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 2:52
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Learn how to use the real-time cell confluency algorithm on the EVOS M3000 Imaging System for unbiased, accurate cell confluency measurements.
      FluoCells imaged on the EVOS M3000 imaging system using GFP and RFP EVOS light cubes
      FluoCells prepared slide #1 (Cat. No. F36924) showing bovine pulmonary artery endothelial cells (BPAE) labeled with MitoTracker Red CMXRos (Cat. No. M7512) for mitochondria, and then fixed, permeabilized and stained with Alexa Fluor 488 phalloidin (Cat. No. A12379) for F-actin. This two-channel image was acquired on an EVOS M3000 Imaging System using GFP and RFP EVOS Light Cubes (Cat. No. AMEP4951 and AMEP4952) and an Olympus 60X 1.42 NA oil-immersion objective (Cat. No. AMEP4910).
      iPSC cells expressing GFP imaged on the EVOS M3000 Imaging System
      Fixed tumoroids expressing GFP grown in OncoPro Tumoroid Culture Medium Kit (Cat. No. A5701201) on Nunc Cell-Culture Treated Multidishes (Cat. No. 140685). Two-channel image was acquired on an EVOS M3000 Imaging System using an EVOS 20X Objective (Cat.No. AMEP4982) for phase contrast and an EVOS GFP Light Cube (Cat. No. AMEP4951).
      Video Player is loading.
      Current Time 0:00
      Duration 0:19
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:19
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • default, selected
        Achieve optimal video quality when recording images captured with transmitted light on the EVOS M3000 Imaging System. Shown here are iPSCs recorded using an EVOS 4X Objective (Cat. No. AMEP4980) for phase contrast.
        Specifications