Essential 8 Flex Medium
Gibco Essential 8 Flex Medium is formulated to extend the activity of key heat-sensitive components found in PSC medium, including FGF2, to enable truly weekend-free culture with a flexible feeding schedule.
  • Flexible feeding schedule—maintain pluripotency over a full 2-day period without feeding cells
  • Robust cultures—optimized stability of heat sensitive components, like FGF2, increases maintenance of pluripotency
  • Easy to transition—same setup, applications and protocol as the original Essential 8 Medium formulation, but without the daily feeding
  • Proven—based on the original Gibco Essential 8 Medium formulation, maintains pluripotency and normal karyotypes in long-term culture
Returning to the lab?
Maximize your time with Essential 8 Flex Medium
Essential 8 Flex Medium is designed to eliminate the need to manage cultures daily which allows for more flexible feeding schedules, including up to two consecutive feed-free days, so that your cultures can be adapted to your schedule. 
Gibco PSC Media are manufactured according to cGMP for medical devices, 21 CFR Part 820
View webinar
Video Player is loading.
Current Time 0:00
Duration 38:38
Loaded: 0%
Stream Type LIVE
Remaining Time 38:38
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Routine PSC expansion and maintenance without the need for daily feeding
    Essential 8 Flex Medium is a serum-free, xeno-free medium that supports the culture and expansion of pluripotent stem cells (PSCs) while eliminating the need for daily feeding schedules (Figure 1).  Just like the original Essential 8 Medium formulation, the Essential 8 Flex Medium is xeno-free and does not include BSA (bovine serum albumin) or HSA (human serum albumin) components, helping to minimize batch variability and maintain long-term health and pluripotency of PSCs (Figures 2–3).
    Illustration demonstrating the Essential 8 Flex Medium flexible feeding schedule and a typical 7-days-per-week feeding schedule for human PSCs when using traditional media.
    Figure 1. Culture schedule comparison. Unlike other feeder-free media, Gibco Essential 8 Flex Medium eliminates the need to manage cultures daily, enabling a truly weekend-free schedule for expansion and maintenance of PSCs. The Essential 8 Flex medium workflow can be adjusted to a once-per-week passage if required to fit existing workflows. Simply change day 1 to a feed day in the workflow above.
    Line graph showing how Essential 8 Flex Medium preserves FGF2 activity when viewing growth factor activity as a function of time.
    Karyotype shown as 22 pairs of somatic chromosomes and one pair of sex chromosomes stained by G-banding.
    Figure 2. FGF2 activity over time in PSC culture medium. Unlike other feeder-free PSC culture media, Essential 8 Flex Medium has been optimized to extend the activity of unstable components such as FGF2. Extended activity allows for routine culture without the daily feeding.
    Figure 3. Long-term stability of cells cultured in Essential 8 Flex medium.  Healthy karyotypes of pluripotent stem cells cultured in Essential 8 Flex medium have been observed after long term culture of cells.
    Robust pluripotent stem cell cultures
    FGF2 levels widely fluctuate in standard PSC culture medium, even with daily feeding (Figure 4). The use of Essential 8 Flex medium significantly limits these fluctuations, producing more robust, stable culture conditions with the added value of a flexible feeding schedule.
    Line graph showing relatively stable FGF2 activity with Essential 8 Flex Medium over the course of a week. While cells experience large fluctuations in FGF2 activity in a competitor's medium.
    Figure 4. Essential 8 Flex medium limits fluctuations in levels of active FGF2. Measurements of heat-mediated loss of active FGF2 were used to simulate active FGF2 levels in PSC cultures over the course of a week. Standard PSC culture medium (red curve) cultures would be fed on a daily basis while Essential 8 Flex medium (blue curves) cultures would be follow the suggested protocol outlined in Figure 1.
    Complete media system
    The following components are recommended to create a complete media system for weekend-free routine expansion and maintenance of PSCs with Essential 8 Flex Medium.
    Essential 8 Flex Protocol
    Matrix
    Passaging reagent
    Webinar: Essential 8 Flex: Eliminating daily feeding in a feeder-free, xeno-free PSC culture system
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00