Cells cultured in 2D can differ in terms of both physiology and cellular responses compared with cells in vivo. These differences have led to a surge in the popularity of using 3D culture techniques. Mounting evidence suggests that culturing cells in 3D is more representative of the in vivo environment, creating more physiological cell models, even to the extent that the gene expression profiles of cells from 3D cultures more accurately reflect clinical expression profiles than those observed in 2D cultures [1,2]. 
Spheroids
Spheroids, or sphere cultures, have become an especially exciting area of 3D in vitro culture due to their great potential for use in studies that investigate growth and function of both malignant and normal tissues. These sphere cultures have contributed considerably to our knowledge of cellular responses thanks to the accuracy with which they reflect the in vivo system. This is primarily because cells do not normally grow or interact in isolation, but instead form complex interactions with other cells and the surrounding microenvironment. Thus, the creation of a 3D environment that incorporates spheroids more closely mimics in vivo conditions, allowing researchers to incorporate cell-cell interactions, nutrient gradients, and diffusion kinetics in their in vitro models.
Spheroids offer particular benefits in cancer biology, where they contribute immense value in examining the growth and behavior of tumors since they share several key histomorphological and functional traits that include the formation of cell-cell contacts, decreased proliferation, increased survival rates, and a hypoxic core [3,4]. As more researchers recognize the benefits that spheroid cultures provide as a cell model, development efforts have increased to better aid spheroid generation, culture, and scale-up. Researchers are now moving toward advanced culture methods, employing hypoxic conditions, or coculturing with different cell types to develop increasingly accurate in vitro models of disease and physiology. 
Spheroids and organoids are both three-dimensional cell culture systems that more accurately mimic the in vivo environment compared to traditional two-dimensional cultures. However, organoids are more complex structures derived from stem cells that self-organize into miniature, functional replicas of organs, displaying multiple cell types and tissue-specific architecture. The primary difference between spheroids and organoids lies in their complexity and functionality; spheroids are generally simpler and less organized, while organoids exhibit higher levels of cellular differentiation and organization, closely resembling the structure and function of actual organs. This distinction makes organoids particularly valuable for studying organ development, disease modeling, and personalized medicine, whereas spheroids are often used for more straightforward applications such as drug screening and basic cell biology research.
Brief history of spheroid development
Researchers have cultured cells in aggregates since the 1950s [5], but it wasn’t until 1971 when the term “spheroid” was coined in work using Chinese hamster V79 lung cells as a model of nodular carcinomas, which happened to form perfect spheres [6]. Robert Sutherland’s early research provided some of the first glimpses into not only the effects of nutrition and oxygenation on cell growth, but also allowed for the determination of the growth fraction following treatment with drugs or radiation.
By the 1980s, Mina Bissell and her team at Lawrence Berkeley National Laboratory began pioneering the use of 3D techniques for more accurate in vivo cell models. This shift away from traditional 2D culture systems was first published in a paper highlighting the importance of the extracellular matrix (ECM) along with the crucial role of the microenvironment [7]. These observations were critical for driving the uptake of spheroid culture as a widespread and biologically relevant system with obvious advantages over the widely used monolayer culture methods.
Since then, the field has expanded rapidly to investigate a number of topics from small-scale disease modeling to large-scale, high-throughput screening (HTS) platforms attempting to combat the rising attrition rates seen in existing drug discovery programs.
The ECM: an influential network
Industry has responded to these changes and supported spheroid culture in research through the development of specialized equipment and protocols for culture and maintenance, including plates, synthetic coatings, and cellular scaffolding. There are several common methods used in the generation of spheroids. These include the liquid overlay technique [8], spinner flask [9], gyratory [10], and hanging drop methods [11], or more recently, using suspension culture in individual wells for high-throughput analysis [12]. Following the initial generation of spheroids, the task of maintaining and culturing them can make use of a wide selection of techniques. Depending on the intended application, spheroid culture can involve extracellular matrices or scaffolds, modified surfaces, rotating bioreactors, microcarriers, magnetic levitation, hanging drop plates, or magnetic 3D bioprinting.
Successfully generating and culturing spheroids has a lot to do with the ECM. The ECM is generally composed of soluble proteins and insoluble collagen fibers. While collagen forms the rigid structures that allow tissues to tolerate mechanical stresses like stretching, the proteins within the ECM are involved in a variety of other processes. Proteoglycans, for example, can aid in signaling, binding growth factors, and binding hormones, while multiadhesive matrix proteins like laminin and fibronectin can bind both collagen and other ECM components.
The points at which the ECM makes contact with a cell’s plasma membrane are known as focal adhesions. These vary between tissues but generally consist of integrin molecules that associate with both the intracellular and ECM components—making these ECM components functional units of intracellular signaling.
The ECM is also important when it comes to adhesion not only between cells, but also to the culture vessel. When culturing spheroids, the ECM proteins mediating adhesion will automatically adhere to the surface of a culture vessel. This can interfere with complete spheroid formation and may possibly result in the formation of multiple spheroids or satellite colonies. In an attempt to optimize spheroid formation, manufacturers have developed a number of synthetically modified culture vessel surfaces that specifically inhibit the adsorption of ECM proteins from initiating adhesion between the cell and the culture vessel, thereby prompting cell–cell aggregation and spheroid formation in vitro.
Nunclon Sphera surface is excellent for culturing cancer spheroids
The Thermo Scientific Nunclon Sphera hydrophilic polymer-coated surface has been shown to minimize surface variability. This polymer coating discourages ECM adsorption to the surface, thereby supporting the formation of consistent spheroids (Figure 1).
Extracellular matrix adsorption
Figure 1. Extracellular matrix adsorption. The adsorption of collagen I and fibronectin to the Nunclon Sphera surface is extremely low compared to the standard cell culture–treated surface. Student’s t-test, P < 0.01.
By combining a hydrophilic polymer coating with U-bottom–shaped wells, it is possible to culture spheroids without the production of satellite colonies. HCT 116 human colon carcinoma cells were seeded into Nunclon Sphera 96-well U-bottom plates in complete DMEM. Similarly, cells were seeded into 96-well U-bottom nontreated plates in complete DMEM containing 3% methylcellulose. Using different seeding densities of HCT 116 human colon carcinoma cells, it was shown that single spheroids with well-defined edges can be consistently generated in each individual well (Figure 2).
Advantages of Nunclon Sphera plates over nontreated plates and methylcellulose-containing medium
Figure 2. Advantages of Nunclon Sphera plates over nontreated plates and methylcellulose-containing medium. (A) High and consistent quality of cancer spheroids grown in the Nunclon Sphera plate. (B) Early formation of single cancer spheroids in the Nunclon Sphera 96-well U-bottom plate. (Courtesy of Professor Dolznig from the Institute of Medical Genetics at the Medical University of Vienna.)<
To demonstrate spheroid growth, A549 human adenocarcinoma cells and HCT 116 human colon carcinoma cells were cultured at different densities in Nunclon Sphera plates for 2 weeks. Both cell types displayed adequate spheroid growth as demonstrated by size measurements (Figure 3A). Additionally, the cell health of A549 and HCT 116 spheroids were assessed by Invitrogen PrestoBlue cell viability assay (Figure 3B). Data was normalized against spheroid size for better quantitative comparison—a higher ratio indicates healthier spheroids. Cell viability of cancer spheroids was further confirmed by Invitrogen LIVE/DEAD fluorescence staining assay (Figure 3C). All parameters indicated that cancer spheroids grown on Nunclon Sphera plates were healthy and robust, and that the Nunclon Sphera 96-well U-bottom plate is a reliable and convenient tool for both routine and high-throughput cancer spheroid applications.
Assessments-of-spheroid-growth
Figure 3. Assessments of spheroid growth, cell health, and viability on Nunclon Sphera plates. (A) Growth kinetics of A549 and HCT 116 cancer spheroids on Nunclon Sphera plates were evaluated over period of 13 days. Data represents the mean ± SD of 3 replicates for each cell number. (B) Spheroid cell health assessments on Nunclon Sphera plates were performed using the PrestoBlue cell viability assay with data normalized by spheroid size. (C) Spheroid cell viability was evaluated by LIVE/DEAD staining assay, where live cells are stained green and dead cells are stained red. Scale bar = 1,000 μm.
The hypoxic culture condition
In addition to specialized culture vessels, culturing spheroids requires precisely controlled abiotic conditions such as temperature, humidity, and pH. Gas condition is another vital requirement of cell culture, and typically this has meant mimicking atmospheric oxygen tension supplemented with 5–10% carbon dioxide. Yet, while atmospheric levels of oxygen are approximately 20%, the levels within the human body range from 12% to as low as 1%. In light of this, some researchers have taken to culturing their cells under hypoxic conditions.
The role of oxygen was seen as early as 1972 when Alan Richter and colleagues improved plating efficiency of mouse and rat embryonic tissues by cultivating in 1–3% oxygen [13]. The 21st century is seeing cell culture truly coming of age, taking positions in everything from routine cell culture to cell therapy and the development of personalized medicines. These applications have rekindled an interest in the levels of oxygen used in cell culture, and over the past decade or so, the hypoxic element came to the forefront of spheroid culture.
Cells cultured under hypoxic conditions grow faster, live longer, and show lower stress. A cell culture incubator that controls nitrogen gas, in addition to carbon dioxide, is the best way to achieve hypoxic conditions. So-called tri-gas incubators, such as the Thermo Scientific Heracell VIOS Incubator, optimize low-oxygen cultures to offer optimal growth and culture stability. However, the term “tri-gas” is a misnomer as only carbon dioxide and nitrogen are supplied, thereby reducing the internal oxygen levels to as low as 1%.
Detecting hypoxic conditions in real time is often carried out using a chemical that generates a fluorescent signal under specific conditions. A specialized hypoxia probe, in the form of a fluorogenic compound that is live-cell permeable and begins to fluoresce when oxygen levels fall below 5%, provides robust and reproducible measurements of hypoxia in cells (Figure 4). This reagent is preferable to using pimonidazole adducts that only respond to very low levels of oxygen (at a partial pressure of ≤10 mHg), below levels at which hypoxia may occur, potentially yielding false negative results. The Invitrogen Image-iT Hypoxia Reagent has a greater range of sensitivity and responds quickly to changing levels of oxygen, making it ideal for detecting hypoxic conditions in 3D cultures, spheroids, or neurons, for example [14,15].
Detection of hypoxic conditions
Figure 4. Detection of hypoxic conditions. A549 cells were grown on Thermo Scientific Nunc 35 mm glass-bottom dishes in complete medium at a density of 105 cells/dish. The cells were incubated in Gibco FluoroBrite DMEM with 5 μM Image-iT Hypoxia Reagent (red) at (A) 20%, (B) 5%, (C) 2.5%, and (D) 1% oxygen for one hour on an Invitrogen EVOS Onstage Incubator attached to an EVOS FL Auto Imaging System. The images were taken after one hour of incubation at each oxygen level. The hypoxia signal can be detected at oxygen levels as low as 5%, with increasing signal intensities at 2.5% and 1%.
Spheroids in cancer biology
Spheroid culture methods have made substantial contributions to the advances being made in our basic understanding of cell biology, as well as providing insights into cancer biology. The multicellular tumor spheroid (MCTS) model, using spheroids between 200–500 μm, has lent itself to cancer biology as it more accurately mimics the physiology of tumors, as mentioned earlier. Spheroids in this model develop chemical gradients of oxygen, nutrients, and catabolites just like a tumor in vivo, as well as possess similar histomorphological and functional features [16]. Internally, spheroids possess the same hypoxic core seen in solid tumors (Figure 5) where cells rapidly outgrow the blood supply, leaving the center of the tumor with an extremely low oxygen concentration. Chronically hypoxic regions of tumors are highly resistant to therapy as they are especially difficult to penetrate with chemotherapy [17].
A single HeLa spheroid used in the assessment of hypoxic cores
Figure 5. A single HeLa spheroid used in the assessment of hypoxic cores. HeLa cells were plated at a density of 1,000 cells/well. After two days of culture on Nunclon Sphera 96-well U-bottom plates, HeLa spheroids were stained with Image-iT Hypoxia Reagent (red) and Invitrogen NucBlue Live ReadyProbes Reagent (blue). Images were taken on a confocal microscope.
While the ability of cancer spheroids to replicate key elements of tumors, such as hypoxia, necrosis, angiogenesis, and cell adhesion [20] is intriguing, 3D cell cultures have also been used for studies of viability, clonogenicity, LD50, and metastatic potential under a broad spectrum of conditions. The versatility afforded by the spheroid system has been a game-changer in how we understand and develop treatments for cancer.
This gradient of oxygen in spheroids, progressing from normoxic cells at the periphery to hypoxic cells at the core, provides an excellent model for assessing novel pharmacological agents and drug delivery methods. MCTS models can be used to validate compounds that are activated under hypoxic conditions, thereby targeting the hypoxic core specifically, as well as evaluating drugs and signaling pathways [18,19].
Conclusions
The spheroid system of cell culture has major implications not only for our fundamental understanding of how the interplay between cells, tissues, and the ECM affects pathological states such as cancer, but also for the development of more robust drug screening programs and improved organotypic models.
  • The Nunclon Sphera surface demonstrates extremely low ECM binding properties; it therefore effectively discourages cell attachment and promotes spheroid formation
  • Nunclon Sphera 96-well U-bottom plates support consistent formation and growth of cancer spheroids across commonly used cancer cell lines
  • The evidence for hypoxic cores in cancer spheroids indicates that 3D cancer spheroid culture on Nunclon Sphera plates presents an ideal in vitro system for modeling tumor growth
Methods: Cancer spheroid culture
Cancer cell lines were maintained in Thermo Scientific Nunc Cell Culture Treated EasYFlasks before they were subjected to spheroid culture. To form cancer spheroids, cells were seeded in Nunclon Sphera 96-well U-bottom plates at densities of 100–5,000 cells/well in 200 μL/well of Gibco DMEM with GlutaMAX Supplement and 10% FBS, 1X MEM Non-Essential Amino Acids, 100 U/mL Penicillin- Streptomycin, and 25 mM HEPES. Nontreated plates were similarly seeded in the complete DMEM medium containing 3% methylcellulose. The plates were briefly centrifuged at 250 x g for 5 minutes. The cells were then incubated at 37°C and 5% CO2, and fed every 72 hr by carefully removing 100 μL of medium from each well and replenishing with 100 μL of fresh growth medium using a multichannel pipette. The formation and growth of spheroids were examined using an Invitrogen EVOS imaging system.
References
  1. Fennema E, Rivron N, Rouwkema J, et al. (2013) Spheroid culture as a tool for creating 3D complex tissues. Trends Biotechnol 31(2):108–15. 
  2. Hirschhaeuser F, Menne H, Dittfeld C, et al. (2010) Multicellular tumor spheroids: an underestimated tool is catching up again. J Biotechnol 148(1):3–15. 
  3. Heylman C, Sobrino A, Shirure VS, et al. (2014) A strategy for integrating essential three-dimensional microphysiological systems of human organs for realistic anticancer drug screening. Exp Biol Med 239(9):1240–54. 
  4. Tanner K, Gottesman MM (2015) Beyond 3D culture models of cancer. Sci Transl Med 7(283):283ps9. 
  5. Moscona A (1957) The development in vitro of chimeric aggregates of dissociated embryonic chick and mouse cells. Proc Natl Acad Sci USA 43(1):184–94. 
  6. Sutherland RM, McCredie JA, Inch WR (1971) Growth of multicell spheroids in tissue culture as a model of nodular carcinomas. J Natl Cancer Inst 46(1):113–20. 
  7. Bissell MJ, Hall HG, Parry G (1982) How does the extracellular matrix direct gene expression? J Theor Biol 99(1):31–68. 
  8. Carlsson J, Yuhas JM (1984) Liquid-overlay culture of cellular spheroids. Recent Results Cancer Res 95:1–23. 
  9. Sutherland RM, Durand RE (1984) Growth and cellular characteristics of multicell spheroids. Recent Results Cancer Res 95:24–49. 
  10. Moscona A (1961) Rotation-mediated histogenetic aggregation of dissociated cells: a quantifiable approach to cell interactions in vitro. Exp Cell Res 22:455–75. 
  11. Kelm JM, Timmins NE, Brown CJ, et al. (2003) Method for generation of homogeneous multicellular tumor spheroids applicable to a wide variety of cell types. Biotechnol Bioeng 83(2):173–80. 
  12. Ivascu A, Kubbies M (2006) Rapid generation of single-tumor spheroids for high-throughput cell function and toxicity analysis. J Biomol Screen 11(8):922–32. 
  13. Richter A, Sanford KK, Evans VJ (1972) Influence of oxygen and culture media on plating efficiency of some mammalian tissue cells. J Natl Cancer Inst 49(6):1705–12. 
  14. Mandavilli BS, Chen A, Robinson V, et al. (2015) Intracellular detection of hypoxia in live cells. Presentation at American Association for Cancer Research (AACR), Poster 3007.
  15. Zhang S, Hosaka M, Yoshihara T, et al. (2010) Phosphorescent light-emitting iridium complexes serve as a hypoxia-sensing probe for tumor imaging in living animals. Cancer Res 70(11):4490–8. 
  16. Thoma CR, Zimmerman M, Agarkova I, et al. (2014) 3D cell culture systems modeling tumor growth determinants in cancer target discovery. Adv Drug Del Rev 69–70:29–41. 
  17. Das V, Bruzzese F, Konecny P, et al. (2015) Pathophysiologically relevant in vitro tumor models for drug screening. Drug Disc Today 20(7):848–55. 
  18. Dufau I, Frongia C, Sicard F, et al. (2012) Multicellular tumor spheroid model to evaluate spatio-temporal dynamics effect of chemotherapeutics: application to the gemcitabine/CHK1 inhibitor combination in pancreatic cancer. BMC Cancer 12:15. 
  19. Mead S, Foster R (2015) A multicellular tumor model: Applications for evaluating drugs and signaling pathways in a 3D system. Application note available from http://www.horizondiscovery.com/ 
  20. Benien P, Swami A (2014) 3D tumor models: history, advances and future perspectives. Future Oncol 10(7):1311–27. 
Related links
Share:  
Resources
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Media Formulation Tool
Find the right Gibco media formulation for DMEM, DMEM/F-12, MEM, and RPMI-1640 media.
Related products
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products
For Research Use Only. Not for use in diagnostic procedures.