This resource is dedicated to guiding researchers through the complex process of differentiating pluripotent stem cells (PSCs) into neural organoids, which serve as sophisticated models for studying brain development, neurological diseases, and potential therapeutic applications. Find detailed protocols, recommendations, and practical tips to optimize your differentiation procedures and transfection techniques. Enhance your understanding and capabilities in creating and utilizing neural organoids for advanced research in neuroscience and regenerative medicine.
Pluripotent stem cells
PSCs are a type of stem cell capable of differentiating into nearly all cell types of the body, including cells from the three primary germ layers: ectoderm, mesoderm, and endoderm. This ability allows PSCs to give rise to a wide variety of specialized cells, making them incredibly valuable for scientific research and potential therapeutic applications. PSCs are potentially useful in treating diseases because they can be used to generate healthy cells and tissues to replace damaged or diseased ones. This opens up possibilities for regenerative medicine, where PSCs could be used to treat conditions such as spinal cord injuries, Parkinson's disease, diabetes, and heart disease, by restoring normal function through the regeneration of healthy tissue. Additionally, PSCs provide a powerful platform for drug testing and disease modeling, allowing researchers to study disease mechanisms and screen for new drugs in a controlled laboratory environment.
Neural organoids
Neural organoids are three-dimensional, miniaturized, and simplified versions of brain tissue derived from pluripotent stem cells. These structures mimic the complex architecture and functionality of the human brain, providing a valuable model for studying brain development, neurological diseases, and potential treatments. Neural cell differentiation refers to the process by which pluripotent stem cells or neural progenitor cells develop into specialized neural cells, such as neurons, astrocytes, and oligodendrocytes. This differentiation is guided by specific signals and growth factors that mimic the natural developmental cues within the body. By using neural organoids, researchers can observe and manipulate neural cell differentiation in a controlled environment, gaining insights into the mechanisms of brain development and the pathology of neurodegenerative diseases and exploring new therapeutic approaches.
Workflow for the creation of neural organoids and spheroids
Recent advances in cell culture techniques have focused on creating 3-dimensional (3D) systems in an attempt to represent in vivo cell–cell relationships and microenvironments in vitro. Various tissue engineering technologies such as bioprinting, microfluidics, and organs-on-chips have been used successfully to generate 3D cultures [1,2]. Remarkable progress has also been made utilizing adult and pluripotent stem cells (ASCs and PSCs) to generate 3D organ-like (i.e., organoid) cell models [3–5]. PSC-based methods frequently start by aggregating cells in suspension culture to form clusters called embryoid bodies (EBs). Cells in these clusters are capable of differentiating into many types and can undergo self-organization and self-morphogenesis to create a complex cell model that better mimics the in vivo cell–cell interactions and microanatomy of a given tissue type. Some PSC-based approaches also require the encapsulation of cells within a natural or synthetic extracellular matrix (ECM)-like substrate [6–8]. In all methods, the application of growth factors, small molecules, and other media supplements is used to guide the formation of organoid systems based on principles inferred from studies of embryogenesis and adult stem cell biology. There are now many published methods for generating a variety of organoid types that resemble different parts of the brain, as well as the liver, intestine, and kidney, to name a few.
The unknown compatibility of multiple reagents from different vendors that span the organoid workflow is an issue that many researchers experience. This issue can have dramatic consequences for the successful generation of the desired organoid system and its reproducibility between laboratories. Established workflows for generating neural organoids from PSCs typically follow a specific sequence of steps that begin with standard PSC culture followed by EB formation, neural induction, neural patterning, and organoid growth [7, 9–12] (Figure 1). The composition of the cell culture medium at each of these steps is critical for the successful differentiation of PSCs. Importantly, the differentiation capacity of a given PSC line must be determined empirically, and some optimization of the differentiation method may be needed for the PSC line of choice. In this application note, we demonstrate the use of feeder-free Gibco StemFlex Medium, Gibco Geltrex matrix, and Thermo Scientific Nunclon Sphera Microplates to create neural organoids and spheroids.
Diagram listing the 5 basic steps for organoid formation
Figure 1. The essential steps of neural organoid formation from PSC cultures.
Experimental details and results
PSC culture
Prior to differentiation, H9 human embryonic stem cells (ESCs) and Gibco Human Episomal Induced Pluripotent Stem Cells (iPSCs, Cat. No. A18945) were maintained using StemFlex Medium and grown on Thermo Scientific Nunclon Delta tissue cultureware coated with a 1:100 dilution of Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1413301). PSC clumps were routinely passaged using Gibco Versene Solution (Cat. No. 15040066).
EB formation
PSCs were cultured in feeder-free conditions using StemFlex Medium (Cat. No. A3349401). When PSC cultures reached 70–80% confluency, they were dissociated into single-cell suspensions using Gibco StemPro Accutase Cell Dissociation Reagent (Cat. No. A1110501), Trypsin/EDTA Solution (Cat. No. R001100), or TrypLE Select Enzyme (Cat. No. 12563011). Cell counts and viability were determined using Gibco Trypan Blue Solution (Cat. No. 15250061) and the Invitrogen Countess II FL Automated Cell Counter (Cat. No. AMQAF1000). About 6–9 x 103 viable cells per well were seeded in StemFlex Medium with Gibco RevitaCell Supplement (Cat. No. A2644501) in Nunclon Sphera 96-well U-bottom microplates (Cat. No. 174925). Nunclon Sphera microplates exhibit virtually no cell attachment, promoting consistent formation of spheroids. EBs formed overnight equally well with all dissociation methods but most efficiently with the addition of RevitaCell Supplement (Figure 2). In the absence of RevitaCell Supplement, small EBs did form but with poor efficiency, as most cells either did not survive or did not self-aggregate (Figure 2). EBs were then cultivated for 3–4 days, with a 75% medium change every other day with StemFlex Medium with RevitaCell Supplement. The resulting EBs were of consistent size that was directly proportional to the number of cells seeded (Figure 3).
microscopic images of embryoid bodies resulting from different dissociation methods and culture supplementation
Figure 2. RevitaCell Supplement dramatically improves EB formation. A comparison of EB formation after isolation of PSCs by different methods demonstrated that EBs formed equally well with each dissociation reagent but only if RevitaCell Supplement was included in the culture medium. Cells that do not contribute to the EB are eventually washed away during media changes and do not typically interfere with subsequent steps; here they were not washed away, to illustrate the efficiency of EB formation.
microscopic images of embryoid bodies (EB) and a graph of EB volume based on initial cell seeding density
Figure 3. Evaluation of EBs formed in StemFlex Medium with RevitaCell Supplement. (A, B) These images show representative EBs from two different PSC lines after 4 days of culture. (C) EB size is directly proportional to the number of cells seeded. The graph shows the consistency in size between 8 replicates for each cell density that was evaluated. Data were calculated by measuring the area of each EB using ImageJ software. The area was then used to calculate the approximate EB volume, which is plotted on the y-axis.
Neural induction and patterning
Following EB formation, the cell aggregates were induced to differentiate into neural lineages by performing 3–4 successive 75% volume medium changes to serially dilute and remove the prior culture medium. Neural induction medium was composed of Gibco DMEM/F-12 with GlutaMAX Supplement (Cat. No. 10565018) and N-2 Supplement (Cat. No. 17502001). EBs were cultured for 8–9 days with a 75% volume medium change every other day until the outer layers of the EB formed a bright “ring” in contrast to the darker center (Figure 4). By day 10, each EB that displayed this phenotype was encapsulated in undiluted Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1413201) and incubated at 37°C to gel. The use of Geltrex matrix for this application has been independently demonstrated elsewhere [13]. Droplets of Geltrex matrix containing EBs were then transferred to a differentiation medium consisting of DMEM/F-12 with GlutaMAX Supplement (Cat. No. 10565018) and Gibco Neurobasal Medium (Cat. No. 21103049) with GlutaMAX Supplement (Cat. No. 35050061), MEM with NEAA (Cat. No. 10370021), 2-mercaptoethanol (Cat. No. 21985023), insulin (Cat. No. 12585014), N-2 Supplement (Cat. No. 17502001), and B-27 Supplement Minus Vitamin A (Cat. No. 12587010). Encapsulated samples were then transferred to Nunclon Sphera 6-well or 24-well plates (Cat. No. 174932, 174930) with a density of 3–5 or 1–2 droplets per well, respectively.
Growth and maturation
The samples were cultured in a growth and maturation medium of the same formulation as the previous incubation medium except this medium contained B-27 Supplement (Cat. No. 17504044). From this point on, neural organoids were cultured on an orbital shaker at 80–85 rpm and the medium was changed every 2–3 days. Neuroepithelia become easily visible within about a week. These samples can be continuously cultured for many weeks (Figure 5A, B) or until analysis is performed (e.g., cellular organization, marker expression). For example, Figure 5C indicates the presence of multiple neural cell types present at day 39 of culture. Gene expression analysis shows that these organoids still contain neural stem and progenitor cells, based on SOX1, SOX2, and PAX6 expression, as well as immature neuronal markers such as DCX and MAP2. However, markers of specific neural regions such as TBR1 (deep layer neurons), FOXG1 (forebrain tissue), and SLC6A1 (encodes GABA1 transporter expressed in cerebral cortical tissue, hippocampus, and other tissues) were also detected, indicating the presence of more differentiated cell types.
microscopic images showing day 7 and day 10 embryoid bodies
Figure 4. Neural induction and patterning.(A) Brightfield image showing a day 7 EB. (B) Day 10 neuralized EB immediately before encapsulation in Geltrex matrix.
microscopic images of neural organoids and graphs of gene expression analysis results for 9 neural cell markers
Figure 5. Phenotypic characterization and gene expression analysis of neural organoids.(A, B) Brightfield images of neural organoids on day 31 or day 24 of culture show convoluted neuroepithelial structures. (C) Gene expression analyses of day 39 neural organoid cultures indicate the presence of multiple neural cell types, including neural stem cells and neurons. Expression levels were calculated using the 2–ΔΔCt method, relative to undifferentiated H9 human ESCs or Gibco Human Episomal iPSCs. Samples from two experiments are shown. (D) Summary of Applied Biosystems TaqMan® Assays used for gene expression analysis.
Neural organoids differentiation
Together, these data demonstrate the compatibility of feeder-free StemFlex Medium and Nunclon Sphera 96-well U-bottom microplates with EB formation and neural organoid differentiation. Furthermore, we demonstrate the effectiveness of Geltrex matrix for the encapsulation and morphogenesis of neural organoids. In all, the results indicate that these three products can be successfully integrated with existing Gibco basal media and supplements that are commonly used for studies involving neural organoids.
Ordering information
Catalog # Name Size Price (USD) Qty
A1413201 Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix, 1 mL Each
82.50
17504044 B-27™ Supplement (50X), serum free, 10 mL Each
141.65
Online Exclusive
152.00
Save 10.35 (7%)
12587010 B-27™ Supplement (50X), minus vitamin A, 10 mL Each
170.65
Online Exclusive
184.00
Save 13.35 (7%)
10565018 DMEM/F-12, GlutaMAX™ supplement, 500 mL Each
65.65
Online Exclusive
71.75
Save 6.10 (9%)
17502001 N-2 Supplement (100X), 50 mL Each
1,264.00
21103049 Neurobasal™ Medium Each
110.00
A3349401 StemFlex™ Medium Each
432.00
A2644501 RevitaCell™ Supplement (100X) Each
247.00
174925 96 Well Round (U) Bottom Plate, Sphera Low-Attachment Surface, Pack of 1 Case of 8
206.65
Online Exclusive
224.00
Save 17.35 (8%)
174930 24 Well Plate, Sphera Low-Attachment Surface, Pack of 1 Case of 7
164.65
Online Exclusive
177.00
Save 12.35 (7%)
174932 6 Well Plate, Sphera Low-Attachment Surface, Pack of 1 Case of 7
169.65
Online Exclusive
181.00
Save 11.35 (6%)
References
  1. Bhatia SN, Ingber DE (2014) Microfluidic organs-on-chips. Nat Biotechnol 32:760–72. 
  2. Murphy SV, Atala A (2014) 3D bioprinting of tissues and organs. Nat Biotechnol 32:773–785. 
  3. Bartfeld S, Clevers H (2017) Stem cell–derived organoids and their application for medical research and patient treatment. J Mol Med (Berl) 95:729–738. 
  4. Clevers H (2016) Modeling development and disease with organoids. Cell 165: 1586–1597. 
  5. Drost J, Clevers H (2018) Organoids in cancer research. Nat Rev Cancer 18:407–418. 
  6. Sato T, Vries RG, Snippert HJ, et al. (2009) Single Lgr5 stem cells build cryptvillus structures in vitro without a mesenchymal niche. Nature 459:262–265. 
  7. Lancaster MA, Renner M, Martin CA, et al. (2013) Cerebral organoids model human brain development and microcephaly. Nature 501:373–379. 
  8. Gjorevski N, Sachs N, Manfrin A, et al. (2016) Designer matrices for intestinal stem cell and organoid culture. Nature 539:560–564. 
  9. Mariani J, Simonini MV, Palejev D, et al. (2012) Modeling human cortical development in vitro using induced pluripotent stem cells. Proc Natl Acad Sci USA 109:12770–12775. 
  10. Phillips AW, Nestor JE, Nestor MW (2017) Developing hiPSC derived serum free embryoid bodies for the interrogation of 3-D stem cell cultures using physiologically relevant assays. J Vis Exp 125:e55799. 
  11. Sloan SA, Darmanis S, Huber N, et al. (2017) Human astrocyte maturation captured in 3D cerebral cortical spheroids derived from pluripotent stem cells. Neuron 95:779–790.e6. 
  12. Thomas CA, Tejwani L, Trujillo CA, et al. (2017) Modeling of TREX1-dependent autoimmune disease using human stem cells highlights L1 accumulation as a source of neuroinflammation. Cell Stem Cell 21:319–331.e8. 
  13. Iefremova V, Manikakis G, Krefft O, et al. (2017) An organoid-based model of cortical development identifies non-cell-autonomous defects in Wnt signaling contributing to Miller-Dieker syndrome. Cell Rep 19:50–59. 
Resources
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Media Formulation Tool
Find the right Gibco media formulation for DMEM, DMEM/F-12, MEM, and RPMI-1640 media.
Related products
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products
For Research Use Only. Not for use in diagnostic procedures.