Front angle on the Automated Thermal Cycler (ATC)
Looking to automate your PCR workflow? The Applied Biosystems Automated Thermal Cycler (ATC) offers the flexibility, reliability, and performance needed in a complete PCR automation system.
Request quote Request demo Virtual tour
What is automated PCR?
Automated PCR refers to the use of automated systems and instruments to perform the polymerase chain reaction (PCR) technique, which is a widely used molecular biology method for amplifying DNA samples that helps in reducing manual errors and cross-contamination while providing accurate, reproducible results by saving manual hands-on time.
PCR automation system designed for easy robotic integration
  • Flexible modular design saves space on deck
  • Easy-to-install software solutions for robotic integration: SiLA Rapid Integration™ standard-compatible
  • Software and prewritten application programming interfaces (APIs)
  • Free software for stand-alone operation allows assay optimization before integration
Optimized features for automated PCR workflows
Video Player is loading.
Current Time 0:00
Duration 3:36
Loaded: 4.54%
Stream Type LIVE
Remaining Time 3:36
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    • Automated lid for hands-free operation with a liquid handler or plate stacker
    • Heated cover slides forward to cover the plate nest, enabling thermal contact and minimizing evaporation
    • Automated plate ejectors help ensure consistent plate positioning for half-skirted or fully skirted 96-well plates
    See ATC-compatible robotics
    We are currently working with robotics/liquid handling system manufacturers on the development of drivers compatible with ATC. Please contact our Technical Support for any questions regarding driver development and robotic integration.
    ManufacturerModelManufacturer contact information
    Beckman Coulter Inc.
    • Biomek i7 Automated Workstation
    • Biomek i5 Automated Workstation
    • Biomek FXP Automated Workstation
    • Biomek NXP Automated Workstation
    • Biomek 4000 Automated Workstation
    lshotline@beckman.com
    Hamilton Robotics
    • VANTAGE Liquid Handling System
    • Microlab STAR
    • Microlab NIMBUS
    marketingrequests@hamiltoncompany.com
    Hudson Robotics Inc.
    • SOLO™ Pipettor
    • PlateCrane EX™ robot arm
    • PlateCrane SciClops™ robot arm
    http://hudsonrobotics.com/contact-us/request-service/
    Tecan
    • Fluent™
    • Freedom Evo™ Series
    https://lifesciences.tecan.com/support/helpdesk
    Thermo Fisher Scientific
    • Thermo Scientific Orbitor
    • Thermo Scientific Spinnaker
    • Thermo Scientific F7
    • Thermo Scientific Momentum
    customercare@thermofisher.com
    • If your robot is listed here, contact us or the robot manufacturer for driver and integration information.
    • If you don't see your system listed here, we can work with you and your robot manufacturer on integration options.
    Stay in control with Thermal Cycler Fleet Control Software
    Diagram of multiple thermal cyclers connected to one tablet to represent Fleet Control Software
    Manage multiple users, methods, and instruments with a single intuitive interface securely behind your firewall.
    Learn more about Thermal Cycler Fleet Control Software
    Ordering information
    High-performance PCR and qPCR plastics for optimal results
    Five multi-colored PCR tube plates
    Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for end-point PCR applications. Quickly find the right fit and compatible plastics for your desired model and block formats by using our plastics selection guide.
    Explore online PCR and qPCR plastics selection tool
    Resources
    Thermal Cycler Software and Firmware Files
    Download the latest updates for your instrument
    Interested in private label thermal cyclers?
    Submit an inquiry
    Support
    Service
    We provide a full range of service options for all Applied Biosystems thermal cyclers.
    View service plans
    Multiplex Dynamic Temperature Verification Service
    Instrument qualification service: Have one of our engineers perform temperature qualifications at your site.
    View temperature service options
     Download MDTV flyer
    For Research Use Only. Not for use in diagnostic procedures.