bf5a- Apps/Techniques/Products/Resources/Contact Us
Nanotechnology is quickly redefining how we live. In the textile industry, they’re leading to intelligent, adaptable clothing that better repels stains, odors, and water. Consumer electronics, such as smartphones and other devices we use on a daily basis, are powered by transistors that are becoming ever smaller, faster, and better thanks to nanotechnology. In medicine, nanoparticles are being developed to deliver next-generation drugs and vaccines. It even has applications in the energy sector, where it is improving the efficiency of solar and wind power.
The versatility of nanoparticles arises from the fact that atoms and molecules have fundamentally different properties at the nanoscale than when they are combined to form larger particles or bulk materials. Some are better at conducting electricity and heat. Some are stronger, have different magnetic properties, or better reflect light. Materials at the nanoscale also have far greater surface areas than bulk materials, allowing nanoparticles to act as chemical catalysts in industrial applications or in automotive catalytic converters.
Quantifying the composition, sizes and shapes of nanoparticles is a crucial first step in furthering our understanding of their unique properties. Using intuitive scanning or transmission electron microscopes (S/TEM) in combination with energy dispersive x-ray spectroscopy (EDX), researchers around the globe are obtaining nanometer-, and even sub-nanometer-, scale images and chemical data, giving important insights into the arrangement and potential functions of nanoparticles.
The need for large area correlative imaging at high resolution has recently increased, as it allows researchers to preserve the context of their observations while also providing statistically robust data. Thermo Scientific Maps Software enables automated, unattended acquisition of images across a sample that are then stitched together to create one large final image. Additionally, Thermo Scientific Avizo Software allows for on-the-fly processing of statistics such as size, surface area, perimeter, distribution, and chemical composition of nanoparticles. Images and chemical information from different microscopes can be correlated to retain relevant context.
Resources
Applications
Process control using electron microscopy
Process control using electron microscopy
Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
Quality control and failure analysis using electron microscopy
Quality control and failure analysis
Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
Fundamental Materials Research_R&D_Thumb_274x180_144DPI
Fundamental Materials Research
Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
Style Sheet for Komodo Tabs
Techniques
Atomic-Scale Elemental Mapping with EDS
Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
Learn more ›
EDS Elemental Analysis
Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
Learn more ›
Particle analysis
Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
Learn more ›
Automated Particle Workflow
The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
Learn more ›
Atomic-Scale Elemental Mapping with EDS
Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
Learn more ›
EDS Elemental Analysis
Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
Learn more ›
Particle analysis
Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
Learn more ›
Automated Particle Workflow
The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
Learn more ›
Products
Style Sheet for Instrument Cards Original
STEM Sample Holder
  • Delivers high contrast under low voltage
  • Accommodates a range of materials
  • Includes BF, DF, and HAADF imaging modes
Talos F200i TEM
  • Compact design with X-TWIN objective lens
  • Available with S-FEG, X-FEG, and X-CFEG
  • Flexible and fast EDS options for comprehensive elemental analysis
Talos F200S TEM
  • Intuitive and easy-to-use automation software
  • Available with Super-X EDS for rapid quantitative chemical analysis
  • High-throughput with simultaneous multi-signal acquisition
Talos F200X TEM
  • High-resolution, EDS cleanliness, and quality in 2D as well as 3D
  • X-FEG and X-CFEG available for the highest brightness and energy resolution
  • High accuracy and repeatable results with integrated Thermo Scientific Velox Software
Thermo Scientific Talos F200C transmission electron microscope (TEM)
Talos F200C TEM
  • High-contrast and high-quality TEM and STEM imaging
  • 4k x 4k Ceta CMOS camera options for large FOV and high read-out speeds
  • Large pole piece gap and multiple in situ options
Thermo Scientific Talos L120C transmission electron microscope (TEM)
Talos L120C TEM
  • High versatility and stability
  • 4k x 4k Ceta CMOS camera for speed and large FOV
  • TEM magnification range of 25X to 650kX
  • EDS and STEM options for compositional analyses
K-Alpha XPS
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
Nexsa G2 XPS
  • Micro-focus X-ray sources
  • Unique multi-technique options
  • Dual-mode ion source for monoatomic & cluster ion depth profiling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Athena Software
Imaging Data Management
  • Ensure traceability of images, data, metadata and experimental workflows
  • Simplify your imaging workflow​
  • Improve collaboration
  • Secure and manage data access​
Thermo Scientific Maps electron microscopy software
Maps Software
  • Acquire high-resolution images over large areas
  • Easily find regions of interest
  • Automate image acquisition process
  • Correlate data from different sources
Avizo Software
Materials Science
  • Support for multi-data/multi-view, multi-channel, time series, very large data
  • Advanced multi-mode 2D/3D automatic registration
  • Artifact reduction algorithms
Elemental Mapping
  • Fast and reliable information on the distribution of elements within the sample or the selected line
  • Easily exported and reported results
ParticleMetric
  • Integrated software in ProSuite for online and offline analysis
  • Correlating particle features such as diameter, circularity, aspect ratio and convexity
  • Creating image datasets with Automated Image Mapping
Velox
  • An experiments panel on the left side of the processing window.
  • Live quantitative mapping
  • Interactive detector layout interface for reproducible experiment control and setup
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.