Gibco StemScale PSC Suspension Medium
A new dimension to PSC culture
The Gibco StemScale™ PSC Suspension Medium is a scalable, easy-to-use medium that supports large-scale generation of human pluripotent stem cells (PSCs) in suspension culture and formulated to address the current technical challenges in aggregating PSC spheroid culture systems.
  • Maximal expansion capability—achieves 5- to 10-fold expansion per passage and 3x the expansion capability compared to other commercially available PSC suspension media systems
  • Simplified workflow—offers easy media changes without the use of microcarriers or cell strainers during culture or passaging
  • Scalable across formats—supports scale-up of PSC cultures across multiple vessel formats: multiwell plates, shake flasks, and bioreactors
  • Consistency across PSC lines—delivers reliable and consistent spheroid formation while maintaining pluripotency and cell viability
Order now  See the data  Customer stories
CTS StemScale medium product photo
Simplify the transition from bench to clinical applications
Gibco CTS (Cell Therapy Systems) StemScale PSC Suspension Medium has a similar formulation to StemScale PSC Suspension Medium with specific modifications that follow regulatory guidance. The xeno-free formulation is intended for translational and clinical applications and enables comparable performance, including pluripotency, genomic stability, and cell yield.
See the data
Customer stories
A robust, scalable solution for PSC expansion and downstream neural differentiation
Dr. Cláudia Miranda shares how StemScale PSC Suspension Medium is enabling her research as a scalable solution for high-fold expansion of human pluripotent stem cells and downstream neural differentiation from PSC spheroids in suspension culture.
“StemScale is robust and allows us to replicate the results despite the cell line that we use.”
Generating cardiac organoids from PSC spheroids
Rebecca Sereda talks about her cardiomyocyte research at Harvard University and using StemScale PSC Suspension Medium to create human PSC cardiac organoids at high efficiency in a 3D model system.
“StemScale has provided us a simple and easy way to produce the cells we need to do the science we want.”
Superior expansion capability
Gibco StemScale PSC Suspension Medium supports the growth of pluripotent stem cells in suspension through the self-assembly of spheroids and is ideal for applications where large numbers of high-quality PSCs are required.
StemScale medium delivers 5- to 10-fold expansion per passage and 3x the expansion capability of other media (Figure 1).
Scaling expansion using StemScale Medium
Figure 1. Scaling expansion using StemScale medium. After 30 passages and 20 weeks in culture, StemScale medium delivers greater than 10x cell expansion per passage (left plot). When compared against a medium from supplier medium “M”, StemScale medium generates up to 3x expansion across the same period (right plot).
StemScale medium helps to maintain cell viability and pluripotency
Figure 2. StemScale medium helps to maintain cell viability and pluripotency. StemScale medium delivers superior cell viability after 3 passages as compared to another suspension culture system and maintains high pluripotency after 30 passages.
Generate more cells in less time
In comparison to adherent culture, growing PSCs as self-aggregating spheroids in suspension using StemScale Medium allows users to culture greater numbers of cells in a shorter amount of time (Table 1) while reducing the amounts of media and plasticware consumed.
Here we show differences in time and cell number between cells grown in adherent culture and suspension culture starting from a single well of a 6-well plate.
 Suspension scale-upAdherent scale-up
Total days in cultureCulture vessel formatAverage cell yield*Medium consumedCulture vessel formatAverage cell yield*Medium consumed
3 days6-well plate (single well)1.5 x 106 cells6 mL6-well plate (single well)1.5 x 106 cells6 mL
7 days6-well plate (full plate)18 x 106 cells30 mL100 mm dish9 x 106 cells48 mL
11 days100 mL bioreactor150 x 106 cells200 mL150 mm dish22 x 106 cells72 mL
15 days500 mL bioreactor750 x 106 cells1,000 mLNunc TripleFlask Treated Cell Culture Flask78 x 106 cells800 mL
19 days3 L bioreactor4.5 x 109 cells6,000 mL2-layer Nunc Cell Factory System300 x 106 cells1,200 mL
23 days10-layer Nunc Cell Factory System1.5 x 109 cells6,000 mL
27 days30-layer Nunc Cell Factory System4.5 x 109 cells18,000 mL
* Values are estimated from data based on 4 days of expansion per culture vessel format. Values for the 3 L bioreactor and 30-layer Thermo Scientific Nunc Cell Factory System have been extrapolated from the smaller bioreactor and Nunc Cell Factory System formats listed in the table.
Table 1. Scale-up for suspension vs adherent culture systems. Scale-up in suspension culture systems using StemScale PSC Suspension Medium vs. scale-up in adherent culture systems.
Pluripotent stem cells grown in StemScale PSC Suspension medium
Figure 3. Pluripotent stem cells grown in StemScale medium. Pluripotent stem cells seeded in StemScale PSC Suspension medium maintain optimal spheroid size and viability over 5 days.
“StemScale medium improves upon the efficiency of stem cell aggregation formation.”
– S. Rieck, ViaCyte
Simplified workflow
Simplified workflow for pluripotent stem cell culture maintenance using StemScale Medium
Figure 4. Simplified workflow (schematic). Easily adopt adherent cultures to suspension cultures using StemScale medium. StemScale medium enables users to skip feeding days, if desired. After initiation of cultures in StemScale medium, Gibco Episomal iPSCs are fed periodically using a 50% medium exchange, every day or every other day.
Chart showing similar fold PSC expansion in StemScale medium with every day and every other day feeding.
Chart shows high percent pluripotency marker expression with flexible feeding in StemScale medium
Figure 5. Cultures fed every other day perform similarly to cultures fed daily in fold change (A) and pluripotency (B).
How-to videos: 3 steps to culturing PSCs in suspension
Watch instructional videos to learn how to culture human pluripotent stem cells in suspension using StemScale PSC Suspension Medium.
Step 1: Initiation of PSC spheroid cultures in StemScale medium
Video Player is loading.
Current Time 0:00
Duration 4:02
Loaded: 4.06%
Stream Type LIVE
Remaining Time 4:02
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Step 2: Maintenance and feeding of PSC spheroids with 50% media change
    Video Player is loading.
    Current Time 0:00
    Duration 2:58
    Loaded: 5.51%
    Stream Type LIVE
    Remaining Time 2:58
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Step 3: Passaging PSC spheroids in suspension culture
      Video Player is loading.
      Current Time 0:00
      Duration 4:19
      Loaded: 3.79%
      Stream Type LIVE
      Remaining Time 4:19
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Step 1: Initiation of PSC spheroid cultures in StemScale medium
        Video Player is loading.
        Current Time 0:00
        Duration 4:02
        Loaded: 4.06%
        Stream Type LIVE
        Remaining Time 4:02
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Step 2: Maintenance and feeding of PSC spheroids with 50% media change
          Video Player is loading.
          Current Time 0:00
          Duration 2:58
          Loaded: 5.51%
          Stream Type LIVE
          Remaining Time 2:58
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            Step 3: Passaging PSC spheroids in suspension culture
            Video Player is loading.
            Current Time 0:00
            Duration 4:19
            Loaded: 3.79%
            Stream Type LIVE
            Remaining Time 4:19
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
              Expansion across multiple cell lines and vessel formats
              Across four different PSC lines cultured in StemScale Medium, 6- to 11-fold expansion was achieved per passage and 18- to 28-fold expansion was achieved over 15 days.
              Scale-up in suspension culture systems using StemScale PSC Suspension Medium vs. scale-up in adherent culture systems
              Figure 6. Suspension vs adherent culture systems. Scale-up in suspension culture systems using StemScale PSC Suspension Medium vs. scale-up in adherent culture systems.
              Neural and cardiac differentiation from PSC spheroids
              Neural induction is initiated directly from PSC spheroids cultured in StemScale PSC Suspension Medium
              Neural induction of spheroids
              Figure 7. Neural induction of spheroids. Using the PSC Neural Induction Medium (Cat. No. A1647801), large numbers of maturing neural stem cells are initiated directly from PSC spheroids cultured in StemScale medium.
              StemScale cultured spheroids are initiated into DA neuron differentiation workflow
              Dopaminergic neurons are directly initated from PSC spheroids cultured in StemScale medium
              Figure 8. Dopaminergic neurons are directly initiated from PSC spheroids cultured in StemScale medium. In just 25 days, using the PSC Dopaminergic Neuron (DA) Differentiation kit (Cat. No. A3147701) DA neurons differentiated in StemScale medium co-expressed the same amount of FOXA2/Otx2 in suspension culture when compared to a 35-day protocol in adherent culture (data not shown).
              Cardiac organoids differentiated from PSC spheroids
              Cardiomycytes differentiated in suspension from PSC spheroids grown in StemScale medium
              Figure 9. Cardiomyocytes differentiated in suspension from PSC spheroids grown in StemScale medium. (A) Undifferentiated hPSC spheres growing in StemScale medium prior to cardiac induction. (B) Spheroids in rotation culture 12 days after start of cardiac induction. (C) Cardiac organoids stained for the cardiac markers Alpha Sarcomeric Actinin (green) and Cardiac Troponin T (red). Nuclei are counterstained with DAPI (blue).
              Ordering information
              StemScale Medium resources
              Find technical and educational resources needed to support your PSC Suspension culture using StemScale PSC Suspension Medium.
              Want more information? Contact our Technical Support Specialists for more technical and application support of StemScale PSC Suspension Medium
              Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.