Essential 8 Media System
Just what stem cells need.
Nothing they don’t.
Starting with the original formulation, Gibco Essential 8 Medium is our most defined and consistent feeder-free medium for induced pluripotent stem cell culture. It has been optimized for a wide variety of applications to help researchers achieve ultimate control over their PSC culture systems.
Need help?Download brochure
Essential 8 Resources
Gibco PSC Media are manufactured according to cGMP for medical devices, 21 CFR Part 820
Where do you need more control over your PSC Culture?
timer
Routine PSC maintenance and expansion
stress
E8 media
Transition to clinical research
Ordering Information
Where would you like to gain control? Compare Essential 8 for different applications.
PSC maintenance and expansionEssential 8 Medium
  • Consistent—no BSA reducing variability compared to existing feeder-free culture media
  • Cost effective—economical and scalable PSC culture compared to other feeder-free media
  • Robust—reliable and consistent cultures with a xeno-free, cGMP defined medium
Flexible feeding scheduleEssential 8 Flex Medium
  • Eliminate daily feeding—Stabilized FGF2 enables maintenance of pluripotency over a 2-day period without feeding cells and without adjustments to your typical culture protocol
  • Easy to transition—Same set-up, applications, and protocol as the original Essential 8 formulations
  • Proven—Based on the original Essential 8 formulation, maintains pluripotency and normal karyotypes in long-term culture
DifferentiationEssential 6 Medium
  • Enables differentiation—does not contain bFGF, thereby enabling embryoid body formation and directed differentiation
  • Flexibility—provides a flexible growth factor-free base where TGFβ, bFGF, or other proteins, molecules and supplements can be added to suit different applications
Stressful events
Essential 8 Medium + RevitaCell Supplement
  • Minimize the impact of stress—RevitaCell Supplement supports cells in stressful applications through a more specific ROCK inhibitor
  • Non-addictive—Shown to have a non-addictive impact on downstream cell utility unlike other ROCK inhibitors
  • Expanded applications—Enabling use of Essential 8 medium in applications where cells need additional support
Adaptation from feeder-based mediaEssential 8
Adaptation Kit
  • Confidently transition—adapt cells from feeder-dependent to feeder-free culture system with minimal effort and maximum cell viability
  • Physiologically relevant—expand cells with minimal differentiation
  • Scalable—passage stem cell lines as single cells with no impact to genetic stability
Transition from bench to clinical research
CTS Essential 8 Medium
  • Reduces risk—only globally available human pluripotent stem cell (hPSC) culture medium without components derived directly from animals
  • GMP-grade—cGMP manufactured in an FDA registered facility for medical devices
  • Testing and regulatory documentation—FDA Drug Master Files available
Minimize the impact of stress – RevitaCell Supplement supports cells in stressful applications through a more specific ROCK inhibitor
Non-addictive – Shown to have a non-addictive impact on downstream cell utility unlike other ROCK inhibitors
Expanded applications – Enabling use of Essential 8 medium in applications where cells need additional support
Related products
The Essential 8 Media system and accompanying reagents provide the most defined and consistent culture conditions for pluripotent stem cells. Each of the recommended reagents across the following workflow is defined and  xeno-free, allowing for consistent results every time. Below is an example of a workflow advancing from reprogramming to differentiation utilizing the Essential 8 Media system.
Essential 8 Media System
essential 6
CytoTune-iPS 2.0  Sendai Reprogramming Kit  
contains a polycistronic vector which offers an increased reprogramming efficiency up to 1.2%
essential 8
Essential 8 Mediumcontains only the 8 necessary components needed for stem cell culture enabling more consistent culture
psc
PSC Cryopreservation Kitcontains a xeno-free, ready-to-use cryopreservation medium and a chemically defined recovery supplement, RevitaCell Supplement, that consistently recovers 50% more viable cells post-thaw
essential 6
Essential 6 Mediumdoes not contain bFGF which is used to prevent differentiation and maintain pluripotency
For research use only. Not for use in diagnostic procedures. Essential 8 and Essential 6 are trademarks of Cellular Dynamics International, Inc.