Overman rearrangement
In 1974, the American chemist Larry Overman reported the thermal and mercuric ion catalyzed rearrangement of allylic trichloroacetimidates to create the corresponding trichloroacetamides. Subsequently the 1,3-transposition of alcohol and amine functionalities via [3,3]-sigmatropic rearrangement of allylic trichloroacetimidates became known as the Overman rearrangement. The allylic trichloroacetimidates are readily prepared by treating the corresponding alcohols with trichloroacetonitrile in the presence of catalytic amounts of a base, or DBU.
In recent decades, the Overman rearrangement has become an important carbon-nitrogen bond-forming reaction used in many synthesis campaigns. For example, during the total synthesis of sphingofungin E by N. Chida and colleagues, the stereocenter at C5 was formed using the Overman rearrangement of an allylic trichloroacetimidate derived from diacetone-D-glucose.
Mechanism of the Overman rearrangement reaction
Review available Thermo Scientific products for the Overman rearrangement reaction:
Catalog # Name Size Price (USD) Qty
A10565.18 Trichloroacetonitrile, 98% Each
38.65
Special offer
Online exclusive
Ends: 26-Sep-2025
45.30
Save 6.65 (15%)
A10565.30 Trichloroacetonitrile, 98% Each
91.65
Online Exclusive
102.00
Save 10.35 (10%)
A10565.0B Trichloroacetonitrile, 98% Each
291.65
Special offer
Online exclusive
Ends: 26-Sep-2025
343.00
Save 51.35 (15%)
H36949.AE n-Butyllithium, 2.5M in hexane, packaged under Nitrogen in resealable AcroSeal™ bottles Each
64.65
Special offer
Online exclusive
Ends: 30-Jun-2025
76.00
Save 11.35 (15%)
H36949.9Z n-Butyllithium, 2.5M in hexane, packaged under Nitrogen in resealable AcroSeal™ bottles Each
174.65
Online Exclusive
194.00
Save 19.35 (10%)
A12449.14 1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+% Each
38.90
A12449.22 1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+% Each
78.65
Special offer
Online exclusive
Ends: 26-Sep-2025
92.20
Save 13.55 (15%)
A12449.36 1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+% Each
290.00
L13317.AP Xylenes, mixed, 97+% Each
31.65
Online Exclusive
35.40
Save 3.75 (11%)
L13317.AU Xylenes, mixed, 97+% Each
39.65
Special offer
Online exclusive
Ends: 26-Sep-2025
46.30
Save 6.65 (14%)
L13317.0F Xylenes, mixed, 97+% Each
57.65
Special offer
Online exclusive
Ends: 26-Sep-2025
67.60
Save 9.95 (15%)
Vote for your favorite named reaction
Learn more about the most popular named reactions in chemistry based on feedback from our customers. Do you have a favorite reaction? Let us know!
Vote now