Oncomine Immune Repertoire NGS Assays for Hemato-Oncology Research
Next-generation sequencing in lymphoid malignancies
Lymphoid malignancies involve the clonal proliferation of one or more B or T cells. Every B and T cell expresses distinct receptors on its surface, which give rise to a vastly diverse immune repertoire. Using next-generation sequencing (NGS) technology, these unique receptor sequences can be used to assess clonality, detect rare clones, and measure somatic hypermutation (SHM).
NGS offers significant advantages over traditional approaches by providing sequence information, giving a more detailed view into repertoire (sub-clonal and intra-clonal) diversity, offering ultra-high sensitivity and providing greater flexibility to multiplex.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Watch how NGS can better better specificity and resolution than traditional methods for lymphoid malignancy research
      On Demand Webinar: Advances in Immune Repertoire Sequencing for the Study of Lymphoid Neoplasms
      Learn about the value of immune repertoire sequencing in hematological oncology research applications. Dr. DeCoteau
      shares his experience using the new Oncomine
      Pan-Clonality assays.
      Dr. John DeCoteau MD, FRCPC
      Professor of Pathology and Laboratory Medicine
      University of Saskatchewan
      John DeCoteau
      On Demand Webinar: Sequencing IGH Chains for Immuno-Oncology Research
      In this on-demand webinar, Timothy Looney describes the advantages of BCR-seq as a tool for determining the somatic hypermutation status of chronic lymphocytic leukemia (CLL) and highlights emerging translational research applications of BCR-seq as a biomarker discovery tool for adenosine pathway inhibition immunotherapy.
      Timothy Looney, PhD
      Senior Director of Immuno-Oncology
      Quest Diagnostics
      Timothy Looney
      Clonality and rare clone detection
      NGS provides sequence-level resolution for clonality assessment, allowing you to detect expanded clones from poly-clonal samples with very high specificity. Integrated bioinformatic tools let you easily assess clonal lineage to better understand the relationship between two clones.
      NGS offers a greater level of sensitivity when compared to traditional methods like flow cytometry. The ultra-low limit of detection (LoD) of 10-6 (1 in 1,000,000 cells) enables you to detect extremely rare clones that traditional less-sensitive methods can miss.
      Using proprietary Ion AmpliSeq technology, Oncomine immune repertoire assays can target multiple immune receptors in a single reaction, which can lead to increased positive clonality detection rates (>90%) and reduce the need for secondary testing. 
      clonality
      Oncomine pan-clonality assays
      Oncomine TCR Pan-Clonality Assay
      The Oncomine TCR Pan-Clonality Assay specifically interrogates the CDR3 region of the T-cell receptor (TCR) beta and gamma chains. Detect low frequency T-cell clones in peripheral blood, with sensitivity down to 10-6.
      Oncomine TCR Pan-Clonality Assay
      Oncomine BCR-SR Assay
      Assess clonality by targeting the CDR3 region of the IGH receptor. Have the flexibility to process DNA or RNA samples. Use RNA input to detect rare B cell clones with high sensitivity (LOD: 10-6), while maximizing cost efficiency.
      Oncomine BCR-SR Assay
      Oncomine BCR Pan-Clonality Assay
      This powerful and sensitive NGS assay can accurately assess clonality and detect rare clones in a range of sample types including blood, bone marrow, and FFPE preserved tissues.
      Simultaneously sequence multiple receptor targets in a single reaction, including IGH, IgK, and IgL rearrangements, as well as kappa deletion element (KDE)-containing rearrangements.
      Oncomine BCR Pan-Clonality Assay
      Somatic hypermutation analysis assays
      Following V(D)J recombination in developing lymphocytes, the IGHV gene undergoes somatic hypermutation. During this process, a series of point mutations are introduced to help confer greater repertoire diversity and enable higher affinity for potential antigens.
      The degree of somatic hypermutation is a key biomarker relevant for chronic lymphocytic leukemia (CLL) research. Long-amplicon NGS assays provide highly accurate IGHV SHM quantification, while enabling efficient batch sample processing and simplifying the workflow when compared to traditional Sanger sequencing methods.
      Somatic Hypermutation
      Oncomine IGHV Leader-J Assay
      Sequence from the Leader region of the BCR IGHV gene to assess SHM frequency. The Leader-J Assay adheres to recommendations by the European Research Initiative on CLL (ERIC). These standards in CLL research aid in understanding of the biological and clinical relevance for immunogenetic analysis.
      Oncomine IGHV Leader-J Assay
      Oncomine BCR IGH LR Assay
      Accurately measure the level of SHM from the FR1 to the constant region of the BCR IGHV gene. Identify all isotypes (and subtypes) to expand immune repertoire research possibilities. Process various sample types, including blood and bone marrow, using a low RNA input requirement (25 ng).
      Oncomine BCR IGH LR Assay
      Powerful informatics and visualization tools
      Interactive spectratyping plots make it easy to identify clonal expansion within the broader context of the repertoire. Automated reporting features provide detailed information on each clone, including the CDR3 sequence, SHM frequency, clone frequency, and more.
      informatics
      Scientific posters
      Evaluation of Multiple Myeloma Research Samples by Analysis of B Cell Heavy and Light Chain Receptors in a Single NGS Assay
      aacr 2022 poster
      Somatic Hypermutation Analysis of Chronic Lymphocytic Leukemia Research Samples by DNA or RNA Input IGH Chain Sequencing
      aacr 2022 poster
      EAHP 2021 hemato-oncology symposium on demand
      Watch this on-demand presentation to hear our customers discuss the latest advancements in NGS testing for lymphoid neoplasm samples.
      Ordering information
      Ready to speak to a Thermo Fisher representative?
      We will be happy to answer your questions and provide a demo of our NGS solutions.
      For Research Use Only. Not for use in diagnostic procedures.