Zeba spin desalting columns
Thermo Scientific Zeba Spin Desalting Columns are made of low protein-binding polypropylene and are compatible with a wide range of standard laboratory instruments and consumables.
Our Zeba columns are designed for use with most swinging-bucket or fixed-angle bench- or floor-model centrifuges (please verify head clearance before use), and are available in six volume capacities and two molecular weight cut-offs (MWCOs), 7K and 40K.
The MWCO should be lower than the molecular weight of the target molecule to help ensure it is retained while contaminants are removed. Columns with 7K help provide finer resolution for smaller molecules, useful for purifying smaller proteins (e.g., cytochrome c) or peptides (e.g., insulin). Columns with 40K are better suited for large proteins (e.g., BSA), protein complexes (e.g., hemoglobin) and large enzymes (e.g,. pyruvate kinase).
Choose the right Zeba spin desalting column with 7K MWCO
 zeba desalting spin column: zeba desalting spin columndesalting column kitdesalting column kitdesalting column kitzeba desalting spin plate
Sample volume2–12 µL30–130 µL200–700 µL500–2,000 µL700–4,000 µL4,000–10,000 µL
Resin bed
volume
75 μL0.5 mL2 mL5 mL10 mL20 mL
Product typeMicro Spin Column0.5 mL Spin Column2 mL Spin Column5 mL Spin Column10 mL Spin Column20 mL Spin Column
See all 7K MWCO Zeba Spin Desalting Columns, Plates, and Cartridges ›
Choose the right Zeba spin desalting column with 40K MWCO
 zeba desalting spin column: zeba desalting spin columndesalting column kitdesalting column kitdesalting column kitzeba desalting spin plate
Sample volume5–14 µL70–200 µL200–900 µL300–2,000 µL1,000–4,000 µL4,000–10,000 µL
Resin bed
volume
75 μL0.5 mL2 mL5 mL10 mL20 mL
Product typeMicro Spin Column0.5 mL Spin Column2 mL Spin Column5 mL Spin Column10 mL Spin Column20 mL Spin Column
See all 40K MWCO Zeba Spin Desalting Columns, Plates, and Cartridges ›
Choose the right Zeba product for high throughput and automation
 zeba desalting spin column: zeba desalting spin columndesalting column kitdesalting column kit
MWCO7K 7K 7K 40K 
Sample volume20–100 µL50–250 µL100–1,500 µL20–100 µL
Resin bed volumeN/A1 mL5 mLN/A
Product type96-well filter plates1 mL Desalting Cartridges 5 mL Desalting Cartridges 96-well filter plates
Additional products used to efficiently wash away contaminates and elute your purified sample:
Catalog # Name Size Price (USD) Qty
89877 Zeba™ Micro Spin Desalting Columns, 7K MWCO, 75 μL, 25 Columns Each
172.65
Online Exclusive
190.00
Save 17.35 (9%)
89878 Zeba™ Micro Spin Desalting Columns, 7K MWCO, 75 μL, 50 Columns Each
309.65
Online Exclusive
336.00
Save 26.35 (8%)
89882 Zeba™ Spin Desalting Columns, 7K MWCO, 0.5 mL, 25 Columns Each
162.65
Online Exclusive
176.00
Save 13.35 (8%)
89883 Zeba™ Spin Desalting Columns, 7K MWCO, 0.5 mL, 50 Columns Each
282.65
Online Exclusive
312.00
Save 29.35 (9%)
89889 Zeba™ Spin Desalting Columns, 7K MWCO, 2 mL, 5 Columns Each
68.65
Online Exclusive
75.25
Save 6.60 (9%)
89890 Zeba™ Spin Desalting Columns, 7K MWCO, 2 mL, 25 Columns Each
290.65
Online Exclusive
315.00
Save 24.35 (8%)
89891 Zeba™ Spin Desalting Columns, 7K MWCO, 5 mL, 5 Columns Each
82.65
Online Exclusive
91.50
Save 8.85 (10%)
89892 Zeba™ Spin Desalting Columns, 7K MWCO, 5 mL, 25 Columns Each
364.65
Online Exclusive
404.00
Save 39.35 (10%)
89893 Zeba™ Spin Desalting Columns, 7K MWCO, 10 mL, 5 Columns Each
103.65
Online Exclusive
115.00
Save 11.35 (10%)
89894 Zeba™ Spin Desalting Columns, 7K MWCO, 10 mL, 25 Columns Each
437.65
Online Exclusive
487.00
Save 49.35 (10%)
89934 Zeba™ Desalting Chromatography Cartridges, 7K MWCO, 1 mL, 5 columns x 1 mL Each
224.65
Online Exclusive
250.00
Save 25.35 (10%)
89935 Zeba™ Desalting Chromatography Cartridges, 7K MWCO, 5 mL, 1 column x 5 mL Each
269.65
Online Exclusive
293.00
Save 23.35 (8%)
89807 Zeba™ Spin Desalting Plates, 7K MWCO, 2 Plates Each
322.65
Online Exclusive
367.00
Save 44.35 (12%)
89808 Zeba™ Spin Desalting Plates, 7K MWCO, 4 Plates Each
608.65
Online Exclusive
660.00
Save 51.35 (8%)
A40002469 Zeba XL Spin Desalting Columns, 7K MWCO, 5 Columns 5 columns
223.65
Online Exclusive
243.00
Save 19.35 (8%)
A40002475 Zeba XL Spin Desalting Columns, 7K MWCO, 10 Columns 10 columns
441.65
Online Exclusive
478.00
Save 36.35 (8%)
Catalog # Name Size Price (USD) Qty
A57756 Zeba™ Micro Spin Desalting Columns, 40K MWCO, 25 Columns, 25 columns Each
174.65
Online Exclusive
186.00
Save 11.35 (6%)
A57758 Zeba™ Micro Spin Desalting Columns, 40K MWCO, 50 Columns, 50 columns Each
307.65
Online Exclusive
328.00
Save 20.35 (6%)
A57759 Zeba™ Spin Desalting Columns, 40K MWCO, 0.5 mL, 25 Columns, 25 columns Each
158.65
Online Exclusive
169.00
Save 10.35 (6%)
A57760 Zeba™ Spin Desalting Columns, 40K MWCO, 0.5 mL, 50 Columns, 50 columns Each
286.65
Online Exclusive
306.00
Save 19.35 (6%)
A57761 Zeba™ Spin Desalting Columns, 40K MWCO, 2 mL, 5 Columns, 5 columns Each
68.65
Online Exclusive
73.50
Save 4.85 (7%)
A57762 Zeba™ Spin Desalting Columns, 40K MWCO, 2 mL, 25 Columns, 25 columns Each
288.65
Online Exclusive
308.00
Save 19.35 (6%)
A57763 Zeba™ Spin Desalting Columns, 40K MWCO, 5 mL, 5 Columns, 5 columns Each
84.65
Online Exclusive
89.50
Save 4.85 (5%)
A57764 Zeba™ Spin Desalting Columns, 40K MWCO, 5 mL, 25 Columns, 25 columns Each
373.65
Online Exclusive
398.00
Save 24.35 (6%)
A57765 Zeba™ Spin Desalting Columns, 40K MWCO, 10 mL, 5 Columns, 5 columns Each
107.65
Online Exclusive
114.00
Save 6.35 (6%)
A57766 Zeba™ Spin Desalting Columns, 40K MWCO, 10 mL, 25 Columns, 25 columns Each
450.65
Online Exclusive
480.00
Save 29.35 (6%)
A57767 Zeba™ Spin Desalting Plates, 40K MWCO, 2 Plates, 2 plates Each
339.65
Online Exclusive
362.00
Save 22.35 (6%)
A57768 Zeba™ Spin Desalting Plates, 40K MWCO, 4 Plates, 4 plates Each
607.65
Online Exclusive
648.00
Save 40.35 (6%)
A40002471 Zeba XL Spin Desalting Columns, 40K MWCO, 5 Columns 5 columns
227.65
Online Exclusive
243.00
Save 15.35 (6%)
A40002476 Zeba XL Spin Desalting Columns, 40K MWCO, 10 Columns 10 columns
448.65
Online Exclusive
478.00
Save 29.35 (6%)
Catalog # Name Size Price (USD) Qty
89879 Pierce™ Micro-Spin Columns Each
92.65
Online Exclusive
104.00
Save 11.35 (11%)
69705 Pierce™ Spin Columns - Screw Cap Each
160.65
Online Exclusive
182.00
Save 21.35 (12%)
69725 Pierce™ Spin Columns - Snap Cap Each
211.00
A40000364 Pierce™ Centrifuge Columns, 34 mL, 25 Columns 25 columns
212.00
Zeba Spin Desalting Columns and Plates video
Video Player is loading.
Current Time 0:00
Duration 1:48
Loaded: 0.00%
Stream Type LIVE
Remaining Time 1:48
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Thermo Scientific Zeba Spin Desalting Columns
    Zeba spin desalting columns quickly and effectively remove salts and other small molecules from your sample using gel-filtration chromatography. They contain ready-to-use size exclusion resin that enables high salt retention and protein recovery. Zeba columns eliminate the need for gravity-filtration, thus speeding up the desalting process. View the video to learn how to desalt your sample using the Zeba Spin Desalting Columns.
    Zeba spin desalting column performance
    Zeba Spin Desalting Columns are available in a variety of sizes and volume capacities compared to competitor offerings (Table 1). Zeba columns can also be used with volumes as small as 5 uL (in micro format) to as large as 10 mL
    Zeba 40K Spin Desalting Columns contain a new and enhanced high-performance size-exclusion chromatography resin. These redeveloped Zeba columns demonstrate improved protein recovery for a variety of proteins in the 40 kDa and above molecular weight range (Table 2).
     Recovery*
    ProteinMolecular Size7K Zeba Column40K Zeba Column
    Bacitracin1.5 kDa15%12%
    Aprotinin6.5 kDa58%37%
    Ubiquitin8.5 kDa78%51%
    Lactalbumin14 kDa88%64%
    Trypsin Inhibitor21 kDa94%76%
    Carbonic Anhydrase30 kDa90%79%
    Ovalbumin42 kDa95%95%
    BSA67 kDa94%93%
    Rabbit IgG150 kDa92%92%
    *Recovery of 100 uL sample volume, with a loaded concentration of 0.5 mg/mL
    Table 2Protein recovery comparison using Zeba Protein Desalting Columns with 7K and 40K MWCO resin.
    Zeba 7K Spin Desalting Columns contain a high-performance size-exclusion chromatography resin that allows for exceptional protein desalting and recovery. When compared to competitors, Zeba 7K desalting columns have superior protein recovery even at dilute protein concentrations (Figure 1).
    Figure 1. Zeba Micro Spin Desalting Columns, 7K MWCO, recovery performance. BSA and ubiquitin samples of varying concentrations were processed using Zeba Micro Spin Desalting Columns, 7K MWCO, alongside comparable competitor products. The data show that, even at very dilute protein concentrations (250 ng), the Zeba Desalting columns outperform those of the competitors, yielding higher protein recovery for both BSA and ubiquitin.
    Zeba 40K Spin Desalting Columns provide superior performance that improves protein recovery compared to competitors (Figure 2). Further, the new resin in the Zeba 40K desalting columns allows for higher desalting and is more effective at small protein removal (Figure 3).
    Figure 2. Exceptional sample recovery of low protein loads with Zeba 40K spin columns. BSA and IgG of varying concentrations were applied in 50µl aliquots to either 0.5 mL Zeba 40K, competitor C, or Competitor B spin columns. All columns were equilibrated with PBS, then centrifuged per manufacturers’ instructions for each resin. Recovered protein (%) in each flow through sample was determined by BCA assay. Flow through samples were performed on SDS-PAGE and stained with either Coomassie Blue or silver stain. Although 12.5 µg total protein load was recovered efficiently by all three resins, lower recoveries were observed with competitor resins at 1 ug and 250 ng protein loads.
    Figure 3. New enhanced Zeba 40K resin is more effective than that of competitors at removing small proteins. Purified protein samples (0.5mg/mL) of varying molecular weights were applied individually in 50 ul aliquots in either 0.5 mL Zeba 40K, Competitor C, or Competitor B spin columns equilibrated in PBS. Columns were centrifuged per manufacturers’ instructions for each resin, and the recovered protein (%) in each flow-through sample was determined by BCA protein assay. As opposed to competitor columns, the Zeba 40K resin was more efficient at removing smaller proteins (evidenced by the lower small protein recovery values).
    Zeba Desalting Handbook
    Thermo Scientific Zeba Resin technology provides consistent performance and peace of mind. The unique, proprietary Zeba Resin enables better protein recovery, even for samples with low protein concentrations.
    Download the Zeba Desalting Handbook ›
    front cover of zeba desalting handbook
    For Research Use Only. Not for use in diagnostic procedures.