48ab - Appl/Samples/Products/Resources/Contact
Sample preparation for scanning/transmission electron microscopy (S/TEM) analysis is considered to be one of the most critical but challenging and time-consuming tasks in materials characterization labs. Conventional methods used to prepare ultra-thin samples required for S/TEM are slow, typically requiring many hours or even days of effort by highly trained personnel. This is further complicated by the variety of different materials and the need for site-specific information. For more than 30 years, Thermo Fisher Scientific has been continuously developing and optimizing sample preparation instruments and introducing software that enable in situ TEM sample preparation that allows researchers to accelerate their research and fast-track innovation.
Thermo Fisher Scientific has 30+ years of TEM sample preparation expertise. In 1993, we introduced DualBeam (FIB-SEM, focused ion beam – scanning electron microscopy) technology to give you cutting-edge sample preparation tools that are accessible, robust, and reliable. This begins with a highly stable column and high-quality source, which allows for excellent performance even at low voltages. Final polishing of the lamella using energies as low as 500 V greatly reduces damage, even to beam sensitive materials, and delivers unrivaled sample quality. For gallium-free sample preparation, Thermo Fisher Scientific has a broad portfolio of plasma FIB products, including the Thermo Scientific Helios Hydra DualBeam, which offers fast switching between four ion beam species; xenon, argon, oxygen, and nitrogen.
30+ years of innovation with TEM sample preparation
Video Player is loading.
Current Time 0:00
Duration 11:12
Loaded: 1.49%
Stream Type LIVE
Remaining Time 11:12
1x
    • Chapters
    • descriptions off, selected
    • en (Main), selected
    Additionally, Thermo Scientific SmartAlign Technology eliminates the need for any user alignments of the electron column, which not only minimizes maintenance, but also increases your productivity. Auto-tuning tools, embedded into the user interface, further increasing your ability to get high-quality images, tuning the image up to 10-times faster than standard manual alignment.
    Quality hardware is supported by a suite of software solutions that enhance and simplify sample preparation. Thermo Scientific AutoTEM 5 Software enables fully automated in situ lamella preparation and lift-out, allowing even novice users to produce quality samples. This greatly expands the accessibility of the instrument, thereby increasing throughput as sample preparation is no longer reliant on expert operation. Moreover, full automation allows maximum utilization of system time with unattended overnight operation, resulting in greatly enhanced productivity.
    TEM sample preparation is part of a multi-scale, multi-modal workflow that includes microCT, (P)FIB-SEM, and (S)TEM. First, data is acquired using the microCT, followed by data reconstruction and analysis using Thermo Scientific Avizo Software. Avizo Software visualizes the data and enables you to precisely locate the suitable regions of interest (ROI) for further exploration of the sample. When the ROI is identified, the workflow progresses through higher-resolution imaging and sample preparation with a plasma-focused ion beam (PFIB) DualBeam to atomic-scale analysis in a transmission electron microscope (TEM). When combined, these technologies provide a complete understanding of sample structure and composition, accelerating development of novel materials.
    Automated S/TEM sample preparation in a multiscale workflow
    2021-MS-TEM-Sample-Prep-Workflow-Infographic-Web_1200x500_416264
    Video Player is loading.
    Current Time 0:00
    Duration 6:18
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 6:18
    1x
      • Chapters
      • descriptions off, selected
      • en (Main), selected
      Joseph Michael on how TEM sample preparation has progressed over the past 30 years
      Joseph Richard Michael, who worked many years at Sandia National Laboratories in Materials, Physical and Chemical Sciences, tells his story of how TEM sample preparation changed the way he and his fellow materials scientists were able to do their research.
      Video Player is loading.
      Current Time 0:00
      Duration 1:01:40
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 1:01:40
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        The Single-Beam and Dual-Beam Revolution: How FIB Instruments Transformed TEM Sample Preparation
        In this podcast we hear David Foord, product director and materials scientist, in talks with some of his colleagues at Thermo Fisher Scientific. What started as a casual internal conversation around the driving force behind 30+ years of ion beam technology was so interesting that we wanted to share David’s story with you. This podcast proves that people at Thermo Fisher Scientific drive the field of materials science forward with solution-focused innovation that always circles around one question: What problem is this technology going to solve?
        Nanoscale Materials Characterization
        Understanding the nanoscale characteristics of materials is the first step in creating the next generation of lighter, stronger, and more energy efficient solutions. With our electron microscopy solutions, we help you simplify this complex material journey and accelerate your nanoscale materials research. Nanoscale workflow is a journey of material from bulk to nanometer scales using our DualBeam FIB-SEM for sample preparation and Transmission Electron Microscopes for material analysis. 
        Resources
        Introducing Helios 5 DualBeam
        Introducing Helios 5 DualBeam
        Learn how advanced automation capabilities, increased robustness and stability enhancements in the Helios 5 and AutoTEM 5 software can significantly increase sample preparation throughout by allowing unattended and even overnight operation.
        Register now
        Webinar TEM Specimen Preparation
        Toward “Damage-Free” TEM specimen preparation by Focused Ion Beam without Gallium
        Diminish the challenges associated with focused ion beam (FIB) sample preparation for high resolution transmission electron microscopy (HR-TEM). Dr. Chengge Jiao, Staff Scientist of Applications Development at Thermo Fisher Scientific, explains how to tailor processing conditions and select the right FIB ion species for certain groups of materials to achieve optimized results.
        Register now
        Webinar TEM Specimen Preparation
        Nano-, micro-, and mesoscale sample preparation by focused ion beam and laser milling
        Listen to Dr. Joakim Reuteler, a FIB expert from ETH Zurich, discuss how gallium focused ion beam, multi-ion species plasma FIB, and ultra-short-pulse laser milling can help you prepare samples for a wide range of applications.
        Register now
        Introducing Helios 5 DualBeam
        Introducing Helios 5 DualBeam
        Learn how advanced automation capabilities, increased robustness and stability enhancements in the Helios 5 and AutoTEM 5 software can significantly increase sample preparation throughout by allowing unattended and even overnight operation.
        Register now
        Webinar TEM Specimen Preparation
        Toward “Damage-Free” TEM specimen preparation by Focused Ion Beam without Gallium
        Diminish the challenges associated with focused ion beam (FIB) sample preparation for high resolution transmission electron microscopy (HR-TEM). Dr. Chengge Jiao, Staff Scientist of Applications Development at Thermo Fisher Scientific, explains how to tailor processing conditions and select the right FIB ion species for certain groups of materials to achieve optimized results.
        Register now
        Webinar TEM Specimen Preparation
        Nano-, micro-, and mesoscale sample preparation by focused ion beam and laser milling
        Listen to Dr. Joakim Reuteler, a FIB expert from ETH Zurich, discuss how gallium focused ion beam, multi-ion species plasma FIB, and ultra-short-pulse laser milling can help you prepare samples for a wide range of applications.
        Register now
        Applications
        Process control using electron microscopy
        Process control using electron microscopy
        Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
        Quality control and failure analysis using electron microscopy
        Quality control and failure analysis
        Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
        Fundamental Materials Research_R&D_Thumb_274x180_144DPI
        Fundamental Materials Research
        Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
        Samples
        Battery Research
        Battery development is enabled by multi-scale analysis with microCT, SEM and TEM, Raman spectroscopy, XPS, and digital 3D visualization and analysis. Learn how this approach provides the structural and chemical information needed to build better batteries.
        Learn more ›
        Polymers Research
        Polymer microstructure dictates the material’s bulk characteristics and performance. Electron microscopy enables comprehensive microscale analysis of polymer morphology and composition for R&D and quality control applications.
        Learn more ›
        Metals Research and Characterization
        Effective production of metals requires precise control of inclusions and precipitates. Our automated tools can perform a variety of tasks critical for metal analysis including; nanoparticle counting, EDS chemical analysis and TEM sample preparation.
        Learn more ›
        Catalysis Research
        Catalysts are critical for a majority of modern industrial processes. Their efficiency depends on the microscopic composition and morphology of the catalytic particles; EM with EDS is ideally suited for studying these properties.
        Learn more ›
        Style Sheet to change H2 style to p with em-h2-header class
        Style Sheet for Komodo Tabs
        Products
        Style Sheet for Instrument Cards Original
        FIB-SEM and Laser Ablation
        • All three beams have same coincident point for accurate and repeatable cut placement
        • Millimeter-scale cross sections with up to 15,000x faster material removal than a typical FIB
        • Statistically relevant deep subsurface and 3D data analysis
        Thermo Scientific Helios Hydra plasma focused ion beam scanning electron microscope (DualBeam)
        Helios Hydra DualBeam
        • 4 fast switchable ion species (Xe, Ar, O, N) for optimized PFIB processing of a widest range of materials
        • Ga-free TEM sample preparation
        • Extreme high resolution SEM imaging
        Helios 5 HX/Helios 5 UX/Helios 5 FX DualBeam
        • Fully automated, high-quality, ultra-thin TEM sample preparation
        • High throughput, high resolution subsurface and 3D characterization
        • Rapid nanoprototyping capabilities
        Helios 5 PFIB DualBeam
        • Gallium-free STEM and TEM sample preparation
        • Multi-modal subsurface and 3D information
        • Next-generation 2.5 μA xenon plasma FIB column
        Spectra 300
        • Highest-resolution structural and chemical information at the atomic level
        • Flexible high-tension range from 30-300 kV
        • Three lens condenser system
        Spectra 200
        • High-resolution and contrast imaging for accelerating voltages from 30-200 kV
        • Symmetric S-TWIN/X-TWIN objective lens with wide-gap pole piece design of 5.4 mm
        • Sub-Angstrom STEM imaging resolution from 60 kV-200 kV
        Talos F200i TEM
        • Compact design with X-TWIN objective lens
        • Available with S-FEG, X-FEG, and X-CFEG
        • Flexible and fast EDS options for comprehensive elemental analysis
        Talos F200S TEM
        • Intuitive and easy-to-use automation software
        • Available with Super-X EDS for rapid quantitative chemical analysis
        • High-throughput with simultaneous multi-signal acquisition
        Talos F200X TEM
        • High-resolution, EDS cleanliness, and quality in 2D as well as 3D
        • X-FEG and X-CFEG available for the highest brightness and energy resolution
        • High accuracy and repeatable results with integrated Thermo Scientific Velox Software
        Thermo Scientific Scios 2 plasma focused ion beam scanning electron microscope (DualBeam)
        Scios 2 DualBeam
        • Full support of magnetic and non-conductive samples
        • High throughput subsurface and 3D characterization
        • Advanced ease of use and automation capabilities
        AutoTEM 5
        • Fully automated in situ S/TEM sample preparation
        • Support of top-down, planar and inverted geometry
        • Highly configurable workflow
        • Easy to use, intuitive user interface
        Contact us
        Style Sheet for Support and Service footer
        Style Sheet for Fonts
        Style Sheet for Cards
        Electron microscopy services for
        the materials science
        To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.