Talk with Us
Phadia 250 Laboratory System
Fully automated, immunoassay system, aiding in the diagnosis of autoimmune and allergic diseases.
Ideal for labs performing 400-2,000 tests/per week
Size: With table cart: 73 x 50 x 30 in
Maximum throughput: Up to 60 results/hour
Reliability: 98% uptime
Sample type: Serum, plasma
Sample tubes for sample rack: 10 and 16 position rack options, Height: 50-110mm, Diameter: 10-17mm
Testing capabilities: ImmunoCAP Allergy solutions & EliA Autoimmunity solutions
Testing methodology: fluoroenzyme immunoassay (FEIA)
See all details
Ready to gain effortless workflow efficiency with Phadia 250 instrument?
Pop the hood and get a good look at the instrument’s capabilities.
Let the Phadia 250 system be the backbone in maximizing your lab’s workflow.
Set up and walkway
With a 15-minute, low hands-on start-up time and automatic shutdown and rinse capability, gain extended periods of walkaway time.1
28-day stored calibration curve
Using a master calibration curve by isotypes, Phadia 250 system requires less upkeep and generates less waste.1
Easily scalable systems
The Phadia™ Prime software’s intuitive and user-friendly interface to allow for seamless system management as well as the ability to scale and network multiple Phadia systems from one computer.1
Don’t stay shackled to the hood.
Gain the freedom to walk away and let the Phadia 250 system do the heavy lifting.
Enhance your lab today.
Chat with us
Save on reagents and time
Phadia 250 system test menu
One lab system with the capabilities of running both ImmunoCAP Allergy solutions and EliA Autoimmunity solutions.
Over 550 whole allergens and mixes as well as 100 allergen components that aide in the diagnosis of a wide range of allergies, such as...
  • Peanut and tree nuts
  • Milk and egg
  • Alpha-Gal syndrome
  • Pollen
  • Pet dander
  • And more…
EliA Autoimmunity Solutions
Compare Phadia Laboratory Systems
See how the Phadia 250 system compares to other instruments in our portfolio.
Compare instruments
Overhaul your autoimmunity and allergy diagnostics with effortless efficiency.
References
  1. Phadia® 250 User Manual 12-3904-01/EN Edition 2.2. published 2017 Sep.
  2. Phadia® 1000 User Manual 12-3803-01/EN Edition 2.6. published 2020 July
  3. Phadia® 2500 and Phadia 5000® User Manual EN Edition 1.1. published 2012 March