Blood testing for allergies
Accurately identify allergy triggers for targeted symptom management
A blood allergy test, also called a specific IgE test, is a powerful diagnostic tool that measures the concentration of specific IgE antibodies in the blood.1 It can test for hundreds of allergic triggers, such as pollen, mold, food and animal dander with one blood sample.
Any detectable level of specific IgE indicates that you are sensitized to that allergen. Even a low level can be significant, especially if you experience symptoms when exposed to the allergen. The opposite can also be true. You might have a detectable level of IgE to an allergen, but if you don't show symptoms when exposed, you're not considered clinically allergic.
So, what does that mean? It means if you aren’t sure what are causing symptoms, a specific IgE blood test may help your healthcare provider rule in or rule out allergies. The results of a blood allergy test, a detailed medical history, and a physical examination will help a healthcare provider develop a customized treatment plan for your specific allergic triggers and their symptoms. And even if a clinical allergy isn't diagnosed, that's still some powerful information for a healthcare provider to use to help address symptoms.