Search Thermo Fisher Scientific
E-Gel DNA Ladders and E-Gel Loading Buffer |
Invitrogen E-Gel DNA Ladders are double stranded DNA fragments of defined mass and size suspended pre-mixed with gel loading buffer. These ladders are formulated for optimal performance on Invitrogen E-Gel Precast Agarose Gels. For best results, use with Invitrogen E-Gel Sample Loading Buffer.
Use the DNA ladder suited to a specific application for accurate band analysis. Table 1 lists E-Gel DNA Ladders available by high-to-low bp range. For optimal performance, use the right combination of E-Gel DNA Ladders and E-Gel Precast Agarose Gels as specified in Table 2.
E-Gel 96 High Range DNA Marker | E-Gel 1 Kb Plus DNA Ladder | E-Gel 1 Kb Plus Express DNA Ladder | E-Gel Low Range Quantitative DNA Ladder | E-Gel 50 bp DNA Ladder | E-Gel Sizing DNA Ladder | E-Gel Ultra Low Range DNA Ladder |
---|---|---|---|---|---|---|
400–10,000 bp | 100–15,000 bp | 100–5,000 bp | 100–2,000 bp | 50–2,500 bp | 50–1,500 bp | 10–300 bp |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
E-Gel 1 Kb Plus DNA Ladder | E-Gel 1 Kb Plus Express DNA Ladder | E-Gel 50 bp DNA Ladder | E-Gel 96 High Range DNA Marker | E-Gel Low Range Quantitative DNA Ladder | E-Gel Ultra Low DNA Ladder | Millennium RNA Marker | Century-Plus RNA Ladder | E-Gel Sizing DNA Ladder | |
---|---|---|---|---|---|---|---|---|---|
E-Gel Agarose Gels with SYBR Safe DNA Stain, 1% | ✓ | ||||||||
E-Gel Agarose Gels with SYBR Safe DNA Stain, 2% | ✓ | ||||||||
E-Gel Agarose Gels with SYBR Safe DNA Stain, 4% | ✓ | ✓ | |||||||
E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 1% | ✓ | ✓ | |||||||
E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 2% | ✓ | ✓ | |||||||
E-Gel EX Gel, 1% | ✓ | ✓ | |||||||
E-Gel EX Gel, 2% | ✓ | ✓ | ✓ | ✓ | |||||
E-Gel EX Gel, 4% | ✓ | ✓ | ✓ | ||||||
E-Gel EX Double Comb Agarose Gels, 1% | ✓ | ✓ | |||||||
E-Gel EX Double Comb Agarose Gels, 2% | ✓ | ✓ | |||||||
E-Gel Go! Agarose Gels, 1% | ✓ | ✓ | ✓ | ||||||
E-Gel Go! Agarose Gels, 2% | ✓ | ✓ | |||||||
E-Gel CloneWell II | ✓ | ||||||||
E-Gel SizeSelect II | ✓ | ||||||||
E-Gel NGS | ✓ | ✓ | |||||||
E-Gel 48-well, 1% | ✓ | ✓ | ✓ | ✓ | |||||
E-Gel 48-well, 2% | ✓ | ✓ | ✓ | ||||||
E-Gel 48-well, 4% | ✓ | ✓ | |||||||
E-Gel 96-well, 1% | ✓ | ||||||||
E-Gel 96-well, 2% | ✓ |
E-Gel Sample Loading Buffer is ready to use out of the tube, requiring no further dilution. It contains the unique tracking dyes xylene cyanol FF (XCFF) and tartrazine; these dyes visually track DNA migration during electrophoresis, and indicate when maximum resolution is achieved.
Watch educational and insightful videos on molecular biology techniques.
View live and recorded webinars on research trends in molecular biology.
Access white papers, application notes, and technical notes on molecular biology.
Find the right E-Gel precast gels and DNA ladders for your applications.
For Research Use Only. Not for use in diagnostic procedures.