Choose from a variety of stand-alone PCR enzymes and master mixes optimized for your applications and experimental needs. 
PCR master mix
Our new generation of Platinum DNA polymerases come with novel buffers enabling universal primer annealing at a temperature of 60°C. The universal primer annealing allows all primers to anneal to the template DNA at 60°C, eliminating Tm calculations and enhancing convenience. A combination of the innovative buffers, high-performing enzymes, and reliable hot-start technology offers exceptional PCR results, even in the toughest applications.
A PCR master mix is a pre-mixed solution that contains a thermostable DNA polymerase, dNTPs (deoxynucleotide triphosphates), buffer, and other additives. The optimized master mix formulation helps ensure consistent and reliable results, save time, and reduce the potential for errors during reaction set up. In addition, PCR master mixes can be an economical choice compared to purchasing individual enzymes and reagents.
Video Player is loading.
Current Time 0:00
Duration 2:39
Loaded: 0.00%
Stream Type LIVE
Remaining Time 2:39
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Learn how our commitment to enzyme excellence, including the exceptional reliability and high-fidelity of Platinum DNA Polymerases, are leading the way in the pursuit of science.
    Interactive enzyme selection tool: Quickly find the DNA polymerase you need for your PCR!
    Compare PCR enzyme and master mix by PCR type
    Hot-start PCR
    Hot-start PCR is a type of PCR where DNA polymerase activity is inhibited until the initial denaturation step. This inhibition is achieved through various methods, such as antibodies, affibodies, or chemical modifications of the polymerase. The benefits of hot-start PCR include increased specificity and yield.
    product package and three tubes of reagents
    Platinum II Taq Hot-Start DNA Polymerase
    product package and four tubes of reagents
    AmpliTaq Gold 360 DNA Polymerase
    Hot-start technologyAntibodyChemical
    Enzyme activation time2 min10 min
    Universal annealing temperature of 60°CYesNo
    DNA synthesis speed15 sec/kb60 sec/kb
    Flexible extension step*YesNo
    Inhibitor toleranceYesNo
    Amplification lengthUp to 5 kbUp to 5 kb
    GC-rich formatYesYes
    Stand-alone enzymeColorless†
    Order now
    Colorless
    Order now
    Master mix format
    Colorless
    Order now
    Green**
    Order now
    Colorless
    Order now
    *The extension step can be extended up to 60 sec/kb without the effect of specificity.
    **Contains electrophoresis tracking dyes and a density reagent for direct gel loading of PCR products.
    Green buffer available as separate item for use with stand-alone enzyme.
    Note: The original Platinum Taq DNA Polymerase is available in the formats of stand alone, stand alone with gel-loading dyes, master mix, and master mix with gel-loading dyes.
    High-fidelity PCR
    High fidelity PCR, also known as proofreading PCR, is a technique that utilizes DNA polymerases with a built-in proofreading capability to ensure accurate sequence amplification with no errors introduced in the sequence. 
    Product package and three tubes of reagents
    Platinum SuperFi II DNA Polymerase
    Product package and three tubes of reagents
    Platinum Taq DNA Polymerase High Fidelity
    Fidelity vs. Taq enzyme>300x6x
    Hot start for enhanced specificityYesYes
    Universal annealing temperature of 60°CYesNo
    Amplification lengthUp to 20 kb*Up to 20 kb
    Amplicon overhangsBlunt3′ A/Blunt
    GC-rich amplificationYes
    No
    Stand-alone enzymeColorless
    Order now
    Colorless
    Order now
    Master mix format
    Colorless
    Order now
    Green**
    Order now
    Colorless
    Order now
    *Amplification of >20 kb fragment sizes is possible (up to 40 kb), but may require additional optimization of reaction conditions and primer design.
    **Contains electrophoresis tracking dyes and a density reagent for direct gel loading of PCR products.
    Long-range PCR
    Long-range PCR is a technique used to amplify DNA fragments that are longer than 5 kb, which is larger than the typical limit of standard PCR. It involves modifying the PCR conditions and utilizing specialized DNA polymerases to overcome the challenges associated with amplifying longer DNA sequences. 
    Product photo of package and three tubes of reagents.
    Platinum SuperFi II DNA Polymerase
    Amplification lengthUp to 20 kb*
    Fidelity vs. Taq enzyme>300x
    Hot start for enhanced specificityYes
    Universal annealing temperature of 60°CYes
    GC-rich amplificationYes
    DNA synthesis speed15–30 sec/kb
    Stand-alone enzymeColorless
    Order now
    Master mix format
    Colorless
    Order now
    Green**
    Order now
    *Amplification of >20 kb fragment sizes is possible (up to 40 kb), but may require additional optimization of reaction conditions and primer design.
    **Contains electrophoresis tracking dyes and a density reagent for direct gel loading of PCR products.
    White paper Assembly of large PCR amplicons by the Gibson Assembly method
    Multiplex PCR
    Multiplex PCR is a technique that allows for the concurrent amplification of multiple target DNA sequences within a single PCR reaction. It involves the use of multiple primer sets, each specific to a different target sequence, but amplified in a single reaction mixture.
    Product package and three tubes of reagents
    Platinum SuperFi II DNA Polymerase
    Product package and two tubes of reagents
    Platinum Multiplex PCR Master Mix
    No. of amplicons in single reaction Up to 15-plexUp to 20-plex
    Amplification lengthUp to 2.5 kbUp to 2.5 kb
    Hot start for enhanced specificityYesYes
    Fidelity vs. Taq enzyme>300x1x
    Universal annealing temperature of 60°CYesNo
    GC-rich amplificationYes
    No
    Stand-alone enzymeColorless
    Order now
    N/A
    Master mix format
    Colorless
    Order now
    Green*
    Order now
    Colorless
    Order now
    *Contains electrophoresis tracking dyes and a density reagent for direct gel loading of PCR products.
    GC-rich PCR
    GC-rich PCR is amplification of DNA fragments with high GC content. GC-rich regions of DNA often form stable secondary structures, such as hairpins or G-quadruplexes, that pose a challenge to efficient DNA amplification. Our enzymes are designed to amplify DNA sequences with >65% GC content.
    Product photo of package and three tubes of reagents.
    Platinum SuperFi II DNA Polymerase
    product package and three tubes of reagents
    Platinum II Taq DNA Polymerase
    Fidelity vs. Taq enzyme>300x1x
    Hot start for enhanced specificityYesYes
    Efficient amplification of >65% GC sequencesYes
    Yes
    Universal annealing temperature of 60°CYesYes
    Speed15–30 sec/kb15 sec/kb
    Amplification lengthUp to 20 kbUp to 5 kb
    Amplicon overhangsBlunt3′ A
    Stand-alone enzymeColorless
    Order now
    Colorless
    Order now
    Master mix format
    Colorless
    Order now
    Green*
    Order now
    Colorless
    Order now
    Green*
    Order now
    *Contains electrophoresis tracking dyes and a density reagent for direct gel loading of PCR products.
    Direct PCR
    Direct PCR is a streamlined technique that allows for DNA amplification directly from various biological samples without DNA extraction or purification steps.
    Product photo of package, four tubes of reagents, and one vial of lysis buffer
    Platinum Direct PCR Universal Master Mix
    Works across samples of various originsYes
    Universal annealing temperature of 60°CYes
    Hot start for enhanced specificityYes
    Fidelity vs. Taq enzyme1x
    GC-rich amplificationYes
    Amplification length Up to 8 kb*
    Speed20 sec/kb
    Stand-alone enzymeN/A
    Master mix formatGreen**
    Order now
    *Using the lysis protocol.
    **Contains electrophoresis tracking dyes and a density reagent for direct gel loading of PCR products.
    Application note Detection of target DNA in cell samples by direct PCR
    Hot-start PCR
    Hot-start PCR is a type of PCR where DNA polymerase activity is inhibited until the initial denaturation step. This inhibition is achieved through various methods, such as antibodies, affibodies, or chemical modifications of the polymerase. The benefits of hot-start PCR include increased specificity and yield.
    product package and three tubes of reagents
    Platinum II Taq Hot-Start DNA Polymerase
    product package and four tubes of reagents
    AmpliTaq Gold 360 DNA Polymerase
    Hot-start technologyAntibodyChemical
    Enzyme activation time2 min10 min
    Universal annealing temperature of 60°CYesNo
    DNA synthesis speed15 sec/kb60 sec/kb
    Flexible extension step*YesNo
    Inhibitor toleranceYesNo
    Amplification lengthUp to 5 kbUp to 5 kb
    GC-rich formatYesYes
    Stand-alone enzymeColorless†
    Order now
    Colorless
    Order now
    Master mix format
    Colorless
    Order now
    Green**
    Order now
    Colorless
    Order now
    *The extension step can be extended up to 60 sec/kb without the effect of specificity.
    **Contains electrophoresis tracking dyes and a density reagent for direct gel loading of PCR products.
    Green buffer available as separate item for use with stand-alone enzyme.
    Note: The original Platinum Taq DNA Polymerase is available in the formats of stand alone, stand alone with gel-loading dyes, master mix, and master mix with gel-loading dyes.
    High-fidelity PCR
    High fidelity PCR, also known as proofreading PCR, is a technique that utilizes DNA polymerases with a built-in proofreading capability to ensure accurate sequence amplification with no errors introduced in the sequence. 
    Product package and three tubes of reagents
    Platinum SuperFi II DNA Polymerase
    Product package and three tubes of reagents
    Platinum Taq DNA Polymerase High Fidelity
    Fidelity vs. Taq enzyme>300x6x
    Hot start for enhanced specificityYesYes
    Universal annealing temperature of 60°CYesNo
    Amplification lengthUp to 20 kb*Up to 20 kb
    Amplicon overhangsBlunt3′ A/Blunt
    GC-rich amplificationYes
    No
    Stand-alone enzymeColorless
    Order now
    Colorless
    Order now
    Master mix format
    Colorless
    Order now
    Green**
    Order now
    Colorless
    Order now
    *Amplification of >20 kb fragment sizes is possible (up to 40 kb), but may require additional optimization of reaction conditions and primer design.
    **Contains electrophoresis tracking dyes and a density reagent for direct gel loading of PCR products.
    Long-range PCR
    Long-range PCR is a technique used to amplify DNA fragments that are longer than 5 kb, which is larger than the typical limit of standard PCR. It involves modifying the PCR conditions and utilizing specialized DNA polymerases to overcome the challenges associated with amplifying longer DNA sequences. 
    Product photo of package and three tubes of reagents.
    Platinum SuperFi II DNA Polymerase
    Amplification lengthUp to 20 kb*
    Fidelity vs. Taq enzyme>300x
    Hot start for enhanced specificityYes
    Universal annealing temperature of 60°CYes
    GC-rich amplificationYes
    DNA synthesis speed15–30 sec/kb
    Stand-alone enzymeColorless
    Order now
    Master mix format
    Colorless
    Order now
    Green**
    Order now
    *Amplification of >20 kb fragment sizes is possible (up to 40 kb), but may require additional optimization of reaction conditions and primer design.
    **Contains electrophoresis tracking dyes and a density reagent for direct gel loading of PCR products.
    White paper Assembly of large PCR amplicons by the Gibson Assembly method
    Multiplex PCR
    Multiplex PCR is a technique that allows for the concurrent amplification of multiple target DNA sequences within a single PCR reaction. It involves the use of multiple primer sets, each specific to a different target sequence, but amplified in a single reaction mixture.
    Product package and three tubes of reagents
    Platinum SuperFi II DNA Polymerase
    Product package and two tubes of reagents
    Platinum Multiplex PCR Master Mix
    No. of amplicons in single reaction Up to 15-plexUp to 20-plex
    Amplification lengthUp to 2.5 kbUp to 2.5 kb
    Hot start for enhanced specificityYesYes
    Fidelity vs. Taq enzyme>300x1x
    Universal annealing temperature of 60°CYesNo
    GC-rich amplificationYes
    No
    Stand-alone enzymeColorless
    Order now
    N/A
    Master mix format
    Colorless
    Order now
    Green*
    Order now
    Colorless
    Order now
    *Contains electrophoresis tracking dyes and a density reagent for direct gel loading of PCR products.
    GC-rich PCR
    GC-rich PCR is amplification of DNA fragments with high GC content. GC-rich regions of DNA often form stable secondary structures, such as hairpins or G-quadruplexes, that pose a challenge to efficient DNA amplification. Our enzymes are designed to amplify DNA sequences with >65% GC content.
    Product photo of package and three tubes of reagents.
    Platinum SuperFi II DNA Polymerase
    product package and three tubes of reagents
    Platinum II Taq DNA Polymerase
    Fidelity vs. Taq enzyme>300x1x
    Hot start for enhanced specificityYesYes
    Efficient amplification of >65% GC sequencesYes
    Yes
    Universal annealing temperature of 60°CYesYes
    Speed15–30 sec/kb15 sec/kb
    Amplification lengthUp to 20 kbUp to 5 kb
    Amplicon overhangsBlunt3′ A
    Stand-alone enzymeColorless
    Order now
    Colorless
    Order now
    Master mix format
    Colorless
    Order now
    Green*
    Order now
    Colorless
    Order now
    Green*
    Order now
    *Contains electrophoresis tracking dyes and a density reagent for direct gel loading of PCR products.
    Direct PCR
    Direct PCR is a streamlined technique that allows for DNA amplification directly from various biological samples without DNA extraction or purification steps.
    Product photo of package, four tubes of reagents, and one vial of lysis buffer
    Platinum Direct PCR Universal Master Mix
    Works across samples of various originsYes
    Universal annealing temperature of 60°CYes
    Hot start for enhanced specificityYes
    Fidelity vs. Taq enzyme1x
    GC-rich amplificationYes
    Amplification length Up to 8 kb*
    Speed20 sec/kb
    Stand-alone enzymeN/A
    Master mix formatGreen**
    Order now
    *Using the lysis protocol.
    **Contains electrophoresis tracking dyes and a density reagent for direct gel loading of PCR products.
    Application note Detection of target DNA in cell samples by direct PCR
    Taq DNA polymerases: Native and recombinant enzymes
    Native DNA polymerases are naturally occurring enzymes found in various organisms such as bacteria and archaea. Taq DNA polymerase is one of the best-known thermostable DNA polymerases used in PCR amplification of DNA targets. The native enzyme is purified from Thermus aquaticus YT1. The native Taq DNA polymerase is often preferred for amplification of bacterial DNA sequences homologous to E. coli sequences. The recombinant enzyme is a cloned version of native Taq polymerases with improved properties and functionalities.
    Platinum DNA polymerases for excellent results
    The new generation of Platinum DNA polymerases come with novel buffers, enabling primer annealing at a universal temperature of 60°C. A combination of the innovative buffers, high-performing enzymes, and reliable hot-start technology offers exceptional PCR results even for challenging applications.
    Benefits of platinum PCR enzymes
    • Enhanced PCR specificity—Antibody-mediated hot-start enzymes with Platinum technology
    • Circumvent multiple PCR runs—Ability to co-cycle different PCR assays
    • Detecting multiple targets from one template—Ability to multiplex up to 15 targets
    • Fewer steps for pipetting and reagent setup—Necessary reaction components and direct gel-loading dyes included in the green master mixes
    • Simplify PCR optimization steps—Protocol with a universal annealing temperature
    • Robust PCR with difficult targets—High tolerance to inhibitor-containing samples and efficient amplification of GC-rich sequences
    • Enabling automation or robotic setup—Assembled reactions stable on the benchtop for 8–24 hours
    Education icon
     PCR enzymes education resources
    Access PCR learning resources—a knowledge base of all things PCR.
    Webinar icon
    Molecular biology webinars
    View our webinars for current date to learn more about molecular biology topics.
    Video icon
    PCR enzymes videos
    Discover product features and tips to optimize your PCR experiments.
    Tools icon
    Support center
    Find tips, troubleshooting help, and resources for your end-point PCR and cDNA applications.
    Computer mouse icon
    Contact us
    Email or call our Technical Application Scientists for questions regarding PCR enzymes and master mixes.
    Check icon
    OEM and custom PCR products
    Discover custom manufacturing solutions, product labeling, and packaging capabilities tailored to your specifications.
    Stylesheet for Classic Wide Template adjustments
    For Research Use Only. Not for use in diagnostic procedures.