In 1903, German chemist Bernhard Tollens observed that the reaction between acetophenone and formaldehyde in the presence of ammonium chloride led to the formation of a tertiary amine. In 1917, German chemist Carl Mannich also prepared a tertiary amine from antipyrine using the same conditions, and recognized that this reaction was general. Since then, the condensation of a CH-activated compound such as an aldehyde or ketone with a primary or secondary amine or ammonia and a non-enolizable aldehyde or ketone to prepare aminoalkylated derivatives has come to be known as the Mannich reaction.
The product of this reaction is a substituted beta-amino carbonyl compound which is often known as a Mannich base. Mannich bases are useful intermediates for synthesis since they can undergo a variety of transformations. These can include beta-elimination to afford alpha- or beta-unsaturated carbonyl compounds (Michael acceptors), reaction with organolithium or Grignard reagents to produce beta-amino alcohols, or even substitution of the dialkylamino group with nucleophiles to create functionalized carbonyl compounds. One of the best-known applications of the Mannich reaction is its use in conjunction with an aza-Cope rearrangement to generate heterocycles.
Mechanism of the Mannich reaction
Review available Thermo Scientific products for the Mannich reaction:
Catalog # Name Size Price (USD) Qty
A16163.0F Formaldehyde, 37% w/w aq. soln., stab. with 7-8% methanol Each
52.65
Online Exclusive
58.00
Save 5.35 (9%)
A12727.AP Acetophenone, 99% Each
44.65
Special offer
Online exclusive
51.90
Save 7.25 (14%)
A15000.30 Ammonium chloride, 98+% Each
31.65
Special offer
Online exclusive
36.90
Save 5.25 (14%)
A12727.0F Acetophenone, 99% Each
110.65
Online Exclusive
123.00
Save 12.35 (10%)
A16163.AP Formaldehyde, 37% w/w aq. soln., stab. with 7-8% methanol Each
27.65
Special offer
Online exclusive
32.10
Save 4.45 (14%)
A12727.0D Acetophenone, 99% Each
256.65
Online Exclusive
285.00
Save 28.35 (10%)
A15000.0B Ammonium chloride, 98+% Each
41.65
Special offer
Online exclusive
48.50
Save 6.85 (14%)
A15000.0I Ammonium chloride, 98+% Each
127.65
Special offer
Online exclusive
150.00
Save 22.35 (15%)
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers. Do you have a favorite reaction? Let us know!
Vote now