In-the-Lab_BXP62049m
Proper controls are essential to ensure success in every RNAi experiment. We offer a selection of RNAi technologies to ensure that researchers carry out efficient, reproducible, high-quality gene silencing experiments. Our siRNA controls allow you to:
  • Determine the role of non-specific cellular responses in your phenotype
  • Achieve greater knockdown by optimizing transfection conditions
  • Ensure ongoing experimental success
Recommended experimental controls
It is recommended to use controls from the same product line as your experimental siRNA (e.g., Silencer Select siRNA) to control for the effects of chemical modifications. Experiments with unmodified custom siRNA should use Silencer siRNA controls.
Type of controlRecommended use
Negative controlsNon-targeting siRNAs control for non-specific effects related to siRNA delivery to provide a baseline for target gene silencing
Positive controlsValidated siRNAs to achieve high levels of knockdown to ensure optimal transfection conditions. They typically target a constitutively expressed gene for use in a wide range of cell types.
Fluorescent transfection controlsAssess and monitor transfection efficiency with a fluorescently labeled siRNA control
Additional RNAi experimental guidelinesSee below for guidance on experimental best practice; use of multiple siRNA sequences to the same target, titration of siRNA, use of untransfected controls, and downstream controls.
Negative controls
Negative control siRNAs are most often a non-targeting siRNA - designed not to target any gene - for determining the non-specific effects of siRNA delivery and for providing a baseline to compare to siRNA-treated samples. Negative controls may also be a functional siRNA that is known to have no impact on your phenotype of interest, which should be determined empirically.
We recommend using one or more negative controls in every RNAi experiment. You should match the control siRNA to the experimental siRNA type (for example, use only Silencer Select siRNA controls for experiments using Silencer Select siRNA).
Positive controls
Positive controls provide confidence in your RNAi experiments by confirming that experimental conditions were met to achieve robust silencing. Positive controls are validated siRNAs that are known to achieve high levels of knockdown (>70%), typically for a constitutively expressed or housekeeping gene. A positive control should be used to optimize siRNA delivery conditions and to reconfirm high levels of delivery in each RNAi experiment. When a positive control fails to produce the anticipated phenotype, carefully evaluate your experimental conditions and decide if some factors need to be adjusted.
You should match the control siRNA to the experimental siRNA type (for example, use only Silencer Select siRNA controls for experiments using Silencer Select siRNA).
Fluorescently labeled transfection controls
To achieve the highest transfection efficiency possible, it is advised to first optimize transfection conditions for your cell lines. Fluorescently labeled RNAs can help with this by providing a qualitative, visual indicator of transfection success. Keep in mind that it is also important to monitor transfection in each experiment with a validated positive control siRNA for knockdown.
For optimal siRNA transfection, we have many cell type-specific transfection protocols for siRNA delivery using Lipofectamine RNAiMAX Transfection Reagent to help you get started.
Additional RNAi experimental guidelines
In addition to the proper use of controls, these guidelines outline experimental best-practices for RNAi gene silencing.
Correlate phenotypes with reduced target mRNA levels
A phenotypic assay is often used as an indicator of a successful RNAi-mediated loss-of-function experiment. However, an observed change in your cells due to target gene silencing should be correlated with the loss of the corresponding mRNA levels using qRT-PCR. If one or more of the gene-targeting siRNAs gives a phenotype but not a loss in mRNA, then the phenotype could be due to a non-specific response or off-target gene silencing.
Use multiple siRNA sequences to the same target
siRNA sequences with partial homology to other targets may contribute to off-target activity. Gene profiling experiments have shown that duplexes with partial homology to other transcripts can cleave the target or act like a microRNA (miRNA), inhibiting translation of the target mRNA. To ensure that knockdown of the intended gene can be attributed to the observed phenotype, the results should be confirmed by at least two unique siRNA reagents that target non-overlapping regions of the target mRNA.
Titration of siRNA
Silencer Select siRNA and Stealth RNAi siRNA can be very effective even at low concentrations, so you should aim to use the lowest effective level to avoid altering the cells’ normal processes. Titrating down the dose of the siRNA enables you to reduce off-target or nonspecific effects while achieving robust knockdown.
Untransfected controls
Cells that are not transfected or treated with siRNA are an important control for the overall reproducibility of your experiment. These cell populations control for the impact of media changes,reactivity to assay conditions, or other variables, while providing a secondary baseline for negative/non-targeting controls in determination of cell viability.
Downstream detection assays
Before transfecting cells with siRNA, we recommend validating the reagents that will be used in qRT-PCR and western blots to measure mRNA and protein levels. Validating qRT-PCR primers or antibodies for your positive control and target genes before performing knockdown experiments ensures that these reagents are sensitive enough to detect changes in your target gene’s expression due to knockdown. Without sufficient sensitivity, it can be difficult to interpret knockdown results from genes or proteins with low expression levels.
Silencer Select negative controls
  • Non-targeting controls with no significant sequence similarity to mouse, rat, or human gene sequences
  • Chemically modified for reduced off-target effects as found in other Silencer Select siRNAs
  • Tested by microarray analysis and shown to have minimal effects on gene expression
  • Assessed in multiparametric cell-based assays and are proven to have no significant effect on cell proliferation, viability, or morphology in the cell lines tested.
Silencer Select siRNA positive controls
  • Extensively validated for optimizing and monitoring transfection efficiency in human, mouse, and rat cells.
  • Functionally tested in several common cell lines
  • Include Silencer Select siRNA modifications for enhanced specificity
Silencer Select controls—order now
Catalog # Name Size Price (USD) Qty
4390843 Silencer™ Select Negative Control No. 1 siRNA, 5 nmol Each
269.65
Online Exclusive
282.00
Save 12.35 (4%)
4390844 Silencer™ Select Negative Control No. 1 siRNA, 40 nmol Each
748.65
Online Exclusive
784.00
Save 35.35 (5%)
4404020 Silencer™ Select Negative Control No. 1 siRNA, in vivo ready Each
3,205.00
4390846 Silencer™ Select Negative Control No. 2 siRNA, 5 nmol Each
271.65
Online Exclusive
282.00
Save 10.35 (4%)
4390847 Silencer™ Select Negative Control No. 2 siRNA, 40 nmol Each
759.65
Online Exclusive
790.00
Save 30.35 (4%)
4390849 Silencer™ Select GAPDH Positive Control siRNA, 5 nmol Each
264.65
Online Exclusive
282.00
Save 17.35 (6%)
4390850 Silencer™ Select GAPDH Positive Control siRNA, 40 nmol Each
736.65
Online Exclusive
786.00
Save 49.35 (6%)
4455877 Silencer™ Select MALAT1 Positive Control siRNA Each
285.00
Stealth RNAi siRNA negative controls
  • Negative controls that offer low, medium, and high levels of GC content to match that of the experimental Stealth RNAi siRNAs
  • Medium GC is recommended for experiments using numerous unique siRNA designs
Stealth RNAi siRNA positive controls
  • Validated reagents targeting housekeeping genes or common reporter genes
  • Ideal as a control for assessing knockdown and optimizing RNAi experiments
Stealth RNAi controls—order now
Silencer siRNA negative controls
  • No significant sequence similarity to mouse, rat, or human gene sequences
  • Tested in cell-based screens and proven to have no significant effect on cell proliferation, viability, or morphology.
  • Recommended for use in experiments with any Silencer siRNA or unmodified siRNA (e.g. custom synthesis)
Silencer siRNA positive controls
  • Validated siRNA controls for optimizing siRNA experiments
  • Functionally tested in several common cell lines
  • Recommended for use in experiments with unmodified siRNA (e.g. custom synthesis)
Silencer controls—order now
Catalog # Name Size Price (USD) Qty
AM4611 Silencer™ Negative Control No. 1 siRNA, 5 nmol Each
203.65
Online Exclusive
214.00
Save 10.35 (5%)
AM4635 Silencer™ Negative Control No. 1 siRNA, 40 nmol Each
597.65
Online Exclusive
630.00
Save 32.35 (5%)
AM4636 Silencer™ Negative Control No. 1 siRNA, 5 x 40 nmol Each
1,238.65
Online Exclusive
1,304.00
Save 65.35 (5%)
4404021 Silencer™ Negative Control No. 1 siRNA Each
2,570.00
AM4613 Silencer™ Negative Control No. 2 siRNA, 5 nmol Each
203.65
Online Exclusive
214.00
Save 10.35 (5%)
AM4637 Silencer™ Negative Control No. 2 siRNA, 40 nmol Each
592.65
Online Exclusive
624.00
Save 31.35 (5%)
AM4615 Silencer™ Negative Control No. 3 siRNA Each
213.00
AM4641 Silencer™ Negative Control No. 4 siRNA Each
206.65
Online Exclusive
213.00
Save 6.35 (3%)
AM4642 Silencer™ Negative Control No. 5 siRNA Each
213.00
AM4624 Silencer™ GAPDH siRNA (human, mouse, rat), 5 nmol Each
253.65
Online Exclusive
261.00
Save 7.35 (3%)
AM4631 Silencer™ GAPDH siRNA (human, mouse, rat), 40 nmol Each
599.65
Online Exclusive
618.00
Save 18.35 (3%)
AM4632 Silencer™ GAPDH siRNA (human, mouse, rat), 5 x 40 nmol Each
1,257.65
Online Exclusive
1,296.00
Save 38.35 (3%)
AM4633 Silencer™ GAPDH siRNA (human), 40 nmol Each
618.00
AM4605 Silencer™ GAPDH siRNA (human), 5 nmol Each
261.00
AM4626 Silencer™ GFP (eGFP) siRNA Each
253.65
Online Exclusive
261.00
Save 7.35 (3%)
AM4629 Silencer™ Firefly Luciferase (GL2 + GL3) siRNA Each
261.00
AM4639 Silencer™ KIF11 (Eg5) siRNA (human, mouse, rat) Each
251.00
AM4620 Silencer™ FAM-labeled Negative Control No. 1 siRNA Each
241.65
Online Exclusive
255.00
Save 13.35 (5%)
AM4621 Silencer™ Cy™3-labeled Negative Control No. 1 siRNA Each
289.65
Online Exclusive
307.00
Save 17.35 (6%)
AM4649 Silencer™ Cy™3-labeled GAPDH siRNA (human, mouse, rat) Each
296.00
AM4650 Silencer™ FAM-labeled GAPDH siRNA (human, mouse, rat) Each
235.65
Online Exclusive
243.00
Save 7.35 (3%)
Fluorescently labeled transfection controls
An siRNA or oligo with a fluorophore conjugate, such as FAM or Cy3, can be useful as a visual indicator of transfection success, especially during transfection optimization when conditions such as cell density and the amount of transfection reagent are being varied.It should be noted that intracellular fluorescence is not a substitute for quantitation of transfection efficiency as measured by knockdown from a validated positive control.
Transfection controls—order now
Catalog # Name Size Price (USD) Qty
AM4620 Silencer™ FAM-labeled Negative Control No. 1 siRNA Each
241.65
Online Exclusive
255.00
Save 13.35 (5%)
AM4621 Silencer™ Cy™3-labeled Negative Control No. 1 siRNA Each
289.65
Online Exclusive
307.00
Save 17.35 (6%)
AM4649 Silencer™ Cy™3-labeled GAPDH siRNA (human, mouse, rat) Each
296.00
AM4650 Silencer™ FAM-labeled GAPDH siRNA (human, mouse, rat) Each
235.65
Online Exclusive
243.00
Save 7.35 (3%)
14750100 BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control Each
470.65
Online Exclusive
500.00
Save 29.35 (6%)
13750062 BLOCK-iT™ Fluorescent Oligo, for electroporation Each
1,180.65
Online Exclusive
1,258.00
Save 77.35 (6%)
Negative controls
Negative control siRNAs are most often a non-targeting siRNA - designed not to target any gene - for determining the non-specific effects of siRNA delivery and for providing a baseline to compare to siRNA-treated samples. Negative controls may also be a functional siRNA that is known to have no impact on your phenotype of interest, which should be determined empirically.
We recommend using one or more negative controls in every RNAi experiment. You should match the control siRNA to the experimental siRNA type (for example, use only Silencer Select siRNA controls for experiments using Silencer Select siRNA).
Positive controls
Positive controls provide confidence in your RNAi experiments by confirming that experimental conditions were met to achieve robust silencing. Positive controls are validated siRNAs that are known to achieve high levels of knockdown (>70%), typically for a constitutively expressed or housekeeping gene. A positive control should be used to optimize siRNA delivery conditions and to reconfirm high levels of delivery in each RNAi experiment. When a positive control fails to produce the anticipated phenotype, carefully evaluate your experimental conditions and decide if some factors need to be adjusted.
You should match the control siRNA to the experimental siRNA type (for example, use only Silencer Select siRNA controls for experiments using Silencer Select siRNA).
Fluorescently labeled transfection controls
To achieve the highest transfection efficiency possible, it is advised to first optimize transfection conditions for your cell lines. Fluorescently labeled RNAs can help with this by providing a qualitative, visual indicator of transfection success. Keep in mind that it is also important to monitor transfection in each experiment with a validated positive control siRNA for knockdown.
For optimal siRNA transfection, we have many cell type-specific transfection protocols for siRNA delivery using Lipofectamine RNAiMAX Transfection Reagent to help you get started.
Additional RNAi experimental guidelines
In addition to the proper use of controls, these guidelines outline experimental best-practices for RNAi gene silencing.
Correlate phenotypes with reduced target mRNA levels
A phenotypic assay is often used as an indicator of a successful RNAi-mediated loss-of-function experiment. However, an observed change in your cells due to target gene silencing should be correlated with the loss of the corresponding mRNA levels using qRT-PCR. If one or more of the gene-targeting siRNAs gives a phenotype but not a loss in mRNA, then the phenotype could be due to a non-specific response or off-target gene silencing.
Use multiple siRNA sequences to the same target
siRNA sequences with partial homology to other targets may contribute to off-target activity. Gene profiling experiments have shown that duplexes with partial homology to other transcripts can cleave the target or act like a microRNA (miRNA), inhibiting translation of the target mRNA. To ensure that knockdown of the intended gene can be attributed to the observed phenotype, the results should be confirmed by at least two unique siRNA reagents that target non-overlapping regions of the target mRNA.
Titration of siRNA
Silencer Select siRNA and Stealth RNAi siRNA can be very effective even at low concentrations, so you should aim to use the lowest effective level to avoid altering the cells’ normal processes. Titrating down the dose of the siRNA enables you to reduce off-target or nonspecific effects while achieving robust knockdown.
Untransfected controls
Cells that are not transfected or treated with siRNA are an important control for the overall reproducibility of your experiment. These cell populations control for the impact of media changes,reactivity to assay conditions, or other variables, while providing a secondary baseline for negative/non-targeting controls in determination of cell viability.
Downstream detection assays
Before transfecting cells with siRNA, we recommend validating the reagents that will be used in qRT-PCR and western blots to measure mRNA and protein levels. Validating qRT-PCR primers or antibodies for your positive control and target genes before performing knockdown experiments ensures that these reagents are sensitive enough to detect changes in your target gene’s expression due to knockdown. Without sufficient sensitivity, it can be difficult to interpret knockdown results from genes or proteins with low expression levels.
Silencer Select negative controls
  • Non-targeting controls with no significant sequence similarity to mouse, rat, or human gene sequences
  • Chemically modified for reduced off-target effects as found in other Silencer Select siRNAs
  • Tested by microarray analysis and shown to have minimal effects on gene expression
  • Assessed in multiparametric cell-based assays and are proven to have no significant effect on cell proliferation, viability, or morphology in the cell lines tested.
Silencer Select siRNA positive controls
  • Extensively validated for optimizing and monitoring transfection efficiency in human, mouse, and rat cells.
  • Functionally tested in several common cell lines
  • Include Silencer Select siRNA modifications for enhanced specificity
Silencer Select controls—order now
Catalog # Name Size Price (USD) Qty
4390843 Silencer™ Select Negative Control No. 1 siRNA, 5 nmol Each
269.65
Online Exclusive
282.00
Save 12.35 (4%)
4390844 Silencer™ Select Negative Control No. 1 siRNA, 40 nmol Each
748.65
Online Exclusive
784.00
Save 35.35 (5%)
4404020 Silencer™ Select Negative Control No. 1 siRNA, in vivo ready Each
3,205.00
4390846 Silencer™ Select Negative Control No. 2 siRNA, 5 nmol Each
271.65
Online Exclusive
282.00
Save 10.35 (4%)
4390847 Silencer™ Select Negative Control No. 2 siRNA, 40 nmol Each
759.65
Online Exclusive
790.00
Save 30.35 (4%)
4390849 Silencer™ Select GAPDH Positive Control siRNA, 5 nmol Each
264.65
Online Exclusive
282.00
Save 17.35 (6%)
4390850 Silencer™ Select GAPDH Positive Control siRNA, 40 nmol Each
736.65
Online Exclusive
786.00
Save 49.35 (6%)
4455877 Silencer™ Select MALAT1 Positive Control siRNA Each
285.00
Stealth RNAi siRNA negative controls
  • Negative controls that offer low, medium, and high levels of GC content to match that of the experimental Stealth RNAi siRNAs
  • Medium GC is recommended for experiments using numerous unique siRNA designs
Stealth RNAi siRNA positive controls
  • Validated reagents targeting housekeeping genes or common reporter genes
  • Ideal as a control for assessing knockdown and optimizing RNAi experiments
Stealth RNAi controls—order now
Silencer siRNA negative controls
  • No significant sequence similarity to mouse, rat, or human gene sequences
  • Tested in cell-based screens and proven to have no significant effect on cell proliferation, viability, or morphology.
  • Recommended for use in experiments with any Silencer siRNA or unmodified siRNA (e.g. custom synthesis)
Silencer siRNA positive controls
  • Validated siRNA controls for optimizing siRNA experiments
  • Functionally tested in several common cell lines
  • Recommended for use in experiments with unmodified siRNA (e.g. custom synthesis)
Silencer controls—order now
Catalog # Name Size Price (USD) Qty
AM4611 Silencer™ Negative Control No. 1 siRNA, 5 nmol Each
203.65
Online Exclusive
214.00
Save 10.35 (5%)
AM4635 Silencer™ Negative Control No. 1 siRNA, 40 nmol Each
597.65
Online Exclusive
630.00
Save 32.35 (5%)
AM4636 Silencer™ Negative Control No. 1 siRNA, 5 x 40 nmol Each
1,238.65
Online Exclusive
1,304.00
Save 65.35 (5%)
4404021 Silencer™ Negative Control No. 1 siRNA Each
2,570.00
AM4613 Silencer™ Negative Control No. 2 siRNA, 5 nmol Each
203.65
Online Exclusive
214.00
Save 10.35 (5%)
AM4637 Silencer™ Negative Control No. 2 siRNA, 40 nmol Each
592.65
Online Exclusive
624.00
Save 31.35 (5%)
AM4615 Silencer™ Negative Control No. 3 siRNA Each
213.00
AM4641 Silencer™ Negative Control No. 4 siRNA Each
206.65
Online Exclusive
213.00
Save 6.35 (3%)
AM4642 Silencer™ Negative Control No. 5 siRNA Each
213.00
AM4624 Silencer™ GAPDH siRNA (human, mouse, rat), 5 nmol Each
253.65
Online Exclusive
261.00
Save 7.35 (3%)
AM4631 Silencer™ GAPDH siRNA (human, mouse, rat), 40 nmol Each
599.65
Online Exclusive
618.00
Save 18.35 (3%)
AM4632 Silencer™ GAPDH siRNA (human, mouse, rat), 5 x 40 nmol Each
1,257.65
Online Exclusive
1,296.00
Save 38.35 (3%)
AM4633 Silencer™ GAPDH siRNA (human), 40 nmol Each
618.00
AM4605 Silencer™ GAPDH siRNA (human), 5 nmol Each
261.00
AM4626 Silencer™ GFP (eGFP) siRNA Each
253.65
Online Exclusive
261.00
Save 7.35 (3%)
AM4629 Silencer™ Firefly Luciferase (GL2 + GL3) siRNA Each
261.00
AM4639 Silencer™ KIF11 (Eg5) siRNA (human, mouse, rat) Each
251.00
AM4620 Silencer™ FAM-labeled Negative Control No. 1 siRNA Each
241.65
Online Exclusive
255.00
Save 13.35 (5%)
AM4621 Silencer™ Cy™3-labeled Negative Control No. 1 siRNA Each
289.65
Online Exclusive
307.00
Save 17.35 (6%)
AM4649 Silencer™ Cy™3-labeled GAPDH siRNA (human, mouse, rat) Each
296.00
AM4650 Silencer™ FAM-labeled GAPDH siRNA (human, mouse, rat) Each
235.65
Online Exclusive
243.00
Save 7.35 (3%)
Fluorescently labeled transfection controls
An siRNA or oligo with a fluorophore conjugate, such as FAM or Cy3, can be useful as a visual indicator of transfection success, especially during transfection optimization when conditions such as cell density and the amount of transfection reagent are being varied.It should be noted that intracellular fluorescence is not a substitute for quantitation of transfection efficiency as measured by knockdown from a validated positive control.
Transfection controls—order now
Catalog # Name Size Price (USD) Qty
AM4620 Silencer™ FAM-labeled Negative Control No. 1 siRNA Each
241.65
Online Exclusive
255.00
Save 13.35 (5%)
AM4621 Silencer™ Cy™3-labeled Negative Control No. 1 siRNA Each
289.65
Online Exclusive
307.00
Save 17.35 (6%)
AM4649 Silencer™ Cy™3-labeled GAPDH siRNA (human, mouse, rat) Each
296.00
AM4650 Silencer™ FAM-labeled GAPDH siRNA (human, mouse, rat) Each
235.65
Online Exclusive
243.00
Save 7.35 (3%)
14750100 BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control Each
470.65
Online Exclusive
500.00
Save 29.35 (6%)
13750062 BLOCK-iT™ Fluorescent Oligo, for electroporation Each
1,180.65
Online Exclusive
1,258.00
Save 77.35 (6%)
Shop for everyday lab essentials to power your transfection steps
Transfection lab essentials
We've made it easier for you to quickly find and buy products for every step in your transfection workflow.
Shop now
Questions?
Technical inquires:
Our Technical Application Scientists are available to help assist you at techsupport@thermofisher.com
Ordering & Order Status inquires:
If you have questions about pre-designed RNAi orders and order status, please contact us at genomicorders@thermofisher.com
If you have any questions about Custom RNAi orders and order status, please contact us at RNAiSupport@thermofisher.com
For Research Use Only. Not for use in diagnostic procedures.