Introduction
The BLOCK-iT™ Dicer RNAi Transfection Kit and the BLOCK-iT™ Complete Dicer RNAi Kit facilitate generation of purified diced siRNA duplexes (d-siRNA) that are suitable for use in RNAi analysis of a target gene in mammalian cells. The kits contain the BLOCK-iT™ Dicer Enzyme for dicing dsRNA, reagents to purify the d-siRNA, and an optimized transfection reagent for highly efficient delivery of d-siRNA to mammalian cells.
Note:   The BLOCK-iT™ Complete Dicer RNAi Kit also includes the BLOCK-iT™ RNAi TOPO® Transcription Kit to facilitate high-yield generation of purified dsRNA. For more information, refer to the BLOCK-iT™ RNAi TOPO® Transcription Kit manual. This manual is supplied with the BLOCK-iT™ Complete Dicer RNAi Kit, but is also available for downloading from www.invitrogen.com or by contacting Technical Service (page 30).
Advantages of the BLOCK-iT™ Dicer RNAi Transfection Kit
Using the BLOCK-iT™ Dicer RNAi Transfection Kit and the BLOCK-iT™ Complete Dicer RNAi Kit to generate d-siRNA for RNAi analysis in mammalian provides the following advantages:
  • Provides a cost-effective means to enzymatically generate a pool of d-siRNA that cover a larger portion of the target gene (e.g. 500 bp to 1 kb) without the need for expensive chemical synthesis of siRNA.
  • Provides the BLOCK-iT™ Dicer Enzyme and an optimized protocol to facilitate generation of the highest yields of d-siRNA from a dsRNA substrate.
  • Includes BLOCK-iT™ RNAi Purification reagents for efficient purification of d-siRNA. Purified d-siRNA can be quantitated, enabling highly reproducible RNAi analysis.
  • Includes the Lipofectamine® 2000 Reagent for the highest efficiency transfection in a wide variety of mammalian cell lines.
Purpose of this Protocol
  • A description of the components in the BLOCK-iT™ Dicer RNAi Transfection Kit and an overview of the pathway by which d-siRNA facilitates gene knockdown in mammalian cells.
  • Guidelines to produce dsRNA corresponding to the target gene. For detailed instructions to produce dsRNA, refer to the BLOCK-iT™ RNAi TOPO® Transcription Kit manual.
  • Guidelines and instructions to use the BLOCK-iT™ Dicer Enzyme to cleave dsRNA to generate a complex pool of d-siRNA.
  • Instructions to purify d-siRNA.
  • Guidelines and instructions to transfect purified d-siRNA into mammalian cells using Lipofectamine® 2000 Reagent for RNAi studies.
The RNAi Pathway and How Dicer Works
The RNAi Pathway
RNAi describes the phenomenon by which dsRNA induces potent and specific inhibition of eukaryotic gene expression via the degradation of complementary messenger RNA (mRNA), and is functionally similar to the processes of post-transcriptional gene silencing (PTGS) or cosuppression in plants (Cogoni et al., 1994; Napoli et al., 1990; Smith et al., 1990; van der Krol et al., 1990) and quelling in fungi (Cogoni and Macino, 1999; Cogoni and Macino, 1997; Romano and Macino, 1992). In plants, the PTGS response is thought to occur as a natural defense against viral infection or transposon insertion (Anandalakshmi et al., 1998; Jones et al., 1998; Li and Ding, 2001; Voinnet et al., 1999).
In eukaryotic organisms, dsRNA produced in vivo or introduced by pathogens is processed into 21-23 nucleotide double-stranded short interfering RNA duplexes (siRNA) by an enzyme called Dicer, a member of the RNase III family of double-stranded RNA-specific endonucleases (Bernstein et al., 2001; Ketting et al., 2001). Each siRNA then incorporates into an RNA-induced silencing complex (RISC), an enzyme complex that serves to target cellular transcripts complementary to the siRNA for specific cleavage and degradation (Hammond et al., 2000; Nykanen et al., 2001).
For more information about the RNAi pathway and the mechanism of gene silencing, refer to recent reviews (Bosher and Labouesse, 2000; Hannon, 2002; Plasterk and Ketting, 2000; Zamore, 2001).
Performing RNAi Analysis in Mammalian Cells
A number of kits including the BLOCK-iT™ RNAi TOPO® Transcription Kit now exist to facilitate in vitro production of dsRNA that is targeted to a particular gene of interest. The dsRNA may be introduced directly into some invertebrate organisms or cell lines, where it functions to trigger the endogenous RNAi pathway resulting in inhibition of the target gene. Long dsRNA duplexes cannot be used directly for RNAi analysis in most somatic mammalian cell lines because introduction of long dsRNA into these cell lines induces a non-specific, interferon-mediated response, resulting in shutdown of translation and initiation of cellular apoptosis (Kaufman, 1999). To avoid triggering the interferon-mediated host cell response, dsRNA duplexes of less than 30 nucleotides must be introduced into cells (Stark et al., 1998). For optimal results in gene knockdown studies, the size of the dsRNA duplexes (i.e. siRNA) introduced into mammalian cells is further limited to 21-23 nucleotides.
Using the Kit for RNAi Analysis
The BLOCK-iT™ Dicer RNAi Transfection Kit and the BLOCK-iT™ Complete Dicer RNAi Kit facilitate in vitro production of a complex pool of 21-23 nucleotide siRNA duplexes that is targeted to a particular gene of interest. The kits use a recombinant human Dicer enzyme (see below for more information) to cleave a long dsRNA substrate (produced with the BLOCK-iT™ RNAi TOPO® Transcription Kit) into a pool of 21-23 nucleotide d-siRNA that may be transfected into mammalian cells. Introduction of d-siRNA into the cells then triggers the endogenous RNAi pathway, resulting in inhibition of the target gene. For a diagram of the process, see the figure below.
BLOCK-iT™ Dicer RNAi Transfection Kit
BLOCK-iT™ Dicer Enzyme
BLOCK-iT™ Dicer is a recombinant human enzyme (Myers et al., 2003; Provost et al., 2002) that cleaves long dsRNA processively into 21-23 nucleotide d-siRNA duplexes with 2 nucleotide 3' overhangs. The Dicer enzyme is a member of the RNase III family of double-stranded RNA-specific endonucleases, and consists of an ATP-dependent RNA helicase domain, a Piwi/Argonaute/Zwille (PAZ) domain, two RNase III domains, and a dsRNA-binding domain (Bernstein et al., 2001; Zamore, 2001). In addition to its role in the generation of siRNA, Dicer is also involved in the processing of short temporal RNA (stRNA) (Hutvagner et al., 2001; Ketting et al., 2001) and microRNA (miRNA) (Carrington and Ambros, 2003) from stable hairpin or stem-loop precursors.
Materials
Shipping/Storage
The BLOCK-iT™ Dicer RNAi Kits are shipped as described below. Upon receipt, store each item as detailed below. For more detailed information about the reagents supplied with the BLOCK-iT™ RNAi TOPO® Transcription Kit, refer to the BLOCK-iT™ RNAi TOPO® Transcription Kit manual.
Box Component Shipping Storage
1BLOCK-iT™ Dicer Enzyme KitDry ice-20 ° C
2BLOCK-iT™ RNAi Purification KitRoom temperatureRoom temperature
3Lipofectamine® 2000 ReagentBlue ice+4 ° C (do not freeze)
4 - 6BLOCK-iT™ RNAi TOPO® Transcription KitBLOCK-iT™ TOPO® Linker Kit and BLOCK-iT™ RNAi Transcription Kit: Dry ice BLOCK-iT™ RNAi Purification Kit: Room temperatureBLOCK-iT™ TOPO® Linker Kit and BLOCK-iT™ RNAi Transcription Kit: -20 ° C BLOCK-iT™ RNAi Purification Kit: Room temperature
BLOCK-iT™ Dicer Enzyme Kit
The following reagents are included with the BLOCK-iT™ Dicer Enzyme Kit (Box 1). Store the reagents at -20 ° C.
Reagent
Composition
Amount
BLOCK-iT Dicer Enzyme
1 U/µl in a proprietary buffer
300 µl
10X Dicer Buffer
Proprietary
150 µl
50X Dicer Stop Buffer
0.5 mM EDTA, pH 8.0
30 µl
RNase-Free Water
--
1.5 ml
Unit Definition
One unit of BLOCK-iT™ Dicer enzyme cleaves 1 µg of double-stranded RNA (dsRNA) in 16 hours at 37 °  C.
BLOCK-iT™ RNAi Purification Kit
The following reagents are included with the BLOCK-iT™ RNAi Purification Kit (Box 2). Store reagents at room temperature. Use caution when handling the RNA Binding Buffer. (see the next page for more information) 
Note: Catalog no. K3650-01 includes two boxes of BLOCK-iT™ RNAi Purification reagents. One box is supplied with the BLOCK-iT™ RNAi TOPO® Transcription Kit for purification of sense and antisense single-stranded RNA (ssRNA). The second box is supplied for purification of diced siRNA (d-siRNA).
Reagent
Composition
Amount
RNA Binding Buffer
Proprietary
1.8 ml
5X RNA Wash Buffer
Proprietary
2.5 ml
RNase-Free Water
--
800 µl
RNA Spin Cartridges
--
10
RNA Recovery Tubes
--
10
siRNA Collection Tubes*
--
5
50X RNA Annealing Buffer
500 mM Tris-HCl, pH 8.0
1 M NaCl
50 mM EDTA, pH 8.0
50 µl
*siRNA Collection Tubes are used for purification of dsiRNA only, and are not required for the purification of the ssRNA.
The RNA Binding Buffer supplied in the BLOCK-iT™ RNAi Purification Kit contains guanidine isothiocyanate. This chemical is harmful if it comes in contact with the skin or is inhaled or swallowed. Always wear a laboratory coat, disposable gloves, and goggles when handling solutions containing this chemical.
Do not add bleach or acidic solutions directly to solutions containing guanidine isothiocyanate or sample preparation waste. Guanidine isothiocyanate forms reactive compounds and toxic gases when mixed with bleach or acids.
Lipofectamine® 2000 Reagent
Each BLOCK-iT™ Dicer RNAi Kit includes Lipofectamine® 2000 Reagent (Box 3) for high efficiency transfection of d-siRNA into mammalian cells. Lipofectamine® 2000 Reagent is supplied as follows:
Size: 0.75 ml
Concentration: 1 mg/ml
Storage: +4 °C; do not freeze
BLOCK-iT™ RNAi TOPO® Transcription Kit
The BLOCK-iT™ Complete Dicer RNAi Kit (Catalog no. K3650-01) includes the BLOCK-iT™ RNAi TOPO® Transcription Kit to facilitate production of double-stranded RNA (dsRNA) from your gene of interest. Refer to the BLOCK-iT™ RNAi TOPO® Transcription Kit manual for a detailed description of the reagents provided with the kit and instructions to produce dsRNA.Accessory Products
Introduction
The products listed in this section may be used with the BLOCK-iT™ Dicer RNAi Kits. For more information, refer to our Web site (www.invitrogen.com) or call Technical Service.
Accessory Products
Some of the reagents supplied in the BLOCK-iT™ Dicer RNAi Kits as well as other products suitable for use with the kit are available separately from Invitrogen. Ordering information is provided below. 
Ordering Information
Catalog # Name Size Price (USD) Qty
11668027 Lipofectamine™ 2000 Transfection Reagent, 0.75 mL Each
519.65
Online Exclusive
590.00
Save 70.35 (12%)
11668019 Lipofectamine™ 2000 Transfection Reagent, 1.5 mL Each
924.00
31985062 Opti-MEM™ I Reduced Serum Medium, 100 mL Each
29.65
Online Exclusive
34.34
Save 4.69 (14%)
31985070 Opti-MEM™ I Reduced Serum Medium, 500 mL Each
78.75
10010023 PBS, pH 7.4, 500 mL Each
32.76
EC63152BOX Novex™ TBE Gels, 20%, 12-well, 10 Gels/Box Each
216.65
Online Exclusive
243.00
Save 26.35 (11%)
K145501 β-Gal Assay Kit Each
506.00
Generating Double-Stranded RNA (dsRNA)
Introduction
Before you can use the BLOCK-iT™ Dicer Enzyme to produce short interfering RNA (siRNA), you must generate double-stranded RNA (dsRNA) substrate representing your target sequence of interest. Guidelines and recommendations to generate dsRNA are provided below.
For optimal, high-yield production of dsRNA, we recommend using the BLOCK-iT™ RNAi TOPO® Transcription Kit available from Invitrogen (Catalog no. K3500-01). The BLOCK-iT™ RNAi TOPO® Transcription Kit supplies the reagents necessary to generate T7 promoter-based DNA templates from any Taq-amplified PCR product, then use these templates in in vitro transcription reactions to generate sense and antisense RNA transcripts. The kit also includes reagents to enable purification and annealing of the RNA transcripts to produce high yields of dsRNA that are ready-to-use in the dicing reaction.
For detailed protocols and guidelines to generate dsRNA from your target gene sequence, refer to the BLOCK-iT™ RNAi TOPO® Transcription Kit manual. This manual is supplied with the BLOCK-iT™ Complete Dicer RNAi Kit , but is also available for downloading from our Web site ( www.invitrogen.com) or by contacting Technical Service .
Choosing the Target Sequence
When performing RNAi analysis, your choice of target sequence can significantly affect the degree of gene knockdown observed. In addition, the size of the target sequence and the resulting dsRNA can affect the yields of d-siRNA produced. Consider the following factors when choosing your target sequence.
  • Select a target sequence that covers a reasonable portion of the gene of interest and that does not contain regions of strong homology with other genes.
  • Limit the size of the target sequence. Although smaller or larger target sequences are possible, we recommend limiting the initial target sequence to a size range of 500 bp to 1 kb for the following reasons.
a. This balances the risk of including regions of strong homology between the target gene and other  genes that could result in non-specific off-target effects during RNAi analysis with the benefits of using a more complex pool of siRNA.
b. When producing sense and antisense transcripts of the target template, the highest transcription efficiencies are obtained with transcripts in the 500 bp to 1 kb size range. Target templates outside this size range transcribe less efficiently, resulting in lower yields of dsRNA.
c. Double-stranded RNA that is under 1 kb in size is efficiently diced. Larger dsRNA substrates can be used but yields may decline as the size increases.
Note:   The BLOCK-iT™ Dicer RNAi Kits have been used successfully to knock down gene activity with dsRNA substrates ranging from 150 bp to 1.3 kb in size.
Factors to Consider When Generating dsRNA
If you are using your own method or another kit to produce dsRNA, consider the following factors when generating your dsRNA. These factors will influence the yields of d-siRNA produced from the dicing reaction.
  • Amount of dsRNA required for dicing: We use 60 µg of dsRNA in a typical 300 µl dicing reaction to recover 12-18 µg of d-siRNA after purification. This amount of d-siRNA is generally sufficient to transfect approximately 150 wells of cells plated in a 24-well format. You should have an idea of the scale and scope of your RNAi experiment to determine how much dsRNA you will need to dice.
  • Note:   If you wish to dice less than 60 µg of dsRNA, you will need to scale down the dicing reaction proportionally.
  • Concentration of dsRNA: The amount of dsRNA in a dicing reaction should not exceed half the reaction volume; therefore, the concentration of your dsRNA should be US  400 ng/µl if you wish to dice 60 µg of dsRNA.
  • Buffering of dsRNA: We recommend storing your dsRNA sample in a buffered solution containing 1 mM EDTA and no more than 100 mM salt (i.e. TE Buffer at pH 7-8 or 1X RNA Annealing Buffer). This helps to stabilize the dsRNA and provides the optimal environment for efficient cleavage by the Dicer Enzyme.
  • Note:    If you have used the BLOCK-iT™ RNAi TOPO® Transcription Kit to produce dsRNA, your dsRNA sample will be in 1X RNA Annealing Buffer (10 mM Tris-HCl, 20 mM NaCl, 1 mM EDTA, pH 8.0).
  • The quality of your dsRNA: To obtain the highest yields of d-siRNA, we recommend using purified dsRNA in the dicing reaction.
Once you have generated your purified dsRNA, we recommend saving an aliquot of the dsRNA for future gel analysis. We generally use agarose or polyacrylamide gel electrophoresis to assess the success of the dicing reaction by comparing an aliquot of the dicing reaction to an aliquot of the dsRNA substrate.
Performing the Dicing Reaction
Introduction
Once you have produced your target dsRNA, you will perform an in vitro dicing reaction using the reagents supplied in the BLOCK-iT™ Dicer Enzyme Kit (Box 1) to generate d-siRNA duplexes of 21-23 nucleotides in size.
BLOCK-iT™ Dicer Enzyme Activity
One unit of BLOCK-iT™ Dicer Enzyme cleaves 1 µg of dsRNA in 16 hours at 37°  C. Note that the Dicer enzyme does not cleave dsRNA to d-siRNA with 100% effi-ciency, i.e. dicing 1 µg of dsRNA does not generate 1 µg of d-siRNA. Under these optimal reaction conditions, the Dicer enzyme cleaves dsRNA to d-siRNA with an efficiency of approximately 25-35%. For example, dicing 60 µg of dsRNA in a 300 µl dicing reaction typically yields 12-18 µg of d-siRNA following purification.
Note the following:
  • It is possible to use more than 60 µg of dsRNA in a 300 µl dicing reaction; however, the BLOCK-iT™ Dicer Enzyme becomes less efficient under these conditions. Although you may generate a higher mass yield of d-siRNA, the % yield of d-siRNA will decrease.
  • Do not increase the amount of BLOCK-iT™ Dicer Enzyme used in the dicing reaction (to greater than 60 units in a 300 µl reaction) or increase the length of the dicing reaction (to greater than 18 hours). Under either of these conditions, the BLOCK-iT™ Dicer Enzyme can bind to d-siRNA and cleave the 21-23 nt duplexes into smaller products, resulting in lower yields of d-siRNA.
Amount of dsRNA to Use
For a typical 300 µl dicing reaction, you will need 60 µg of target dsRNA. If you want to dice less than 60 µg of dsRNA, scale down the entire reaction proportionally.
The total volume of dsRNA added should not exceed half the volume of the reaction. Thus, for best results, make sure that the starting concentration of your dsRNA is US 400 ng/µl. 
Positive Control
If you are using the BLOCK-iT™ Complete Dicer RNAi Kit, and have performed all of the recommended control reactions using the control reagents supplied in the BLOCK-iT™ RNAi TOPO® Transcription portion of the kit, you should have purified dsRNA representing a 1 kb portion of the lacZ gene. We recommend setting up a separate dicing and purification reaction using the control lacZ dsRNA. You can then co-transfect the resulting purified lacZ d-siRNA and the pcDNA™1.2/V5-GW/lacZ control plasmid supplied with the kit into your mammalian cell line as a positive control for the RNAi response in that cell line. Alternatively, you may use the lacZ d-siRNA as a negative control for non-specific, off-target effects in your cell line.
Steps to Avoid RNase Contamination
  • Use RNase-free sterile pipette tips and supplies for all manipulations.
  • Use DEPC-treated solutions as necessary.
  • Wear gloves when handling reagents and solutions, and when performing reactions.
Materials Needed
Have the following reagents on hand before beginning:
  • Purified dsRNA (> 400 ng/µl in 1X RNA Annealing Buffer or TE Buffer, pH 7-8; see page 8 for more information)
  • BLOCK-iT™ Dicer Enzyme (1 U/µl; supplied with the kit, Box 1; keep at -20° C until immediately before use)
  • 10X Dicer Buffer (supplied with the kit, Box 1)
  • RNase-Free Water (supplied with the kit, Box 1)
  • 50X Dicer Stop Buffer (supplied with the kit, Box 1)
Dicing Procedure
Follow the procedure below to perform the dicing reaction. Make sure that the volume of dsRNA added does not exceed half the volume of the reaction (i.e. US  150 µl).
  1. Set up a 300 µl dicing reaction on ice using the following reagents in the order shown.
  2. Reagent
    Sample
    10X Dicer Buffer
    30 µl
    RNase-Free Water
    up to 210 µl
    Purified dsRNA (60 µg)
    1-150 µl
    BLOCK-iT Dicer Enzyme (1 U/µl)
    60 µl
    Total volume
    300 µl
  3. Mix reaction gently and incubate for 14-18 hours at 37° C.
    Note: Do not incubate the reaction for longer than 18 hours as this may result in a lower yield of d-siRNA due to cleavage of d-siRNA by the Dicer enzyme.
  4. Add 6 µl of 50X Dicer Stop Solution to the reaction.
  5. Check the integrity of your d-siRNA, if desired.
  6. Proceed to purify the d-siRNA (see Purifying Diced siRNA (d-siRNA) or store the dicing reaction overnight at -20° C.
Checking the Integrity of d-siRNA
You may verify the integrity of your d-siRNA using polyacrylamide or agarose gel electrophoresis, if desired. We suggest running an aliquot of your dicing reaction (0.5-1 µl of a 300 µl reaction; equivalent to 100-200 ng of dsRNA) on the appropriate gel and comparing it to an aliquot of your starting dsRNA. Be sure to include an appropriate molecular weight standard. We generally use the following gels and molecular weight standard:
  • Agarose gel: 4% E-Gel® (Invitrogen, Catalog no. G5000-04)
  • Polyacrylamide gel: 20% Novex® TBE Gel (Invitrogen, Catalog no. EC63152BOX)
  • Molecular weight standard: 10 bp DNA Ladder (Invitrogen, Catalog no. 10821-015)
What You Should See
When analyzing an aliquot of the dicing reaction by gel electrophoresis, we generally see the following:
  • A predominant band of approximately 21-23 nt representing the d-siRNA.
  • 4% E-Gel®: A high molecular weight smear representing uncleaved dsRNA and partially cleaved products. Generally, this band does not resolve well on an agarose gel and runs close to the well.
  • Novex® 20% TBE Gel: A high molecular weight band and a smear representing uncleaved dsRNA and partially cleaved products. The dsRNA band generally resolves better on a polyacrylamide gel.
  • For an example of expected results obtained from agarose gel electrophoresis , see the next page. If the band representing d-siRNA is weak or if you do not see a band, see Troubleshooting.
Example of Expected Results
In this experiment, purified dsRNA representing a 1 kb region of the lacZ gene was generated following the recommended protocols and using the reagents supplied in the BLOCK-iT™ RNAi TOPO® Transcription Kit. The lacZ dsRNA was diced using the procedure on page 10. Aliquots of the dicing reaction (equivalent to 200 ng of dsRNA) and the initial dsRNA substrate were analyzed on a 4%
E-Gel®. 
Results:   A prominent band representing d-siRNA of the expected size is clearly visible in the dicing reaction sample (lane 3). This band is not visible in the initial dsRNA substrate sample (lane 2).
d-siRNA Expected Results
Purifying Diced siRNA (d-siRNA)
Introduction
This section provides guidelines and instructions to purify the d-siRNA produced in the dicing reaction. Use the BLOCK-iT™ RNAi Purification reagents (Box 2) supplied with the kit.
Before proceeding to transfection, note that you must purify the d-siRNA produced in the dicing reaction to remove contaminating long dsRNA duplexes. Transfection of unpurified d-siRNA can trigger the interferon-mediated response and cause host cell shutdown and cellular apoptosis. When purifying d-siRNA, follow the purification procedure provided on page 16 exactly as instructed. This procedure is optimized to allow removal of contaminating long dsRNA and recovery of high yields of d-siRNA.
Experimental Outline
To purify d-siRNA, you will:
  1. Add RNA Binding Buffer and isopropanol to the dicing reaction to denature the proteins and to enable the contaminating dsRNA to bind to the column.
  2. Add half the volume of the sample to an RNA spin cartridge. The dsRNA binds to the silica-based membrane in the cartridge, and the d-siRNA and denatured proteins flow through the cartridge. Save the flow-through.
  3. Transfer the RNA spin cartridge to an siRNA Collection Tube and add the remaining sample to the RNA spin cartridge. Repeat Step 2. Save the flow-through.
  4. Pool the flow-throughs from Step 2 and Step 3 in the siRNA Collection Tube and add isopropanol to the sample to enable the d-siRNA to bind to the column.
  5. Add the sample to a second RNA spin cartridge. The d-siRNA bind to the membrane in the cartridge.
  6. Wash the membrane-bound d-siRNA to eliminate residual RNA Binding Buffer, isopropanol, and any remaining impurities.
  7. Elute the d-siRNA from the RNA spin cartridge with water.
  8. Add 50X RNA Annealing Buffer to the eluted d-siRNA to stabilize the d-siRNA for storage.
The figure below illustrates the d-siRNA purification process.
d-siRNA purification process
Advance Preparation
Before using the BLOCK-iT™ RNA Purification reagents for the first time, add 10 ml of 100% ethanol to the entire amount of 5X RNA Wash Buffer to obtain a 1X RNA Wash Buffer (total volume = 12.5 ml).
Place a check in the box on the 5X RNA Wash Buffer label to indicate that the ethanol was added. Store the 1X RNA Wash Buffer at room temperature.
The RNA Binding Buffer contains guanidine isothiocyanate. This chemical is harmful if it comes in contact with the skin or is inhaled or swallowed. Always wear a laboratory coat, disposable gloves, and goggles when handling solutions containing this chemical.
Do not add bleach or acidic solutions directly to solutions containing guanidine isothiocyanate or sample preparation waste. Guanidine isothiocyanate forms reactive compounds and toxic gases when mixed with bleach or acids.
Materials Needed
Have the following materials on hand before beginning:
  • Dicing reaction (from Step 5)
  • RNA Binding Buffer (supplied with the kit, Box 2)
  • 2-mercaptoethanol
  • Isopropanol
  • RNA Spin Cartridges (supplied with the kit, Box 2; two for each sample)
  • siRNA Collection Tube (supplied with the kit, Box 2)
  • 1X RNA Wash Buffer (see Advance Preparation, above)
  • RNase-Free Water (supplied with the kit, Box 2)
  • RNA Recovery Tube (supplied with the kit, Box 2)
  • 50X RNA Annealing Buffer (supplied with the kit, Box 2)
  • RNase-free supplies
d-siRNA Purification Procedure                                                                                                                                           
TOP
Use this procedure to purify d-siRNA produced from dicing 60 µg of dsRNA in a 300 µl reaction volume (see Step 5, Performing the Dicing Reaction). If you have digested < 60 µg of dsRNA and have scaled down the volume of your dicing reaction, scale down the volume of your purification reagents proportionally. For example, if you have digested 30 µg of dsRNA in a 150 µl dicing reaction, scale down the volume of purification reagents used by half.
Important:   Before beginning, remove the amount of RNA Binding Buffer needed and add 2-mercaptoethanol to a final concentration of 1% (v/v). Use fresh and discard any unused solution.
  1. To each dicing reaction (~300 µl volume), add 300 µl of RNA Binding Buffer containing 1% (v/v) 2-mercaptoethanol followed by 300 µl of isopropanol to obtain a final volume of 900 µl. Mix well by pipetting up and down 5 times.
  2. Apply half of the sample (~450 µl) to the RNA Spin Cartridge. Centrifuge at 14,000 x g for 15 seconds at room temperature.
  3. Transfer the RNA spin cartridge to an siRNA Collection Tube. Save the flow-through containing d-siRNA from Step 2.
  4. Apply the remaining half of the sample (~450 µl) to the RNA Spin Cartridge. Centrifuge at 14,000 x g for 2 minutes at room temperature.
  5. Remove the RNA Spin Cartridge from the siRNA Collection Tube and discard. Save the flow-through containing d-siRNA.
  6. Transfer the flow-through from Step 2 (~450 µl) to the siRNA Collection Tube containing the flow-through from Step 4 (~450 µl) to obtain a final volume of ~900 µl. Add 600 µl of isopropanol to the sample to obtain a final volume of 1.5 ml. Mix well by pipetting up and down.
  7.  Apply one-third of the sample (~500 µl) to a new RNA Spin Cartridge. Centrifuge at 14,000 x g for 15 seconds at room temperature. Discard the flow-through.
  8. Repeat Step 7 twice, applying one-third of the remaining sample (~500 µl) to the RNA Spin Cartridge each time.
  9. Add 500 µl of 1X RNA Wash Buffer to the RNA Spin Cartridge containing bound d-siRNA. Centrifuge at 14,000 x g for 15 seconds at room temperature. Discard the flow-through.
  10. Repeat the wash step (Step 9).
  11. Centrifuge the RNA Spin Cartridge at 14,000 x g for 1 minute at room temperature to remove residual 1X RNA Wash Buffer from the cartridge and to dry the membrane.
  12. Remove the RNA Spin Cartridge from the Wash Tube, and place it in an RNA Recovery Tube.
  13. Add 30 µl of RNase-Free Water to the RNA Spin Cartridge. Let stand at room temperature for 1 minute, then centrifuge the RNA Spin Cartridge at 14,000 x g for 2 minutes at room temperature to elute the d-siRNA. Proceed to Step 14,
  14. Repeat Step 13, previous page, eluting the d-siRNA into the same RNA Recovery Tube. The total volume of eluted d-siRNA is 60 µl.
  15. Add 1.2 µl of the 50X RNA Annealing Buffer to the eluted d-siRNA to obtain a final concentration of 1X RNA Annealing Buffer. Adding RNA Annealing Buffer to the sample increases the stability of the d-siRNA.
  16. Proceed to quantitate the concentration of your purified d-siRNA (see Determining the Purity and Concentration of d-siRNA, below).
  17. Store the purified d-siRNA at -80 ° C. Depending on the amount of d-siRNA produced and your downstream application, you may want to aliquot the d-siRNA before storage at -80° C.
Important:
   When using the d-siRNA, avoid repeated freezing and thawing as d-siRNA can degrade with each freeze/thaw cycle.
Determining the Purity and Concentration of d-siRNA 
Use the procedure below to determine the purity and concentration of your purified d-siRNA.
  1. Dilute an aliquot of the purified d-siRNA 20-fold into 1X RNA Annealing Buffer in a total volume appropriate for your quartz cuvettes and spectrophotometer.
  2. Measure OD at A260 and A280 in a spectrophotometer. Blank the sample against 1X RNA Annealing Buffer.
  3. Calculate the concentration of the d-siRNA by using the following equation:  d-siRNA concentration (µg/ml) = A260 x Dilution factor (20) x 40 µg/ml
  4. Calculate the yield of the d-siRNA by using the following equation:  d-siRNA yield (µg) = d-siRNA concentration (µg/ml) x vol. of d-siRNA (ml)
  5. Evaluate the purity of the purified d-siRNA by determining the A260/A280 ratio. For optimal purity, the A260/A280 ratio should range from 1.9-2.2.
Verifying the Quality of Your d-siRNA
You may verify the quality of your purified d-siRNA using polyacrylamide or agarose gel electrophoresis, if desired. We suggest running a small aliquot of your purified d-siRNA (0.5-1 µl) on the appropriate gel and comparing it to an aliquot of your dicing reaction (equivalent to 100-200 ng of dsRNA). Be sure to include an appropriate molecular weight standard. For recommended gels and a molecular weight standard, we generally use the same gels and molecular weight standard that we use to analyze the quality of the dicing reaction. 
If the band representing purified d-siRNA is weak or if you do not see a band, see Troubleshooting for tips to purify your d-siRNA.
Example of Expected Results
In this experiment, the lacZ d-siRNA generated in the dicing reaction depicted in the Example above were purified using  the  procedure. Aliquots of the purified lacZ d-siRNA (80 ng) and the lacZ dicing reaction (equivalent to 200 ng of dsRNA) were analyzed on a 4% E-Gel®. 
Results: A prominent band representing purified d-siRNA of the expected size is clearly visible in lane 3. No contaminating dsRNA or other high molecular weight products remain in the purified d-siRNA sample.
purified d-siRNA
How Much d-siRNA to Expect
The typical yield of d-siRNA obtained from dicing 60 µg of dsRNA (500 bp to 1 kb in size) in a 300 µl dicing reaction ranges from 12-18 µg, with a concentration of 200-300 ng/µl. Note that yields may vary depending on the size and quality of the dsRNA.
Transfecting Cells
Introduction
Once you have purified your d-siRNA, you may perform RNAi analysis by transfecting the d-siRNA into the mammalian cell line of interest, and assaying for inhibition of target gene expression. This section provides general guidelines and protocols to transfect your purified d-siRNA into mammalian cells using the Lipofectamine® 2000 Reagent (Box 3) supplied with the kit. Suggested transfection conditions are provided as a starting point. You will need to optimize transfection conditions to obtain the best results for your target gene and mammalian cell line.
Reminder:   You must transfect mammalian cells with purified d-siRNA. Note that transfecting cells with unpurified d-siRNA containing contaminating long dsRNA (i.e. with material directly taken from the dicing reaction) can trigger the interferon-mediated cellular response, resulting in host cell shutdown and cellular apoptosis
.
Factors Affecting Gene Knockdown Levels
A number of factors can influence the degree to which expression of your gene of interest is reduced (i.e. gene knockdown) in an RNAi experiment including:
  • Transfection efficiency
  • Transcription rate of the target gene of interest
  • Stability of the target protein
  • Growth characteristics of your mammalian cell line
Take these factors into consideration when designing your transfection and RNAi experiments.
Lipofectamine® 2000 Reagent
The Lipofectamine® 2000 Reagent supplied with the kit is a proprietary, cationic lipid-based formulation suitable for the transfection of nucleic acids including d-siRNA and siRNA into eukaryotic cells (Ciccarone et al., 1999; Gitlin et al., 2002; Yu et al., 2002). Using Lipofectamine® 2000 to transfect d-siRNA into eukaryotic cells offers the following advantages:
  • Provides the highest transfection efficiency in many cell types
  • Is the most widely used transfection reagent for delivery of d-siRNA or siRNA into eukaryotic cells (Gitlin et al., 2002; Yu et al., 2002)
  • d-siRNA-Lipofectamine® 2000 complexes can be added directly to cells in culture medium in the presence of serum.
  • Removal of complexes, medium change, or medium addition following transfection are not required, although complexes can be removed after 4-6 hours without loss of activity.
Lipofectamine® 2000 is also available separately from Invitrogen
Important Guidelines
Follow these guidelines when transfecting siRNA into mammalian cells using Lipofectamine® 2000:
  1. Cell density: For optimal results, we recommend plating cells such that they will be 30-50% confluent at the time of transfection. Gene knockdown levels are generally assayed 24-72 hours following transfection. Transfecting cells at a lower density allows a longer interval between transfection and assay time, and minimizes the loss of cell viability due to cell overgrowth. Depending on the nature of the target gene, higher or lower cell densities may be suitable with optimization of conditions.
  2. For optimal results, use Opti-MEM® I Reduced Serum Medium (Invitrogen, Catalog no. 31985-062) to dilute Lipofectamine® 2000 and d-siRNA prior to complex formation.
  3. Do not include antibiotics in media used during transfection as this will reduce transfection efficiency and cause cell death.
Materials to Have on Hand
Have the following materials on hand before beginning:
  • Mammalian cell line of interest (make sure that cells are healthy and greater than 90% viable before transfection)
  • Purified d-siRNA of interest (40 ng/µl)
  • Note:    If you have diced 60 µg of dsRNA, the typical yield of d-siRNA obtained after purification is 12-18 µg at a concentration of 200-300 ng/µl)
  • Positive control, if desired (see below)
  • Lipofectamine® 2000 Reagent (supplied with the kit; store at +4 ° C until use)
  • Opti-MEM® I Reduced Serum Medium (Invitrogen, Catalog no. 31985-062; pre-warmed)
  • Sterile tissue culture plates and other tissue culture supplies
Positive Control
If you are using the BLOCK-iT™ Complete Dicer RNAi Kit, and have diced the control lacZ dsRNA, two options exist to use the resulting purified lacZ d-siRNA for RNAi analysis:
  1. Use the lacZ d-siRNA as a negative control for non-specific off-target effects.
  2. Use the lacZ d-siRNA as a positive control to assess the RNAi response in your cell line by co-transfecting the lacZ d-siRNA and the pcDNA™1.2/V5-GW/lacZ reporter plasmid supplied with the kit into your mammalian cells using Lipofectamine® 2000. Assay for knockdown of b-galactosidase expression 24 hours post-transfection using Western blot analysis or activity assay.
Important: Transfection conditions (i.e. cell density and reagent amounts) vary slightly when d-siRNA and plasmid DNA are co-transfected into mammalian cells.
Transfection Procedure
Use this procedure to transfect mammalian cells using Lipofectamine® 2000. Refer to the table in Recommended Reagent Amounts and Volumes, below for the appropriate reagent amounts and volumes to add for different tissue culture formats. Use the recommended Lipofectamine® 2000 amounts as a starting point for your experiments, and optimize conditions for your cell line and d-siRNA.
  1. One day before transfection, plate cells in the appropriate amount of growth medium without antibiotics such that they will be 30-50% confluent at the time of transfection.
  2. For each transfection sample, prepare d-siRNA:Lipofectamine® 2000 complexes as follows:
    • Dilute d-siRNA in the appropriate amount of Opti-MEM® I Reduced Serum Medium without serum. Mix gently.
    • Mix Lipofectamine® 2000 gently before use, then dilute the appropriate amount in Opti-MEM® I Reduced Serum Medium. Mix gently and incubate for 5 minutes at room temperature.
    Note:   Combine the diluted Lipofectamine® 2000 with the diluted d-siRNA within 30 minutes. Longer incubation times may decrease activity.
    • After the 5 minute incubation, combine the diluted d-siRNA with the diluted Lipofectamine® 2000. Mix gently and incubate for 20 minutes at room temperature to allow the d-siRNA:Lipofectamine® 2000 complexes to form (solution may appear cloudy).
  3. Add the d-siRNA:Lipofectamine® 2000 complexes to each well containing cells and medium. Mix gently by rocking the plate back and forth.
  4. Incubate the cells at 37 °C in a CO2 incubator for 24-96 hours as appropriate until you are ready to assay for gene knockdown. It is not necessary to remove the complexes or change the medium; however, growth medium may be replaced after 4-6 hours without loss of transfection activity.
Recommended Reagent Amounts and Volumes
The table below lists the recommended reagent amounts and volumes to use to transfect cells in various tissue culture formats. Use the recommended amounts of d-siRNA (see column 4) and Lipofectamine® 2000 (see column 6) as a starting point for your experiments, and optimize conditions for your cell line and target gene.
Note:   With automated, high-throughput systems, larger complexing volumes are recommended for transfections in 96-well plates.
Culture Vessel
Relative Surface Area (vs. 24-well)
Volume of Plating Medium
d-siRNA (ng) and Dilution Volume (µl)
d-siRNA Amounts (ng) for Optimization
Lipofectamine® 2000 (µl) and Dilution Volume (µl)
Lipofectamine® 2000 Amounts (µl) for Optimization
96-well
0.2
100 µl
20 ng in 25 µl
5-50 ng
0.6 µl in 25 µl
0.2-1.0 µl
24-well
1
500 µl
50 ng in 50 µl
20-200 ng
1 µl in 50 µl
0.5-1.5 µl
6-well
5
2 ml
250 ng in 250 µl
100-1000 ng
5 µl in 250 µl
2.5-6 µl
Optimizing Transfection
To obtain the highest transfection efficiency and low non-specific effects, optimize transfection conditions by varying the cell density (from 30-50% confluence) and the amounts of d-siRNA (see column 5) and Lipofectamine® 2000 (see column 7) as suggested in the tableon the previous page. For cell lines that are particularly sensitive to transfection-mediated cytotoxicity (e.g. HeLa, HT1080), use the lower amounts of Lipofectamine® 2000 suggested in the table above (see column 7).
What You Should See
When performing RNAi experiments using d-siRNA, we generally observe inhibition of the gene of interest within 24 to 96 hours after transfection. The degree of gene knockdown depends on the time of assay, stability of the protein of interest, and on the other factors listed on page 19. Note that 100% gene knockdown is generally not observed, but > 95% is possible with optimized conditions.
For examples of results obtained from RNAi experiments using d-siRNA, see below.
Co-transfecting d-siRNA and Plasmid DNA
If you are using the lacZ d-siRNA as a positive control to assess the RNAi response in your cell line, you will co-transfect the lacZ d-siRNA and the pcDNA™ 1.2/V5-GW/lacZ reporter plasmid into the mammalian cell line and assay for inhibition of b-galactosidase expression after 24 hours. When co-transfecting d-siRNA and plasmid DNA, follow the procedure on the previous page with the following exceptions:
  • Plate cells such that they will be 90% confluent at the time of transfection.
  • Refer to the table below for the recommended amount of d-siRNA (see column 3) and plasmid DNA (see column 4) to transfect in a particular tissue culture format.
Note:   We generally transfect twice the mass of plasmid DNA as d-siRNA.
  • Use the recommended Lipofectamine® 2000 amounts in the table below (see column 6) as a starting point, and optimize conditions for your cell line if desired. To optimize conditions, vary the amount of Lipofectamine® 2000 as suggested in the table below (see column 7).
Culture Vessel
Volume of Plating Medium
d-siRNA (ng)
Plasmid DNA (ng)
Nucleic Acid Dilution Volume
Lipofectamine® 2000 (µl) and Dilution Volume (µl)
Lipofectamine® 2000 Amounts (µl) for Optimization
96-well
100 µl
20 ng
40 ng
25 µl
0.6 µl in 25 µl
0.2-1.0 µl
24-well
500 µl
50 ng
100 ng
50 µl
2 µl in 50 µl
0.5-2.0 µl
6-well
2 ml
250 ng
500 ng
250 µl
10 µl in 250 µl
2.5-10 µl
Assaying for ß-galactosidase Expression
If you perform RNAi analysis using the control lacZ d-siRNA, you may assay for ß-galactosidase expression and knockdown by Western blot analysis or activity assay using cell-free lysates (Miller, 1972). Invitrogen offers the ß-gal Antiserum (Catalog no. R901-25) and the ß-Gal Assay Kit (Catalog no. K1455-01) for fast and easy detection of ß-galactosidase expression. For an example of results obtained from a ß-galactosidase knockdown experiment, see below.
Note:   The ß-galactosidase protein expressed from the pcDNA™1.2/V5-GW/lacZ control plasmid is fused to a V5 epitope and is approximately 119 kDa in size. If you are performing Western blot analysis, you may also use the Anti V5 Antibodies available from Invitrogen (e.g. Anti-V5-HRP Antibody; Catalog no. R961-25 or Anti-V5-AP Antibody, Catalog no. R962-25) for detection
Troubleshooting
Introduction
Use the information in this section to troubleshoot your dicing, purification, and transfection experiments.
Dicing Reaction
The table below lists some potential problems and possible solutions that may help you troubleshoot the dicing reaction.
Problem
Reason
Solution
Weak band representing d-siRNA observed on a poly-acrylamide or agarose gel ( i.e. low yield of d-siRNA)
Poor quality dsRNA
  • Generate dsRNA using the BLOCK-iT RNAi TOPO® Transcription Kit (refer to the BLOCK-iT RNAi TOPO® Transcription Kit manual for instructions).
  • Verify the concentration of your dsRNA.
Didn’t use enough dsRNA in the dicing reaction
  • Use 60 µg of dsRNA in a 300 µl dicing reaction. If you are dicing less dsRNA, scale down the entire dicing reaction proportionally.
  • Make sure that the amount of dsRNA added does not exceed half the reaction volume (i.e. concentration of initial dsRNA substrate must be > 400 ng/µl).
dsRNA was degraded
  • Make sure that the dsRNA sample is in a buffer containing 1 mM EDTA (i.e. TE Buffer, pH 7-8 or 1X RNA Annealing Buffer).
  • Avoid repeated freeze/thaw cycles. Aliquot the dsRNA and store at -80°C.
Incubated the dicing reaction for longer than 18 hours
Do not incubate the dicing reaction for longer than 18 hours.
Incubated the dicing reaction for less than 14 hours
Incubate the dicing reaction at 37 °C for 14-18 hours.
Smear with molecular weight  21 nt observed on a poly-acrylamide gel
Used too much BLOCK-iT Dicer Enzyme in the dicing reaction
Follow the recommended procedure to set up the dicing reaction. Do not use more than 60 units of BLOCK-iT Dicer Enzyme in a 300 µl reaction.
Incubated the dicing reaction for longer than 18 hours
Do not incubate the dicing reaction for longer than 18 hours.
Sample contaminated with RNase
  • Use RNase-free supplies and solutions.
  • Wear gloves when handling reagents and setting up the dicing reaction.
No d-siRNA produced
dsRNA was degraded
  • Make sure that the dsRNA sample is in a buffer containing 1 mM EDTA (i.e. TE Buffer, pH 7-8 or 1X RNA Annealing Buffer).
  • Avoid repeated freeze/thaw cycles. Aliquot the dsRNA and store at -80°C.
Sample was contaminated with RNase
  • Use RNase-free supplies and solutions.
  • Wear gloves when handling reagents and setting
ssRNA used as substrate
If you have used to the BLOCK-iT RNAi TOPO ® Transcription Kit to generate sense and antisense ssRNA, you must anneal the ssRNA to generate dsRNA prior to dicing.
Purifying d-siRNA
The table below lists some potential problems and possible solutions that may help you troubleshoot the purification procedure.
Problem
Reason
Solution
Low yield of purified d-siRNA obtained
Eluted d-siRNA from the RNA Spin Cartridge using TE Buffer
Elute d-siRNA from the RNA Spin Cartridge using water.
Concentration of d-siRNA incorrectly determined
·         Sample diluted into water for spectrophotometry
·         Sample blanked against water
  • Dilute sample in 1X RNA Annealing Buffer for spectrophotometry.
  • Blank sample against 1X RNA Annealing Buffer.
No d-siRNA obtained
Forgot to add ethanol to the 5X RNA Wash Buffer
Add 10 ml of ethanol to the 5X RNA Wash Buffer (2.5 ml) to obtain a 1X RNA Wash Buffer.
Forgot to add isopropanol to the combined flow-throughs from the first RNA Spin Cartridge
You must add isopropanol to the combined flow-throughs from the first RNA Spin Cartridge to enable the d-siRNA to bind to the second RNA Spin Cartridge.
Forgot to keep flow-throughs from the first RNA Spin Cartridge
Keep the flow-throughs from the first RNA Spin Cartridge. The flow-throughs contain the d-siRNA.
dsRNA present in purified d-siRNA sample
Forgot to add isopropanol to the dicing reaction
You must add RNA Binding Buffer containing 1% (v/v) b-mercaptoethanol and isopropanol to the dicing reaction to denature the proteins and enable the dsRNA to bind the first RNA Spin Cartridge.
Added the mixture containing the flow-through and isopropanol from the first RNA Spin Cartridge back onto the first RNA Spin Cartridge
You must add the mixture containing the flow-through and isopropanol from the first RNA Spin Cartridge to a second RNA Spin Cartridge as the first RNA Spin Cartridge contains bound dsRNA.
A260/A280 ratio not in the 1.9-2.2 range
Sample was not washed with 1X RNA Wash Buffer
Wash the RNA Spin Cartridge containing bound d-siRNA twice with 1X RNA Wash Buffer. 
RNA Spin Cartridge containing bound d-siRNA not centrifuged to remove residual 1X RNA Wash Buffer
Centrifuge RNA Spin Cartridge at 14,000 x g for 1 minute at room temperature to remove residual 1X RNA Wash Buffer and to dry the membrane .
Transfection and RNAi Analysis
The table below lists some potential problems and possible solutions that may help you troubleshoot your transfection and knockdown experiment.
Problem
Reason
Solution
Low levels of gene knockdown observed
Low transfection efficiency
  • Antibiotics added to the media during transfection
  • Cells were confluent at the time of transfection
  • Not enough d-siRNA transfected
  • Not enough Lipofectamine® 2000 used
  • Do not add antibiotics to the media during transfection.
  • Plate cells such that they will be 30-50% confluent at the time of transfection.
  • Increase the amount of d-siRNA transfected.
  • Optimize the transfection conditions for your cell line by varying the amount of Lipofectamine® 2000 used.
Didn’t wait long enough after transfection before assaying for gene knockdown
  • Repeat the transfection and wait for a longer period of time after transfection before assaying for gene knockdown.
  • Perform a time course of expression to determine the point at which the highest degree of gene knockdown occurs.
d-siRNA was degraded
  • Make sure that the d-siRNA is stored in 1X RNA Annealing Buffer.
  • Aliquot purified d-siRNA and avoid repeated freeze/thaw cycles.
Cytotoxic effects observed after transfection
Too much Lipofectamine® 2000 Reagent used
Optimize the transfection conditions for your cell line by varying the amount of Lipofectamine® 2000 Reagent used.
Cells transfected with unpurified d-siRNA
Purify d-siRNA using the RNAi Purification reagents supplied with the kit.
Important: Transfecting unpurified d-siRNA is not recommended as the contaminating dsRNA will cause host cell shutdown and apoptosis.
No gene knockdown observed
d-siRNA was degraded
  • d-siRNA was stored in water
  • d-siRNA was repeatedly frozen and thawed
  •  Make sure that the d-siRNA is stored in 1X RNA Annealing Buffer.
  • Aliquot purified d-siRNA and avoid repeated freeze/thaw cycles.
Target region contains no active siRNA
Select a larger target region or a different region.
Non-specific off-target gene knockdown observed
Target sequence contains strong homology to other genes
  • Select a new target sequence.
  • Limit the size range of the target sequence to 1 kb.
References
  1. Anandalakshmi, R., Pruss, G. J., Ge, X., Marathe, R., Mallory, A. C., Smith, T. H., and Vance, V. B. (1998). A Viral Suppressor of Gene Silencing in Plants. Proc. Natl. Acad. Sci. USA 95, 13079-13084.
  2. Bernstein, E., Caudy, A. A., Hammond, S. M., and Hannon, G. J. (2001). Role for a Bidentate Ribonuclease in the Initiation Step of RNA Interference. Nature 409, 363-366.
  3. Bosher, J. M., and Labouesse, M. (2000). RNA Interference: Genetic Wand and Genetic Watchdog. Nature Cell Biol. 2, E31-E36.
  4. Carrington, J. C., and Ambros, V. (2003). Role of MicroRNAs in Plant and Animal Development. Science 301, 336-338.
  5. Ciccarone, V., Chu, Y., Schifferli, K., Pichet, J.-P., Hawley-Nelson, P., Evans, K., Roy, L., and Bennett, S. (1999). Lipofectamine® 2000 Reagent for Rapid, Efficient Transfection of Eukaryotic Cells. Focus 21, 54-55.
  6. Cogoni, C., and Macino, G. (1999). Gene Silencing in Neurospora crassa Requires a Protein Homologous to RNA-Dependent RNA Polymerase. Nature 399, 166-169.
  7. Cogoni, C., and Macino, G. (1997). Isolation of Quelling-Defective (qde) Mutants Impaired in Posttranscriptional Transgene-Induced Gene Silencing in Neurospora crassa. Proc. Natl. Acad. Sci. USA 94, 10233-10238.
  8. Cogoni, C., Romano, N., and Macino, G. (1994). Suppression of Gene Expression by Homologous Transgenes. Antonie Van Leeuwenhoek 65, 205-209.
  9. Elbashir, S. M., Harborth, J., Lendeckel, W., Yalcin, A., Weber, K., and Tuschl, T. (2001). Duplexes of 21-Nucleotide RNAs Mediate RNA Interference in Cultured Mammalian Cells. Nature 411, 494-498.
  10. Elbashir, S. M., Harborth, J., Weber, K., and Tuschl, T. (2002). Analysis of Gene Function in Somatic Mammalian Cells Using Small Interfering RNAs. Methods 26, 199-213.
  11. Gitlin, L., Karelsky, S., and Andino, R. (2002). Short Interfering RNA Confers Intracellular Antiviral Immunity in Human Cells. Nature 418, 430-434.
  12. Hammond, S. M., Bernstein, E., Beach, D., and Hannon, G. J. (2000). An RNA-Directed Nuclease Mediates Genetic Interference in Caenorhabditis elegans. Nature 404, 293-296.
  13. Hannon, G. J. (2002). RNA Interference. Nature 418, 244-251.
  14. Hutvagner, G., McLachlan, J., Pasquinelli, A. E., Balint, E., Tuschl, T., and Zamore, P. D. (2001). A Cellular Function for the RNA-Interference Enzyme Dicer in the Maturation of the let-7 Small Temporal RNA. Science 293, 811-813.
  15. Jones, A. L., Thomas, C. L., and Maule, A. J. (1998). De novo Methylation and Co-Suppression Induced by a Cytoplasmically Replicating Plant RNA Virus. EMBO J. 17, 6385-6393.
  16. Kaufman, R. J. (1999). Double-Stranded RNA-Activated Protein Kinase Mediates Virus-Induced Apoptosis: A New Role for an Old Actor. Proc. Natl. Acad. Sci. USA 96, 11693-11695.
  17. Ketting, R. F., Fischer, S. E., Bernstein, E., Sijen, T., Hannon, G. J., and Plasterk, R. H. (2001). Dicer Functions in RNA Interference and in Synthesis of Small RNA Involved in Developmental Timing in C. elegans. Genes Dev. 15, 2654-2659.
  18. Li, W. X., and Ding, S. W. (2001). Viral Suppressors of RNA Silencing. Curr. Opin. Biotechnol. 12, 150-154.
  19. McManus, M. T., and Sharp, P. A. (2002). Gene Silencing in Mammals by Small Interfering RNAs. Nature Rev. Genet. 3, 737-747.
  20. Miller, J. H. (1972). Experiments in Molecular Genetics (Cold Spring Harbor, New York: Cold Spring Harbor Laboratory).
  21. Myers, J. W., Jones, J. T., Meyer, T., and J.E. Ferrell, J. (2003). Recombinant Dicer Efficiently Converts Large dsRNAs into siRNAs Suitable for Gene Silencing. Nat. Biotechnol. 21, 324-328.
  22. Napoli, C., Lemieux, C., and Jorgensen, R. (1990). Introduction of a Chalcone Synthase Gene into Petunia Results in Reversible Co-Suppression of Homologous Genes in trans. Plant Cell 2, 279-289.
  23. Nykanen, A., Haley, B., and Zamore, P. D. (2001). ATP Requirements and Small Interfering RNA Structure in the RNA Interference Pathway. Cell 107, 309-321.
  24. Plasterk, R. H. A., and Ketting, R. F. (2000). The Silence of the Genes. Curr. Opin. Genet. Dev. 10, 562-567.
  25. Provost, P., Dishart, D., Doucet, J., Frendewey, D., Samuelsson, B., and Radmark, O. (2002). Ribonuclease Activity and RNA Binding of Recombinant Human Dicer. EMBO J. 21, 5864-5874.
  26. Romano, N., and Macino, G. (1992). Quelling: Transient Inactivation of Gene Expression in Neurospora crassa by Transformation with Homologous Sequences. Mol. Microbiol. 6, 3343-3353.
  27. Smith, C. J., Watson, C. F., Bird, C. R., Ray, J., Schuch, W., and Grierson, D. (1990). Expression of a Truncated Tomato Polygalacturonase Gene Inhibits Expression of the Endogenous Gene in Transgenic Plants. Mol. Gen. Genet. 224, 477-481.
  28. Stark, G. R., Kerr, I. M., Williams, B. R., Silverman, R. H., and Schreiber, R. D. (1998). How Cells Respond to Interferons. Annu. Rev. Biochem. 67, 227-264.
  29. van der Krol, A. R., Mur, L. A., Beld, M., Mol, J. N., and Stuitje, A. R. (1990). Flavonoid Genes in Petunia: Addition of a Limited Number of Gene Copies May Lead to a Suppression of Gene Expression. Plant Cell 2, 291-299.
  30. Voinnet, O., Pinto, Y. M., and Baulcombe, D. C. (1999). Suppression of Gene Silencing: A General Strategy Used by Diverse DNA and RNA Viruses of Plants. Proc. Natl. Acad. Sci. USA 96, 14147-14152.
  31. Yu, J. Y., DeRuiter, S. L., and Turner, D. L. (2002). RNA Interference by Expression of Short-interfering RNAs and Hairpin RNAs in Mammalian Cells. Proc. Nat. Acad. Sci. USA 99, 6047-6052.
  32. Zamore, P. D. (2001). RNA Interference: Listening to the Sound of Silence. Nat. Struct. Biol. 8, 746-750.
LT068