Search Thermo Fisher Scientific
TO-PRO-3 is a cell-impermeant dye that gives strong and selective staining of the nucleus in cultured cells and paraffin sections. TO-PRO-3 and other carbocyanine monomers have very strong binding affinity for dsDNA, with dissociation constants in the micromolar range. The long-wavelength fluorescence of TO-PRO-3 is outside the spectral range of tissue autofluorescence as well as commonly used fluorophores in the DAPI and FITC channels.
First, fix and permeabilize cultured cells with a protocol appropriate for your sample.
|
|
![]() |
![]() |
![]() |
![]() |
![]() |
TO-PRO-3 | |
---|---|
Excitation/Emission (nm) | 642/661 |
Standard filter set | Far Red |
EVOS Light Cube | Cy®5 |
Storage conditions | ≤–20°C |
Muntjac fibroblast labeled with probes for actin and the nucleus. The actin cytoskeleton was visualized using the green-fluorescent Alexa Fluor 488 phalloidin. The nucleus was stained with TO-PRO-3 reagent.