sho-readyprobes-red
Membrane integrity–based viability assay
NucRed Dead 647 ReadyProbes Reagent is a cell-impermeant stain that emits bright far-red fluorescence when bound to DNA. Since this reagent stains cells that lack plasma membrane integrity, it is extremely useful to measure viability and to measure cytotoxicity in kinetic live-cell assays. NucRed Dead 647 reagent is also suitable for staining nuclei in fixed-cell preparations and tissue sections.
This protocol can be used for:
  • Identifying dead cells using a fluorescence microscope
This protocol should not be used for:
  • Flow cytometry
You will need the following for this protocol:
Protocol
Grow-red
1. Culture cells in appropriate medium and vessel for microscopy
Add-Dropp-Red
2. Add 2 drops NucRed® Dead 647 reagent per milliliter of medium to label cells
Clock-5
3. Incubate 5–15 minutes
Image
4. Image cells
Spectral information and storage
 NucRed Dead 647 ReadyProbes Reagent
Excitation/Emission642/661 nm
Standard filter setCy5
EVOS Light CubeCy5
Storage conditionsRoom temperature
 Protocol tips
hela647
Tubulin and nuclear staining in HeLa cells. HeLa cells were fixed and permeabilized and stained for tubulin (biotinylated mouse anti-a-tubulin mouse monoclonal antibody followed by an Alexa Fluor 488 goat anti-mouse secondary antibody and counter-stained with NucRed Dead 647 ReadyProbes reagent.
Ordering information
Catalog # Name Size Price (USD) Qty
R37113 NucRed™ Dead 647 ReadyProbes™ Reagent (TO-PRO-3 iodide) Each
339.65
Online Exclusive
368.00
Save 28.35 (8%)