HCS LIVE/DEAD Green Kit
Two-color nuclear staining assay for cell viability
This kit includes Image-iT DEAD Green viability stain for discrimination of dead cells and HCS NuclearMask Deep Red stain or Hoechst 33342 for total cell demarcation. Once the stains in this kit are used on your live-cell sample, the signal from the stains will be retained if the sample is fixed and permeabilized.
This protocol can be used for:
  • Identifying dead cells using high-content imaging
This protocol should not be used for:
  • Flow cytometry
You will need the following for this protocol:
Protocol
Grow-red
1. Culture cells in appropriate medium in a 96-well plate
Add-2
2. Add 2.1 μL Image-iT DEAD Green viability stain (Component A) to 6 mL complete medium to create staining solution
Add-2
3. Add 3 mL 16% paraformaldehyde and 18 μL Hoechst 33342 (Component B) to 9 mL PBS to create fixation solution
Add-gray
4. Add test compound or drug to cells to a total volume of 125 μL and incubate as desired
Add-gray
5. Add 50 μL Image-iT DEAD Green staining solution to each well for a total volume of 175 µL
Clock-30
6. Incubate at 37°C for 30 minutes
Remove
7. Remove medium
Add-gray
11. Wash wells once with PBS
Clock-30
6. Incubate at 37°C for 30 minutes
Remove
7. Remove medium
Remove-gray
8. Add 100 μL fixation solution to each well
Clock-15
9. Incubate at room temperature for 15 minutes
Remove
10. Remove the fixation solution
Wash
11. Wash wells once with PBS
Add-gray
12. Add 100 μL PBS to each well
Image
13. Analyze cells on a high-content imaging instrument
Spectral information and storage
 Image-iT DEAD Green stainHoechst 33342
Excitation/Emission488/515 nm350/461 nm
Standard filter setFITC or GFP 
EVOS Light CubeGFPDAPI
Storage conditions–20°C–20°C
 Protocol tips
  • This stains in this kit are compatible with antibody labeling protocols
  • This kit provides sufficient material for two 96-well plates
Dose-response_
Dose-response for valinomycin in HeLa cells using the HCS LIVE/DEAD Green Kit.
Ordering information
Catalog # Name Size Price (USD) Qty
H10290 HCS LIVE/DEAD™ Green Kit Each
482.00