ReadyProbes Cell Viability Imaging Kit, Blue-Green
Two-color nuclear staining assay for cell viability
This kit can be used to quickly and easily determine the viability of cells. NucBlue Live reagent stains the nuclei of all cells, while NucGreen Dead reagent stains only the nuclei of dead cells.
This protocol can be used for:
  • Identifying live and dead cells using a fluorescence microscope
This protocol should not be used for:
  • Flow cytometry
You will need the following for this protocol:
Protocol
1. Culture cells in appropriate medium and vessel for microscopy
2. Add 2 drops each NucBlue Live and NucGreen Dead reagents per milliliter of medium to label cells
3. Incubate 5–30 minutes
4. Image cells
Spectral information and storage
 NucBlue LiveNucGreen Dead
Excitation/Emission360/460 nm504/523 nm
Standard filter setDAPIFITC/GFP
EVOS Light CubeDAPIGFP
Storage conditionsRoom temperatureRoom temperature
Protocol tips
jurket
Ordering information
Catalog # Name Size Price (USD) Qty
R37609 ReadyProbes™ Cell Viability Imaging Kit, Blue/Green Each
406.00