48ab - Appl/Samples/Products/Resources/Contact
Hot stage microscopy
Real-world materials applications often take place under variable environmental conditions, including high temperatures. The behavior of your heated material as it recrystallizes, melts, and deforms can inform critical macro- and microscopic observations, such as how a manufactured part might respond to stress or how feed materials behave during production. As a sample’s response to heat is a dynamic process, it must be paired with dynamic observation for accurate insight. Modern heating stages in electron microscopes allow for in situ experiments for high-resolution observation of heated materials. These demanding experiments are capable of linking sample morphology, environment, and thermodynamics, and can help you control the corresponding behavior of the bulk material.
SEM heating stages
There are many considerations when operating electron microscopes at elevated temperatures, such as the desired temperature range, sample size, and chemical environment. The following table shows what is possible with Thermo Scientific temperature stages.
Style Sheet for Products Table Specifications
Name
Application
Temperature
Max. Sample Size
Environment
High vacuum heating stage
General-purpose heating, high resolution imaging, in-column detection, fast processes, electron-backscatter diffraction (EBSD)
Up to 1100°C
(EBSD up to 900°C)
10 mm
High vacuum
Environmental SEM (ESEM) stage
Heating in gaseous environment: oxidation or other chemical reactions
Up to 1000°C or 1400°C, depending on the model
5 mm
ESEM
µHeater
Powder heating, chunk lift-out studies (DualBeam), STEM imaging, high-temperature EBSD and EDS, ramp rate of 10,000°C/s
Up to 1200°C
50 µm
Any
Cooling stage, WetSTEM
Precise control over humidity, wetting studies, modest heating
-20°C to +60°C
3 mm
ESEM
Gold on a silicon substrate heated with a SEM hot stage.
Gold on a silicon substrate at approximately 1080°C. The High Vacuum Heating Stage allows all in-lens detectors and imaging modes to be used to image the sample with excellent resolution and contrast.
Resources
Mixture of magnetite and hematite nanoparticles heated at 1030 °C.
Backscattered electron image (left) and EDS maps of iron and oxygen (right) acquired simultaneously.
Texture development on implant material. As the temperature increases from 700 °C to 1300 °C
we can observe a completely different surface structure. Pressure: 120 Pa.
Two-phase Co-Sb alloy during heating to 700°C on the High Vacuum Heating Stage. The antimony-rich
phase sublimated during heating, causing exposure of the second phase.
Hydrophobic paper
Surface of hydrophobic treated paper - water condensation appears as droplets on top of the paper, rather than wetting the fibers. This experiment was carried out by controlling chamber pressure and stage temperature in a Quattro ESEM.
Video Player is loading.
Current Time 0:00
Duration 0:08
Loaded: 100.00%
Stream Type LIVE
Remaining Time 0:08
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
      1. Now Playing
        Up NextHydrophobic paper
        Surface of hydrophobic treated paper - water condensation appears as droplets on top of the paper, rather than wetting the fibers. This experiment was carried out by controlling chamber pressure and stage temperature in a Quattro ESEM.
      2. Now Playing
        Up NextExpancel
        Akzo Nobel Expancel Microspheres, a filler material that expands with increased temperature, is observed in situ using an ESEM heating stage. Working at elevated chamber pressure allows the system to deal with any outgassing that is produced.
      3. Now Playing
        Up NextSuper-absorption
        Sodium polyacrylate is a super-absorbent polymer with broad application in consumer products. Here, humidity is introduced into the SEM chamber and swelling of the material is observed in situ.
      4. Now Playing
        Up NextMelting solder
        The wetting behavior of solder droplets is studied in situ using an ESEM heating stage.
      5. Now Playing
        Up NextMelting of 70% chocolate
        Melting of 70% chocolate, under precisely controlled temperature and humidity conditions.
      6. Now Playing
        Up NextFreezing tomato
        Freezing can affect food structure, such as this tomato surface. Using a cooling stage, formation and sublimation of ice is studied.
    Mixture of magnetite and hematite nanoparticles heated at 1030 °C.
    Backscattered electron image (left) and EDS maps of iron and oxygen (right) acquired simultaneously.
    Texture development on implant material. As the temperature increases from 700 °C to 1300 °C
    we can observe a completely different surface structure. Pressure: 120 Pa.
    Two-phase Co-Sb alloy during heating to 700°C on the High Vacuum Heating Stage. The antimony-rich
    phase sublimated during heating, causing exposure of the second phase.
    Hydrophobic paper
    Surface of hydrophobic treated paper - water condensation appears as droplets on top of the paper, rather than wetting the fibers. This experiment was carried out by controlling chamber pressure and stage temperature in a Quattro ESEM.
    Video Player is loading.
    Current Time 0:00
    Duration 0:08
    Loaded: 100.00%
    Stream Type LIVE
    Remaining Time 0:08
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • default, selected
        1. Now Playing
          Up NextHydrophobic paper
          Surface of hydrophobic treated paper - water condensation appears as droplets on top of the paper, rather than wetting the fibers. This experiment was carried out by controlling chamber pressure and stage temperature in a Quattro ESEM.
        2. Now Playing
          Up NextExpancel
          Akzo Nobel Expancel Microspheres, a filler material that expands with increased temperature, is observed in situ using an ESEM heating stage. Working at elevated chamber pressure allows the system to deal with any outgassing that is produced.
        3. Now Playing
          Up NextSuper-absorption
          Sodium polyacrylate is a super-absorbent polymer with broad application in consumer products. Here, humidity is introduced into the SEM chamber and swelling of the material is observed in situ.
        4. Now Playing
          Up NextMelting solder
          The wetting behavior of solder droplets is studied in situ using an ESEM heating stage.
        5. Now Playing
          Up NextMelting of 70% chocolate
          Melting of 70% chocolate, under precisely controlled temperature and humidity conditions.
        6. Now Playing
          Up NextFreezing tomato
          Freezing can affect food structure, such as this tomato surface. Using a cooling stage, formation and sublimation of ice is studied.
      Applications
      Fundamental Materials Research_R&D_Thumb_274x180_144DPI
      Fundamental Materials Research
      Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
      Samples
      Battery Research
      Battery development is enabled by multi-scale analysis with microCT, SEM and TEM, Raman spectroscopy, XPS, and digital 3D visualization and analysis. Learn how this approach provides the structural and chemical information needed to build better batteries.
      Learn more ›
      Metals Research and Characterization
      Effective production of metals requires precise control of inclusions and precipitates. Our automated tools can perform a variety of tasks critical for metal analysis including; nanoparticle counting, EDS chemical analysis and TEM sample preparation.
      Learn more ›
      Polymers Research
      Polymer microstructure dictates the material’s bulk characteristics and performance. Electron microscopy enables comprehensive microscale analysis of polymer morphology and composition for R&D and quality control applications.
      Learn more ›
      Nanoparticles
      Materials have fundamentally different properties at the nanoscale than at the macroscale. To study them, S/TEM instrumentation can be combined with energy dispersive X-ray spectroscopy to obtain nanometer, or even sub-nanometer, resolution data.
      Learn more ›
      Forensics
      Micro-traces of crime scene evidence can be analyzed and compared using electron microscopy as part of a forensic investigation. Compatible samples include glass and paint fragments, tool marks, drugs, explosives, and GSR (gunshot residue).
      Learn more ›
      Catalysis Research
      Catalysts are critical for a majority of modern industrial processes. Their efficiency depends on the microscopic composition and morphology of the catalytic particles; EM with EDS is ideally suited for studying these properties.
      Learn more ›
      Automotive Materials Testing
      Every component in a modern vehicle is designed for safety, efficiency, and performance. Detailed characterization of automotive materials with electron microscopy and spectroscopy informs critical process decisions, product improvements, and new materials.
      Learn more ›
      Fibers and Filters
      The diameter, morphology and density of synthetic fibers are key parameters that determine the lifetime and functionality of a filter. Scanning electron microscopy (SEM) is the ideal technique for quickly and easily investigating these features.
      Learn more ›
      Products
      Style Sheet for Komodo Tabs
      Apreo ChemiSEM System
      • Integrated SEM imaging and chemical characterization
      • Enhanced automation to simplify workflows
      • Extended source lifetime and schedulable upgrades
      Helios 5 HX/Helios 5 UX/Helios 5 FX DualBeam
      • Fully automated, high-quality, ultra-thin TEM sample preparation
      • High throughput, high resolution subsurface and 3D characterization
      • Rapid nanoprototyping capabilities
      Helios 5 PFIB DualBeam
      • Gallium-free STEM and TEM sample preparation
      • Multi-modal subsurface and 3D information
      • Next-generation 2.5 μA xenon plasma FIB column
      Axia ChemiSEM
      • Live quantitative elemental mapping
      • High fidelity scanning electron microscopy imaging
      • Flexible and easy to use, even for novice users
      • Easy maintenance
      Thermo Scientific Apreo 2 scanning electron microscope (SEM)
      Apreo 2 SEM
      • High-performance SEM for all-round nanometer or sub-nanometer resolution
      • In-column T1 backscatter detector for sensitive, TV-rate materials contrast
      • Excellent performance at long working distance (10 mm)
      Thermo Scientific Prisma E scanning electron microscope (SEM)
      Prisma E SEM
      • Entry-level SEM with excellent image quality
      • Easy and quick sample loading and navigation for multiple samples
      • Compatible with a wide range of materials thanks to dedicated vacuum modes
      Thermo Scientific Verios 5 XHR scanning electron microscope (SEM)
      Verios 5 XHR SEM
      • Monochromated SEM for sub-nanometer resolution over the full 1 keV to 30 keV energy range
      • Easy access to beam landing energies as low as 20 eV
      • Excellent stability with piezo stage as standard
      Thermo Scientific Quattro E scanning electron microscope (SEM)
      Quattro ESEM
      • Ultra-versatile high-resolution FEG SEM with unique environmental capability (ESEM)
      • Observe all information from all samples with simultaneous SE and BSE imaging in every mode of operation
      Phenom ProX G6 Desktop SEM
      • High performance desktop SEM with integrated EDS detector
      • Resolution <6 nm (SE) and <8 nm (BSE); magnification up to 350,000x
      • Optional SE detector
      Phenom Pro G6 Desktop SEM
      • High performance desktop SEM
      • Resolution <6 nm (SE) and <8 nm (BSE); magnification up to 350,000x
      • Optional SE detector
      Phenom Pure G6 Desktop SEM
      • Entry level desktop SEM
      • Resolution <15 nm; magnification up to 175,000x
      • Longlife CeB6 source
      Style Sheet for Instrument Cards Original
      Style Sheet to change H2 style to p with em-h2-header class
      Contact us
      Style Sheet for Support and Service footer
      Style Sheet for Fonts
      Style Sheet for Cards
      Electron microscopy services for
      the materials science
      To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.