Ambient air particulate monitors - Identify, define, and inform
The knowledge to do more than inform can make all the differences when it comes to monitoring particulates. Our ambient air particulate monitors use advanced particulate monitoring technologies such as gravimetric sampling, beta attenuation, and light scattering to ensure your monitoring needs are met. Our Partisol air samplers, Tapered Element Oscillating Microbalance (TEOM), Beta Attenuation, and SHARP air quality monitors reliably measure critical regulatory parameters including PM10 and PM2.5 present in ambient air environment.
Beta attenuation and SHARP particulate monitors
The principle behind beta attenuation particulate sampling instruments (beta gauge) is that energy is absorbed from beta particles as they pass through particulate matter (PM) collected on a filter media. Our beta gauge instruments have been designed to take advantage of this scientific principle to accurately monitor and measure PM concentrations.
Reliable and precise beta gauges
cars, planes, smokestacks showing pollution
Volatile aerosols remain intact for accurate measurement
Our continuous particulate monitors use the radiometric principle of beta attenuation through a known area on a fibrous filter tape to continuously detect the mass of deposited ambient particles.
Thermo Scientific 5014i Beta Continuous Ambient Particulate Monitor
Thermo Scientific 5014i Beta Continuous Ambient Particulate Monitor
Thermo Scientific 5014iQ Beta Attenuation Monitor
Thermo Scientific 5014iQ Beta Attenuation Monitor
Thermo Scientific Model 5030i SHARP Monitor
Thermo Scientific Model 5030i SHARP Monitor
Thermo Scientific 5030iQ SHARP Monitor
Thermo Scientific 5030iQ SHARP Monitor
Thermo Scientific Model 5028i Continuous Particulate Monitor
Thermo Scientific Model 5028i Continuous Particulate Monitor
Partisol air samplers
Our partisol monitors measure particles in air by drawing air through a pre-weighed filter at a known flow rate for specified amounts of time, allowing for unattended daily sampling of particulate matter.
Gravimetric particulate samplers 
industrial smokestacks emitting pollution
Collect high quality samples of fine and coarse ambient particulate matter
Our instruments are designed for operational simplicity; performance audits and stored data retrieval are made possible through an embedded microprocessor and menu-driven software.
Thermo Scientific Partisol 2000i Air Sampler
Thermo Scientific Partisol 2000i Air Sampler
Thermo Scientific Partisol 2000i-D Dichotomous Air Sampler
Thermo Scientific Partisol 2000i-D Dichotomous Air Sampler
Thermo Scientific Partisol 2025i Sequential Air Sampler
Thermo Scientific Partisol 2025i Sequential Air Sampler
Thermo Scientific Partisol 2025i-D Dichotomous Sequential Air Sampler
Thermo Scientific Partisol 2025i-D Dichotomous Sequential Air Sampler
TEOM continuous particulate monitors
Tapered element oscillating microbalances technology (TEOM) are gravimetric solutions that draw conditioned ambient air through a filter at a constant flow rate, continuously weighing the filter and calculating near real-time mass concentrations of particulate matter.
Continuously and reliably monitor particulates in ambient air
industrial city with soot and ash coming out of smokestacks
Measure critical regulatory parameters
Our advanced air sampling technology measures critical regulatory parameters including PM-10 and PM-2.5 mass concentration as it exists in ambient air.
Thermo Scientific 1405 TEOM Continuous Ambient Particulate Monitor
Thermo Scientific 1405 TEOM Continuous Ambient Particulate Monitor
Thermo Scientific 1405-D TEOM Continuous Dichotomous Ambient Particulate Monitor
Thermo Scientific 1405-D TEOM Continuous Dichotomous Ambient Particulate Monitor
Thermo Scientific 1405-F TEOM Continuous Ambient Air Monitor
Thermo Scientific 1405-D 1405-F TEOM Continuous Ambient Air Monitor
Thermo Scientific 1405-DF TEOM Continuous Dichotomous Ambient Air Monitor
Thermo Scientific 1405-DF TEOM Continuous Dichotomous Ambient Air Monitor
Service and product availability may vary by country and are subject to varying regulatory requirements. Please contact your local sales representative for availability.