E-Gel Power Snap Systems are compact benchtop instruments that have integrated nucleic acid electrophoresis and gel imaging functionalities. The instrument has two main components: the instrument device base and the camera to capture gel images. The instrument works with precast E-Gel agarose gels, which are buffer-less gels containing SYBR safe DNA stains, and ladders. This eliminates the need to pour gels, prepare buffers, and handle toxic dyes such as ethidium bromide.
The components of the E-Gel Power Snap systems are designed to save time and streamline the nucleic acid electrophoresis workflow. This system facilitates a simple three-step workflow: load, run, and analyze (Figure 1).
Figure 1. Three-step workflow of E-Gel electrophoresis systems
Advantages of E-Gel electrophoresis device and imaging systems
The two models of the E-Gel Precast Agarose Gel Electrophoresis Systems, including the Power Snap and Power Snap Plus, offer flexible sample throughputs and are upgrades over historical gel analyzers and E-Gel imagers, allowing faster setup, gel runs, and analysis. Both systems include:
Real-time visualization: Amber filter to monitor real-time sample migration
Sample safety: Blue-light transilluminator does not damage DNA samples
Intuitive interface: Easy operation with user-friendly touch screen and preset protocols
Flexible throughput: Gels with 11-, 22-, 48-, and 96-wells can be run (model-dependent)
High resolution camera for image capture, with image editing capabilities
Internal storage: Up to 3 months of experimental data can be stored in the device
Small footprint: 6x smaller than traditional gel documentation systems
Maintain productivity with extended warranties Rapid Exchange (REX) one-year extended warranties are available (at an additional charge) for both the E-Gel Power Snap Plus device and camera. If your instrument needs repair, just call our support center and we will exchange your instrument for a factory-certified, refurbished replacement instrument within one business day from the time we determine a replacement is required. Replacement instruments are typically received within three days of shipment and are yours to keep. All parts, labor, and shipping charges are included, and existing warranty contracts will be transferred to replacement units.
Beginning of dialog window. Escape will cancel and close the window.
End of dialog window.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
E-Gel Power Snap Plus System
Learn how to perform high throughput gel electrophoresis, monitor DNA migration, and capture gel images with the E-Gel Power Snap Plus System. Find out about the image editing and image storage options.
Video Player is loading.
Current Time 0:00
/
Duration 1:06
Loaded: 14.77%
0:00
Stream Type LIVE
Remaining Time -1:06
1x
Chapters
descriptions off, selected
captions settings, opens captions settings dialog
captions off, selected
en (Main), selected
This is a modal window.
Beginning of dialog window. Escape will cancel and close the window.
End of dialog window.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
E-Gel Power Snap Plus How-to: Deconvolution Overview
Discover the unique features of the Power Snap Plus E-Gel system, including how to easily read results with on‑instrument deconvolution and analysis for Invitrogen E-Gel 96-well gels.
Introducing the E-Gel Power Snap
Electrophoresis System
Video Player is loading.
Current Time 0:00
/
Duration 1:16
Loaded: 12.81%
0:00
Stream Type LIVE
Remaining Time -1:16
1x
Chapters
descriptions off, selected
captions settings, opens captions settings dialog
captions off
English Captions, selected
French (France) Captions
Chinese (Simplified) Captions
en (Main), selected
This is a modal window.
Beginning of dialog window. Escape will cancel and close the window.
End of dialog window.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
Invitrogen E-Gel Power Snap System overview
Simplify electrophoresis with the Invitrogen E-Gel Power Snap System, the platform that integrates gel running and imaging functions. Watch to learn the benefits of the E-Gel Power Snap Electrophoresis System.
Video Player is loading.
Current Time 0:00
/
Duration 1:51
Loaded: 8.84%
0:00
Stream Type LIVE
Remaining Time -1:51
1x
Chapters
descriptions off, selected
captions settings, opens captions settings dialog
captions off, selected
en (Main), selected
This is a modal window.
Beginning of dialog window. Escape will cancel and close the window.
End of dialog window.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
Invitrogen E-Gel Power Snap System features
Explore the features of the E-Gel Power Snap Electrophoresis System including the gel image capture and editing functions.
Video Player is loading.
Current Time 0:00
/
Duration 2:21
Loaded: 6.96%
0:00
Stream Type LIVE
Remaining Time -2:21
1x
Chapters
descriptions off, selected
captions settings, opens captions settings dialog
captions off, selected
en (Main), selected
This is a modal window.
Beginning of dialog window. Escape will cancel and close the window.
End of dialog window.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
How to quickly purify DNA with E-Gel CloneWell II precast gels
Watch a demo of how to gel purify DNA using the E-Gel Power Snap System and E-Gel CloneWell II gel. Load samples, run the gel, capture bands of interest in the recovery well, and pipette out purified DNA bands. No additional purification steps needed.
INSTRUCTOR: In this
video, we will show you
Video Player is loading.
Current Time 0:00
/
Duration 1:32
Loaded: 10.59%
0:00
Stream Type LIVE
Remaining Time -1:32
1x
Chapters
descriptions off, selected
captions settings, opens captions settings dialog
captions off
English Captions, selected
en (Main), selected
This is a modal window.
Beginning of dialog window. Escape will cancel and close the window.
End of dialog window.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
How to perform rapid electrophoresis using an E-Gel Power Snap device and E-Gel EX Cassette
Watch a demo of agarose gel electrophoresis using an E-Gel Power Snap System and E-Gel EX Cassette. The ease and simplicity of the process is ideal for research labs.
iBright Image Analysis Software analyzes data from E-Gel images captured by the integrated camera. The gel images and the results of data analysis can be stored on Thermo Fisher’s Connect Platform. iBright Image Analysis is included in the E-Gel Power Snap Plus Electrophoresis System.
Invitrogen E-Gel products to complete your gel electrophoresis system
For best results, the E-Gel Power Snap and Power Snap Plus System must be used in conjunction with other Invitrogen E-Gel line of products, which include:
Invitrogen E-Gel precast agarose gels
E-Gel Precast Agarose Gels are ideal for fast and convenient results of DNA and RNA fragments and are available in a variety of percentages and well formats.