The plastic and glass surfaces of pipette tips and tubes including PCR tubes, microcentrifuge tubes, and conical tubes, are an easily overlooked source of ribonuclease (RNase) contamination. Thermo Fisher offers a range of RNase-free plastics including pipette tips, PCR tubes, microcentrifuge tubes, and conical tubes. Each lot of Invitrogen brand tips and tubes undergoes rigorous testing to ensure that they are nuclease-free (RNase- and DNase-free).
Basics of RNase control Working with RNA
RNase-free, nuclease-free tips and tubes
RNase-free, nuclease-free lab supplies are essential in lab spaces where researchers work with RNA. Common lab supplies such as pipette tips, PCR tubes, microcentrifuge tubes, and conical tubes are offered in an RNase-free format.
RNase-free pipette tips
Invitrogen RNase-free Tips, 200 μL size product photo
For safe transfer of samples and reagents, RNase-free pipette tips come in two different formats: without a barrier/filter and with a barrier/filter (also known as barrier tips or filter tips). Both formats have the following features:
  • E-beam sterilized—nuclease-free and RNase-free
  • Autoclave-safe
  • Compatible with a broad range of pipettors
Ordering information for RNase-free pipette tips
Catalog # Name Size Price (USD) Qty
AM12650 RNase-free Tips, 200 μL size 10 racks
181.00
AM12635 Barrier (Filter) Tips, 10 μL size (compatible with Eppendorf™ pipettors) 10 racks
259.65
Online Exclusive
267.00
Save 7.35 (3%)
AM12645 Barrier (Filter) Tips, 20 μL size 10 racks
257.65
Online Exclusive
265.00
Save 7.35 (3%)
AM12648 Barrier (Filter) Tips, 100 μL size 10 racks
260.00
AM12655 Barrier (Filter) Tips, 200 μL size 10 racks
261.00
RNase-free PCR tubes
nvitrogen PCR tubes with caps, RNase-free, 0.2 mL (8-strip format) product photo
RNase-free PCR tubes are available in different varieties. RNase-free PCR tubes offer thinner walls to allow for better sample temperature regulation, as well as frosted surfaces for easy labeling. Other options include domed or flat caps, as well as 0.2 mL and 0.5 mL formats. Thermo Fisher RNase-free PCR tubes have the following features:
  • Certified nuclease-free and RNase-free
  • Allow uniform contact with the thermal cycler temperature block
  • Autoclave-safe
Ordering information for RNase-free PCR tubes
Catalog # Name Size Price (USD) Qty
AM12225 Thin-walled, frosted lid, RNase-free PCR tubes (0.2 mL) Each
219.65
Online Exclusive
226.00
Save 6.35 (3%)
AM12230 PCR Tubes & Caps, RNase-free, 0.2 mL (8-strip format) Each
269.65
Online Exclusive
278.00
Save 8.35 (3%)
AM12250 Thin-walled, dome cap, RNase-free PCR tubes (0.5 mL) Each
174.00
AM12275 Thin-walled, frosted lid, RNase-free PCR tubes (0.5 mL) Each
181.65
Online Exclusive
187.00
Save 5.35 (3%)
RNase-free microcentrifuge tubes
Invitrogen Nonstick, RNase-free Microfuge Tubes, 0.5 mL product photo
RNase-free microcentrifuge tubes are available in three different sizes (0.5 mL, 1.5 mL, and 2.0 mL) and have marked gradations for convenience. Our microcentrifuge tubes are lot tested for contaminants, such as nucleases including RNases and DNases. All our RNase-free microcentrifuge tubes have the following features:
  • Certified nuclease-free and RNase-free
  • Non-stick, low-binding surface
  • Positive-seal design that prevents caps from opening in boiling water baths
  • Autoclave-safe
Ordering information for RNase-free microcentrifuge tubes
Catalog # Name Size Price (USD) Qty
AM12300 Nonstick, RNase-free Microfuge Tubes, 0.5 mL, 1000 Tubes Each
136.65
Online Exclusive
140.00
Save 3.35 (2%)
AM12350 Nonstick, RNase-free Microfuge Tubes, 0.5 mL, 500 Tubes Each
111.65
Online Exclusive
115.00
Save 3.35 (3%)
AM12400 RNase-free Microfuge Tubes, 1.5 mL, 500 Tubes Each
113.65
Online Exclusive
128.00
Save 14.35 (11%)
AM12425 RNase-free Microfuge Tubes, 2.0 mL, 500 Tubes Each
108.65
Online Exclusive
121.00
Save 12.35 (10%)
AM12475 Nonstick, RNase-free Microfuge Tubes, 2.0 mL, 250 Tubes Each
99.65
Online Exclusive
103.00
Save 3.35 (3%)
RNase-free conical tubes for safe storage of samples and reagents from RNases
RNase-free conical tubes are available in two different sizes (15 mL and 50 mL) and have printed writing space and volume marks for convenience. Our RNase-free conical tubes have the following features:
  • Certified nuclease-free and RNase-free tubes come racked and are pre-assembled with green screw tops
  • Tubes come racked and are pre-assembled with green screw tops
  • Autoclave-safe
Ordering information for RNase-free conical tubes
Catalog # Name Size Price (USD) Qty
AM12500 Conical Tubes (15 mL) (racked) Each
353.65
Online Exclusive
370.00
Save 16.35 (4%)
AM12501 Conical Tubes (50 mL) (racked) Each
204.65
Online Exclusive
213.00
Save 8.35 (4%)
RNase-free pipette tips
Invitrogen RNase-free Tips, 200 μL size product photo
For safe transfer of samples and reagents, RNase-free pipette tips come in two different formats: without a barrier/filter and with a barrier/filter (also known as barrier tips or filter tips). Both formats have the following features:
  • E-beam sterilized—nuclease-free and RNase-free
  • Autoclave-safe
  • Compatible with a broad range of pipettors
Ordering information for RNase-free pipette tips
Catalog # Name Size Price (USD) Qty
AM12650 RNase-free Tips, 200 μL size 10 racks
181.00
AM12635 Barrier (Filter) Tips, 10 μL size (compatible with Eppendorf™ pipettors) 10 racks
259.65
Online Exclusive
267.00
Save 7.35 (3%)
AM12645 Barrier (Filter) Tips, 20 μL size 10 racks
257.65
Online Exclusive
265.00
Save 7.35 (3%)
AM12648 Barrier (Filter) Tips, 100 μL size 10 racks
260.00
AM12655 Barrier (Filter) Tips, 200 μL size 10 racks
261.00
RNase-free PCR tubes
nvitrogen PCR tubes with caps, RNase-free, 0.2 mL (8-strip format) product photo
RNase-free PCR tubes are available in different varieties. RNase-free PCR tubes offer thinner walls to allow for better sample temperature regulation, as well as frosted surfaces for easy labeling. Other options include domed or flat caps, as well as 0.2 mL and 0.5 mL formats. Thermo Fisher RNase-free PCR tubes have the following features:
  • Certified nuclease-free and RNase-free
  • Allow uniform contact with the thermal cycler temperature block
  • Autoclave-safe
Ordering information for RNase-free PCR tubes
Catalog # Name Size Price (USD) Qty
AM12225 Thin-walled, frosted lid, RNase-free PCR tubes (0.2 mL) Each
219.65
Online Exclusive
226.00
Save 6.35 (3%)
AM12230 PCR Tubes & Caps, RNase-free, 0.2 mL (8-strip format) Each
269.65
Online Exclusive
278.00
Save 8.35 (3%)
AM12250 Thin-walled, dome cap, RNase-free PCR tubes (0.5 mL) Each
174.00
AM12275 Thin-walled, frosted lid, RNase-free PCR tubes (0.5 mL) Each
181.65
Online Exclusive
187.00
Save 5.35 (3%)
RNase-free microcentrifuge tubes
Invitrogen Nonstick, RNase-free Microfuge Tubes, 0.5 mL product photo
RNase-free microcentrifuge tubes are available in three different sizes (0.5 mL, 1.5 mL, and 2.0 mL) and have marked gradations for convenience. Our microcentrifuge tubes are lot tested for contaminants, such as nucleases including RNases and DNases. All our RNase-free microcentrifuge tubes have the following features:
  • Certified nuclease-free and RNase-free
  • Non-stick, low-binding surface
  • Positive-seal design that prevents caps from opening in boiling water baths
  • Autoclave-safe
Ordering information for RNase-free microcentrifuge tubes
Catalog # Name Size Price (USD) Qty
AM12300 Nonstick, RNase-free Microfuge Tubes, 0.5 mL, 1000 Tubes Each
136.65
Online Exclusive
140.00
Save 3.35 (2%)
AM12350 Nonstick, RNase-free Microfuge Tubes, 0.5 mL, 500 Tubes Each
111.65
Online Exclusive
115.00
Save 3.35 (3%)
AM12400 RNase-free Microfuge Tubes, 1.5 mL, 500 Tubes Each
113.65
Online Exclusive
128.00
Save 14.35 (11%)
AM12425 RNase-free Microfuge Tubes, 2.0 mL, 500 Tubes Each
108.65
Online Exclusive
121.00
Save 12.35 (10%)
AM12475 Nonstick, RNase-free Microfuge Tubes, 2.0 mL, 250 Tubes Each
99.65
Online Exclusive
103.00
Save 3.35 (3%)
RNase-free conical tubes for safe storage of samples and reagents from RNases
RNase-free conical tubes are available in two different sizes (15 mL and 50 mL) and have printed writing space and volume marks for convenience. Our RNase-free conical tubes have the following features:
  • Certified nuclease-free and RNase-free tubes come racked and are pre-assembled with green screw tops
  • Tubes come racked and are pre-assembled with green screw tops
  • Autoclave-safe
Ordering information for RNase-free conical tubes
Catalog # Name Size Price (USD) Qty
AM12500 Conical Tubes (15 mL) (racked) Each
353.65
Online Exclusive
370.00
Save 16.35 (4%)
AM12501 Conical Tubes (50 mL) (racked) Each
204.65
Online Exclusive
213.00
Save 8.35 (4%)
TechNotes articles
Support
CMD Wide-format style fixes
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments