RNase-free Tips, 200 μL size
Customer information regarding this product here »
RNase-free Tips, 200 μL size
RNase-free Tips, 200 μL size
RNase-free Tips, 200 μL size
Invitrogen™

RNase-free Tips, 200 μL size

Ambion® Tips are RNase- and DNase-free and are provided in ten racks containing 8 x 12 tips each. They areRead more
Have Questions?
Catalog NumberQuantity
AM1265010 racks
Catalog number AM12650
Price (USD)
181.00
10 racks
-
Add to cart
Quantity:
10 racks
Recurring order eligible. Learn more »
Price (USD)
181.00
10 racks
Add to cart
Ask our AI about this Product
Ambion® Tips are RNase- and DNase-free and are provided in ten racks containing 8 x 12 tips each. They are compatible with Eppendorf®, Finnpipette, Oxford Benchmate®, Pipetman™, Proline®, Rainin, Socorex, and other brands of pipettors. Each lot of tips is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 racks
Shipping ConditionRoom Temperature
AutoclavableAutoclavable
FilteredNon-filtered
For Use With (Equipment)Universal Fit
FormatRack
Product LineInvitrogen
SterilitySterile
Tip StyleUniversal Fit
Volume (Metric)200 μL
Unit Size10 racks
Contents & Storage
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
Can I use CTAB rather than SDS in my sample buffer?
Where do I find buffer recipes for your precast protein gels?
Why is LDS (lithium dodecyl sulfate) used in the 4X NuPAGE sample buffer instead of SDS?
What is the recipe for traditional Laemmli Buffer?
If a Tricine gel is accidentally run with buffers used in the Tris-Glycine system, what will happen and why?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

瀏覽此商品的人,也瀏覽



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
22280602Certificate of AnalysisSep 17, 2022AM12650
32320402Certificate of AnalysisAug 26, 2021AM12650
30120402Certificate of AnalysisAug 02, 2021AM12650
19220402Certificate of AnalysisJun 24, 2021AM12650
19720402Certificate of AnalysisJun 21, 2021AM12650
5 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection