packaging, reagent, and dilution-free tubes for Bradford assay
Dye-based protein detection
Bradford assays are dye-binding assays for fast and simple protein quantification. The assay is performed at room temperature and no special equipment is required. Standard and unknown samples are added to pre-formulated Coomassie blue G-250 assay reagent and the resultant blue color is measured at 595 nm following a short room temperature incubation. Bradford protein assays are compatible with most salts, solvents, buffers, thiols, reducing substances, and metal chelating agents encountered in protein samples.
For added ease-of-use, select Bradford assays are now offered with Dilution-Free protein standards that help reduce assay setup time and pipetting errors. Dilution-Free protein standards are a set of prediluted standards packaged in a multichannel pipette-compatible format that enable easy transfer of a full dilution series to your microplate at once.
Selection guide  Technical handbook
Choose the right Bradford protein assay or reagent for your application
 packaging, reagents, and dilution-free tubes for Bradford protein assaypackaging and reagents for Bradford protein assaypackaging and reagents for Bradford protein assay
 Bradford Plus Protein Assay Kit with Dilution-Free BSA Protein StandardsDetergent Compatible Bradford AssayBradford Protein Assay
Features
  • Ready-to-use Bradford assay kit with Dilution-Free BSA Protein Standards for reduced standard curve preparation time
  • Increased linearity and reduced protein-to-protein variation compared to other commercial Bradford assay formulations
  • Ready-to-use modification of the Bradford assay with additional additives to make it compatible with 1% or higher of commonly used detergents, including Triton X-100 and NP-40 detergents
  • Ready-to-use formulation of the popular assay reagent originally described by Bradford in 1976
Assay range (sample volume)125–1,500 µg/mL (5 µL)
Or
1–25 µg/mL (150 µL)
100–1,500 µg/mL (10 µL)
or
2–25 µg/mL (150 µL)
100–1,500 µg/mL (20 µL)
or
1–25 µg/mL (150 µL)
Compatible reagentsBuffer salts, metal ions, reducing agents, chelatorsDetergents, buffer salts, metal ions, reducing agents, chelatorsBuffer salts, metal ions, reducing agents, chelators
IncompatibilitiesDetergents>5% detergent solutionsDetergents
Incubation time10 min10 min10 min
Product size950 mL/kit450 mL/kit950 mL/kit
Cat. No.A55866
Related Bradford Plus Kit:
23236
2324623200
Related product: Pierce Dilution-Free BSA Protein Standards, Multichannel Pipette Compatible, 2 mg/mL
multichannel pipet transferring protein standards from the 8 connecting tube format
Effortless, Fast, Accurate
Select Bradford assays now include Dilution-Free BSA Protein Standards which are a set of seven prediluted BSA standards, uniquely designed for multichannel pipette compatibility.
The Pierce Dilution-Free protein standards minimize tedious dilutions and pipetting errors, delivering accurate results while reducing assay setup time. They are calibrated by direct comparison to purified BSA from the National Institute of Standards and Technology (NIST) to help ensure accurate standard curve generation.
Bradford assay principles
The Bradford assay was first described by Dr. Marion Bradford in 1976 and uses Coomassie G-250 dye in a colorimetric reagent for the detection and quantitation of total protein. Pierce Bradford Plus Protein Assays are modifications of the reagent first reported by Dr. Bradford.
Chemistry of Coomassie-based Bradford protein assays
In an acidic environment, proteins bind to Coomassie dye. This results in a spectral shift from the reddish-brown form of the dye (absorbance maximum at 465 nm) to the blue form (absorbance maximum at 610 nm). The difference between the two dye forms is greatest at 595 nm, making it the optimal wavelength to measure the blue color from the Coomassie dye–protein complex. If desired, the blue color can be measured at any wavelength between 575 nm and 615 nm. At the two extremes (575 nm and 615 nm), there is an approximate 10% decrease in the measured amount of color (absorbance) compared to that obtained at 595 nm. Development of color in Coomassie dye–based protein assays has been associated with the presence of certain basic amino acids—primarily arginine, lysine, and histidine—in the protein. Van der Waals forces and hydrophobic interactions also influence dye–protein binding. The number of Coomassie dye molecules bound to each protein is approximately proportional to the number of positive charges found on the protein. Free amino acids, peptides, and low molecular weight proteins do not produce color with Coomassie dye reagents. In general, the mass of a peptide or protein should be at least 3,000 Da for quantification with this reagent. In some applications, this can be an advantage.
Reaction schematic for Bradford protein assay
Figure 1. Reaction schematic for the Coomassie dye–based Bradford protein assays.
The main disadvantage of Bradford protein assays is their incompatibility with surfactants at concentrations routinely used to solubilize membrane proteins. In general, the presence of a surfactant in the sample, even at low concentrations, causes precipitation of the reagent. This limitation can be overcome by using Detergent Compatible Bradford Assay. In addition, the Coomassie dye reagent is highly acidic, so proteins with poor acid-solubility cannot be assayed with this reagent. Finally, Coomassie reagents result in about twice as much protein-to-protein variation as copper chelation-based assay reagents.
Product highlights
Bradford Plus Protein Assay Kit with Dilution-Free BSA Protein Standards
The Bradford Plus Protein Assay Kit with Dilution-Free BSA Protein Standards reduces assay setup by eliminating tedious standard curve generation. Dilution-Free BSA Protein Standards are available as part of the Bradford Plus Protein Assay or available separately for use with any Pierce Bradford Assay.
Detergent Compatible Bradford Assay
Pierce Detergent Compatible Bradford Assay Kit is a quick and ready-to-use modification of the well-known Bradford Assay for total protein quantitation. This formulation is compatible with up to 1% of commonly used detergents. Comparing Pierce Detergent Compatible Bradford Assay to the Bio-Rad DC Protein Assay, better sensitivity is seen with the Pierce Detergent Compatible Bradford Assay using common detergents. The range of the standard curve for the Pierce Detergent Compatible Bradford assay is 4 times broader than the range for the Bio-Rad DC assay.
Comparison between Bio-Rad DC assay vs Pierce Detergent Compatible Bradford assay
Figure 3. Bradford protein assay vs. the Bio-Rad DC protein assay. Each assay was performed in a microplate using BSA standards spiked with detergent or water (control), and followed the manufacturers’ instructions.
Table 1. Comparing Pierce Detergent Compatible Bradford Assay Kit with Bio-Rad DC Protein Assay Kit.
FeaturesPierce Detergent Compatible Bradford Assay KitBio-Rad DC Protein Assay Kit
Assay measurement (absorbance maximum)595 nm750 nm
Test tube assay sample volume50 μL100 μL
Microplate assay sample volume10 μL5 μL
Assay working range100-1,500 μg/mL200-1,500 μg/mL
Absorbance range (sensitivity)HighLow
Number of reagents in kit1 reagent3 reagents
Setup time10 min30 min
Incubation time10 min15 min
Total20 min45 min
Pierce 660 nm protein assay, catalog number 22662
Pierce 660 nm Protein Assay Kit
Assay range: microplate (sample volume)25 to 2,000 µg/mL (65 µL)
or
50 to 2,000 µg/mL (10 µL)
Incubation time and temperature5
Total assay time75
Absorbance660 nm
Compatible reagentsReducing agents, chelating agents, detergents
Incompatible reagentsHigh levels of detergents or SDS requires addition of Ionic detergent compatibility reagent
UniformityLess protein–protein variation than the Coomassie (Bradford) assay
Product size450 mL/kit
Cat. No.22662
Related product: Pierce Dilution-Free BSA Protein Standards, Multichannel Pipette Compatible, 2 mg/mL
Pierce 660 nm Assay principles
Chemistry of Pierce 660 nm Assay
The Pierce 660nm Protein Assay is based on the binding of a unique dye-metal complex to protein in acidic conditions that causes a shift in the dye's absorption maximum, which is measured at 660 nm. The dye-metal complex is reddish-brown and changes to green upon protein binding. The color change is produced by deprotonation of the dye at low pH facilitated by interactions with positively charged amino acid groups in proteins. Therefore, the dye interacts mainly with basic residues in proteins, such as histidine, arginine and lysine and to a lesser extent tyrosine, tryptophan and phenylalanine.
The color produced in the assay is stable and increases in proportion to a broad range of increasing protein concentrations, even in the presence of detergents and reducing agents that would be incompatible with Bradford and BCA Protein Assays. The optional IDCR may be added to the assay reagent to increase compatibility with high amounts of ionic detergents, allowing samples containing Laemmli SDS sample buffer with bromophenol blue to be measured. The IDCR completely dissolves by thorough mixing and does not have any effect on the assay.
Absorption maximum of Pierce 660 nm Protein Assay reagent
Figure 4. Absorption maximum of the 660 nm Assay Reagent-metal complex shifts proportionally upon binding to BSA. Protein in the presence of the reagent-metal complex produces a significant absorbance shift at a wavelength of 660 nm.
Protein quantification with Pierce 660 nm Protein Assay
The Pierce 660 nm Assay is more linear than coomassie-based Bradford assays and compatible with higher concentrations of most detergents, reducing agents and other commonly used reagents. The accessory Ionic Detergent Compatibility Reagent (IDCR) provides for even broader detergent compatibility, making this one of the only protein assays that is suitable for samples containing Laemmli SDS sample buffer with bromophenol blue. Although the Pierce 660 nm Protein Assay produces a higher level of protein-to-protein variation (37%) than other assays, such as the BCA Protein Assay, the simpler single-reagent format and broader substance compatibility make the Pierce 660 nm Assay more convenient for many routine applications. The Pierce 660nm Protein Assay can be performed in either a test tube or microplate format.
Pierce 660 nm Protein assay standard curve and Bio-Rad Bradford standard curve
Figure 5. Performance comparison and typical color response
A: Performance comparison of the Bio-Rad Bradford Protein Assay versus the Thermo Scientific Pierce 660nm Protein Assay. Assays were performed according the standard test-tube procedure using 100µL of BSA. The Pierce 660nm Protein Assay has a greater linear range (25 to 2000µg) compared with the Bradford Assay (125 to 1000µg). Absorbances were measured at the appropriate wavelengths for each assay (660nm and 595nm, respectively). Typical color response curve using the test tube procedure.
B: Typical color response curved using the test tube procedure. The linear detection ranges are 25 to 2000µg/mL for bovine serum albumin (BSA) and 50 to 2000µg/mL for bovine gamma globulin (BGG). Due to the inherent protein to protein variability of all protein assays (37% for the 660nm Protein Assay), this demonstrates that appropriate standards should be used for the type of unknown samples being measured.
Bradford assays
Catalog # Name Size Price (USD) Qty
22660 Pierce™ 660nm Protein Assay Reagent Each
191.65
Online Exclusive
229.00
Save 37.35 (16%)
A55866 Pierce™ Bradford Plus Protein Assay Kit with Dilution-Free™ BSA Protein Standards, multichannel pipette compatible, 950 mL Each
283.65
Online Exclusive
332.00
Save 48.35 (15%)
23200 Pierce™ Bradford Protein Assay Kit Each
151.65
Online Exclusive
169.00
Save 17.35 (10%)
22662 Pierce™ 660nm Protein Assay Kit Each
269.65
Online Exclusive
310.00
Save 40.35 (13%)
23246 Pierce™ Detergent Compatible Bradford Assay Kit Each
218.65
Online Exclusive
246.00
Save 27.35 (11%)
22663 Ionic Detergent Compatibility Reagent for Pierce™ 660nm Protein Assay Reagent Each
124.65
Online Exclusive
142.00
Save 17.35 (12%)
23236 Pierce™ Bradford Plus Protein Assay Kit, 950 mL Each
240.65
Online Exclusive
282.00
Save 41.35 (15%)
23238 Pierce™ Bradford Plus Protein Assay Reagent Each
189.65
Online Exclusive
218.00
Save 28.35 (13%)
Assay standards
Catalog # Name Size Price (USD) Qty
23208 Pierce™ Bovine Serum Albumin Standard Pre-Diluted Set Each
184.65
Online Exclusive
205.00
Save 20.35 (10%)
23209 Pierce™ Bovine Serum Albumin Standard Ampules, 2 mg/mL Each
86.65
Online Exclusive
100.00
Save 13.35 (13%)
A55863 Pierce™ Dilution-Free™ BSA Protein Standards, Multichannel Pipette Compatible, 0.125-2 mg/mL, 8-channel tubestrip (2 x 1 mL) Each
172.00
23212 Pierce™ Bovine Gamma Globulin Standard Ampules, 2 mg/mL Each
101.65
Online Exclusive
112.00
Save 10.35 (9%)
23213 Pierce™ Bovine Gamma Globulin Standard Pre-Diluted Set Each
187.65
Online Exclusive
207.00
Save 19.35 (9%)
 packaging, reagents, and dilution-free tubes for Bradford protein assaypackaging and reagents for Bradford protein assaypackaging and reagents for Bradford protein assay
 Bradford Plus Protein Assay Kit with Dilution-Free BSA Protein StandardsDetergent Compatible Bradford AssayBradford Protein Assay
Features
  • Ready-to-use Bradford assay kit with Dilution-Free BSA Protein Standards for reduced standard curve preparation time
  • Increased linearity and reduced protein-to-protein variation compared to other commercial Bradford assay formulations
  • Ready-to-use modification of the Bradford assay with additional additives to make it compatible with 1% or higher of commonly used detergents, including Triton X-100 and NP-40 detergents
  • Ready-to-use formulation of the popular assay reagent originally described by Bradford in 1976
Assay range (sample volume)125–1,500 µg/mL (5 µL)
Or
1–25 µg/mL (150 µL)
100–1,500 µg/mL (10 µL)
or
2–25 µg/mL (150 µL)
100–1,500 µg/mL (20 µL)
or
1–25 µg/mL (150 µL)
Compatible reagentsBuffer salts, metal ions, reducing agents, chelatorsDetergents, buffer salts, metal ions, reducing agents, chelatorsBuffer salts, metal ions, reducing agents, chelators
IncompatibilitiesDetergents>5% detergent solutionsDetergents
Incubation time10 min10 min10 min
Product size950 mL/kit450 mL/kit950 mL/kit
Cat. No.A55866
Related Bradford Plus Kit:
23236
2324623200
Related product: Pierce Dilution-Free BSA Protein Standards, Multichannel Pipette Compatible, 2 mg/mL
multichannel pipet transferring protein standards from the 8 connecting tube format
Effortless, Fast, Accurate
Select Bradford assays now include Dilution-Free BSA Protein Standards which are a set of seven prediluted BSA standards, uniquely designed for multichannel pipette compatibility.
The Pierce Dilution-Free protein standards minimize tedious dilutions and pipetting errors, delivering accurate results while reducing assay setup time. They are calibrated by direct comparison to purified BSA from the National Institute of Standards and Technology (NIST) to help ensure accurate standard curve generation.
Bradford assay principles
The Bradford assay was first described by Dr. Marion Bradford in 1976 and uses Coomassie G-250 dye in a colorimetric reagent for the detection and quantitation of total protein. Pierce Bradford Plus Protein Assays are modifications of the reagent first reported by Dr. Bradford.
Chemistry of Coomassie-based Bradford protein assays
In an acidic environment, proteins bind to Coomassie dye. This results in a spectral shift from the reddish-brown form of the dye (absorbance maximum at 465 nm) to the blue form (absorbance maximum at 610 nm). The difference between the two dye forms is greatest at 595 nm, making it the optimal wavelength to measure the blue color from the Coomassie dye–protein complex. If desired, the blue color can be measured at any wavelength between 575 nm and 615 nm. At the two extremes (575 nm and 615 nm), there is an approximate 10% decrease in the measured amount of color (absorbance) compared to that obtained at 595 nm. Development of color in Coomassie dye–based protein assays has been associated with the presence of certain basic amino acids—primarily arginine, lysine, and histidine—in the protein. Van der Waals forces and hydrophobic interactions also influence dye–protein binding. The number of Coomassie dye molecules bound to each protein is approximately proportional to the number of positive charges found on the protein. Free amino acids, peptides, and low molecular weight proteins do not produce color with Coomassie dye reagents. In general, the mass of a peptide or protein should be at least 3,000 Da for quantification with this reagent. In some applications, this can be an advantage.
Reaction schematic for Bradford protein assay
Figure 1. Reaction schematic for the Coomassie dye–based Bradford protein assays.
The main disadvantage of Bradford protein assays is their incompatibility with surfactants at concentrations routinely used to solubilize membrane proteins. In general, the presence of a surfactant in the sample, even at low concentrations, causes precipitation of the reagent. This limitation can be overcome by using Detergent Compatible Bradford Assay. In addition, the Coomassie dye reagent is highly acidic, so proteins with poor acid-solubility cannot be assayed with this reagent. Finally, Coomassie reagents result in about twice as much protein-to-protein variation as copper chelation-based assay reagents.
Product highlights
Bradford Plus Protein Assay Kit with Dilution-Free BSA Protein Standards
The Bradford Plus Protein Assay Kit with Dilution-Free BSA Protein Standards reduces assay setup by eliminating tedious standard curve generation. Dilution-Free BSA Protein Standards are available as part of the Bradford Plus Protein Assay or available separately for use with any Pierce Bradford Assay.
Detergent Compatible Bradford Assay
Pierce Detergent Compatible Bradford Assay Kit is a quick and ready-to-use modification of the well-known Bradford Assay for total protein quantitation. This formulation is compatible with up to 1% of commonly used detergents. Comparing Pierce Detergent Compatible Bradford Assay to the Bio-Rad DC Protein Assay, better sensitivity is seen with the Pierce Detergent Compatible Bradford Assay using common detergents. The range of the standard curve for the Pierce Detergent Compatible Bradford assay is 4 times broader than the range for the Bio-Rad DC assay.
Comparison between Bio-Rad DC assay vs Pierce Detergent Compatible Bradford assay
Figure 3. Bradford protein assay vs. the Bio-Rad DC protein assay. Each assay was performed in a microplate using BSA standards spiked with detergent or water (control), and followed the manufacturers’ instructions.
Table 1. Comparing Pierce Detergent Compatible Bradford Assay Kit with Bio-Rad DC Protein Assay Kit.
FeaturesPierce Detergent Compatible Bradford Assay KitBio-Rad DC Protein Assay Kit
Assay measurement (absorbance maximum)595 nm750 nm
Test tube assay sample volume50 μL100 μL
Microplate assay sample volume10 μL5 μL
Assay working range100-1,500 μg/mL200-1,500 μg/mL
Absorbance range (sensitivity)HighLow
Number of reagents in kit1 reagent3 reagents
Setup time10 min30 min
Incubation time10 min15 min
Total20 min45 min
Pierce 660 nm protein assay, catalog number 22662
Pierce 660 nm Protein Assay Kit
Assay range: microplate (sample volume)25 to 2,000 µg/mL (65 µL)
or
50 to 2,000 µg/mL (10 µL)
Incubation time and temperature5
Total assay time75
Absorbance660 nm
Compatible reagentsReducing agents, chelating agents, detergents
Incompatible reagentsHigh levels of detergents or SDS requires addition of Ionic detergent compatibility reagent
UniformityLess protein–protein variation than the Coomassie (Bradford) assay
Product size450 mL/kit
Cat. No.22662
Related product: Pierce Dilution-Free BSA Protein Standards, Multichannel Pipette Compatible, 2 mg/mL
Pierce 660 nm Assay principles
Chemistry of Pierce 660 nm Assay
The Pierce 660nm Protein Assay is based on the binding of a unique dye-metal complex to protein in acidic conditions that causes a shift in the dye's absorption maximum, which is measured at 660 nm. The dye-metal complex is reddish-brown and changes to green upon protein binding. The color change is produced by deprotonation of the dye at low pH facilitated by interactions with positively charged amino acid groups in proteins. Therefore, the dye interacts mainly with basic residues in proteins, such as histidine, arginine and lysine and to a lesser extent tyrosine, tryptophan and phenylalanine.
The color produced in the assay is stable and increases in proportion to a broad range of increasing protein concentrations, even in the presence of detergents and reducing agents that would be incompatible with Bradford and BCA Protein Assays. The optional IDCR may be added to the assay reagent to increase compatibility with high amounts of ionic detergents, allowing samples containing Laemmli SDS sample buffer with bromophenol blue to be measured. The IDCR completely dissolves by thorough mixing and does not have any effect on the assay.
Absorption maximum of Pierce 660 nm Protein Assay reagent
Figure 4. Absorption maximum of the 660 nm Assay Reagent-metal complex shifts proportionally upon binding to BSA. Protein in the presence of the reagent-metal complex produces a significant absorbance shift at a wavelength of 660 nm.
Protein quantification with Pierce 660 nm Protein Assay
The Pierce 660 nm Assay is more linear than coomassie-based Bradford assays and compatible with higher concentrations of most detergents, reducing agents and other commonly used reagents. The accessory Ionic Detergent Compatibility Reagent (IDCR) provides for even broader detergent compatibility, making this one of the only protein assays that is suitable for samples containing Laemmli SDS sample buffer with bromophenol blue. Although the Pierce 660 nm Protein Assay produces a higher level of protein-to-protein variation (37%) than other assays, such as the BCA Protein Assay, the simpler single-reagent format and broader substance compatibility make the Pierce 660 nm Assay more convenient for many routine applications. The Pierce 660nm Protein Assay can be performed in either a test tube or microplate format.
Pierce 660 nm Protein assay standard curve and Bio-Rad Bradford standard curve
Figure 5. Performance comparison and typical color response
A: Performance comparison of the Bio-Rad Bradford Protein Assay versus the Thermo Scientific Pierce 660nm Protein Assay. Assays were performed according the standard test-tube procedure using 100µL of BSA. The Pierce 660nm Protein Assay has a greater linear range (25 to 2000µg) compared with the Bradford Assay (125 to 1000µg). Absorbances were measured at the appropriate wavelengths for each assay (660nm and 595nm, respectively). Typical color response curve using the test tube procedure.
B: Typical color response curved using the test tube procedure. The linear detection ranges are 25 to 2000µg/mL for bovine serum albumin (BSA) and 50 to 2000µg/mL for bovine gamma globulin (BGG). Due to the inherent protein to protein variability of all protein assays (37% for the 660nm Protein Assay), this demonstrates that appropriate standards should be used for the type of unknown samples being measured.
Resources
Pierce Protein methods – an encyclopedia of articles about protein analysis
Protein assay technical guide – tools and reagents for improved quantitation of total or specific proteins.
Protein assay compatibility table – summarizes compatible substances for several popular protein assays.
Protein Analysis Learning Center – resources for different protein analysis techniques
Protein Biology Application Notes – journal-style articles based on research applications and proof-of-concept experiments
Liberate yourself with Pierce Dilution-Free protein assays
For Research Use Only. Not for use in diagnostic procedures.