packaging and reagents for protein quantitation
These specialized protein assay reagents and kits allow you to estimate and measure specific post-translational modification (PTM) or functional (enzymatic) classes of proteins in biological samples, including glycoproteins and phosphoproteins.
Technical handbook
Glycoprotein Carbohydrate Estimation KitPhosphoprotein Phosphate Estimation Assay Kit
Pierce Glycoprotein Carbohydrate Estimation KitGlycoprotein Standards Set for Pierce Glycoprotein Carbohydrate Estimation KitPierce Phosphoprotein Phosphate Estimation Assay Kit
Pierce Glycoprotein Carbohydrate Estimation KitGlycoprotein Standards Set for Pierce Glycoprotein Carbohydrate Estimation KitPierce Phosphoprotein Phosphate Estimation Assay Kit
Measure protein glycosylation as the percent of purified protein mass compared to included glycoprotein standards; uses periodic acid Schiff-base reagents (PAS).Classify and identify proteins as phosphorylated (serine and threonine) and obtain semi-quantitative assessment of the phosphorylation levels.
Assay overview
  • Qualitative—easily identifies purified proteins as glycoproteins or samples as contaminated with sugars
  • Semiquantitative—estimates the percent carbohydrate content (w/w) of purified glycoprotein by comparison to the included set of glycoprotein standards
  • Simple procedure—completed in less than 75 minutes; All you need is this kit, a microplate, and a plate reader to determine carbohydrate content
  • Adaptable—standard curve format allows for the design of alternative tests for aldehydes and carbohydrate components
Assay overview
  • Specific—measures phosphoserine (p-Ser) and phosphothreonine (p-Thr) only; does not measure phosphotyrosine (p-Tyr)
  • Convenient—test tube and 96-well microplate protocols included and requires less than 90 minutes to perform
  • Semi-quantitative—calculate moles of phosphate (P) per mole of protein using the included phosvitin standard
  • Customizable—format is adaptable to development of specific assays for pure phosphoproteins when previously characterized standards are available
Applications
  • Determine if a purified polyclonal antibody or other protein is glycosylated before attempting to perform carbohydrate-based conjugation or immobilization chemistries with hydrazide reagents
  • Characterize and classify purified protein fractions from an affinity procedure
  • Assess sugar and carbohydrate contamination in non-glycosylated protein samples
Applications
  • Identify proteins as containing phosphoserine or phosphothreonine phosphorylations
  • Estimate the amount of pS and pT phosphorylation
  • Develop specific quantitative assays for well-characterized phosphoproteins of interest
232602325923270
Glycoprotein Carbohydrate Estimation Kit
Phosphoprotein Phosphate Estimation Assay Kit
Catalog # Name Size Price (USD) Qty
23270 Pierce™ Phosphoprotein Phosphate Estimation Assay Kit Each
674.00
Glycoprotein Carbohydrate Estimation KitPhosphoprotein Phosphate Estimation Assay Kit
Pierce Glycoprotein Carbohydrate Estimation KitGlycoprotein Standards Set for Pierce Glycoprotein Carbohydrate Estimation KitPierce Phosphoprotein Phosphate Estimation Assay Kit
Pierce Glycoprotein Carbohydrate Estimation KitGlycoprotein Standards Set for Pierce Glycoprotein Carbohydrate Estimation KitPierce Phosphoprotein Phosphate Estimation Assay Kit
Measure protein glycosylation as the percent of purified protein mass compared to included glycoprotein standards; uses periodic acid Schiff-base reagents (PAS).Classify and identify proteins as phosphorylated (serine and threonine) and obtain semi-quantitative assessment of the phosphorylation levels.
Assay overview
  • Qualitative—easily identifies purified proteins as glycoproteins or samples as contaminated with sugars
  • Semiquantitative—estimates the percent carbohydrate content (w/w) of purified glycoprotein by comparison to the included set of glycoprotein standards
  • Simple procedure—completed in less than 75 minutes; All you need is this kit, a microplate, and a plate reader to determine carbohydrate content
  • Adaptable—standard curve format allows for the design of alternative tests for aldehydes and carbohydrate components
Assay overview
  • Specific—measures phosphoserine (p-Ser) and phosphothreonine (p-Thr) only; does not measure phosphotyrosine (p-Tyr)
  • Convenient—test tube and 96-well microplate protocols included and requires less than 90 minutes to perform
  • Semi-quantitative—calculate moles of phosphate (P) per mole of protein using the included phosvitin standard
  • Customizable—format is adaptable to development of specific assays for pure phosphoproteins when previously characterized standards are available
Applications
  • Determine if a purified polyclonal antibody or other protein is glycosylated before attempting to perform carbohydrate-based conjugation or immobilization chemistries with hydrazide reagents
  • Characterize and classify purified protein fractions from an affinity procedure
  • Assess sugar and carbohydrate contamination in non-glycosylated protein samples
Applications
  • Identify proteins as containing phosphoserine or phosphothreonine phosphorylations
  • Estimate the amount of pS and pT phosphorylation
  • Develop specific quantitative assays for well-characterized phosphoproteins of interest
232602325923270
Glycoprotein Carbohydrate Estimation Kit
Phosphoprotein Phosphate Estimation Assay Kit
Catalog # Name Size Price (USD) Qty
23270 Pierce™ Phosphoprotein Phosphate Estimation Assay Kit Each
674.00
Resources
Pierce Protein methods – an encyclopedia of articles about protein analysis
Protein assay technical guide – tools and reagents for improved quantitation of total or specific proteins.
Protein assay compatibility table – summarizes compatible substances for several popular protein assays.
Protein Analysis Learning Center – resources for different protein analysis techniques
Protein Biology Application Notes – journal-style articles based on research applications and proof-of-concept experiments
For Research Use Only. Not for use in diagnostic procedures.