Molecular characterization of any gene usually includes a thorough analysis of the temporal and spatial distribution of RNA expression. A number of widely used procedures exist for detecting and determining the abundance of a particular mRNA in a total or poly(A) RNA sample. Here, we review four popular methods: Northern blot analysis, nuclease protection assays (NPA), in situ hybridization, and reverse transcription-polymerase chain reaction (RT-PCR).
In theory, each of these techniques can be used to detect specific RNAs and to precisely determine their expression level. However, the methods each have inherent advantages and/or limitations. (See chart.) In general, Northern analysis is the only method that provides information about transcript size, whereas NPAs are the easiest way to simultaneously examine multiple messages (Figure 1). In situ hybridization is used to localize expression of a particular gene within a tissue or cell type, and RT-PCR is the most sensitive method for detecting and quantitating gene expression.
Figure 1. Simultaneous analysis of multiple mRNAs using RPA III™.
Ten micrograms of various mouse tissue total RNAs were hybridized overnight with approximately 50,000 cpm each of seven distinct probe transcripts. Nuclease digestion, product separation on a denaturing 6% acrylamide gel, and a four-hour exposure to film at -80°C were used to assess hybridization levels.
Northern Analysis
Northern analysis remains the standard for detection and quantitation of mRNA levels despite the advent of more sensitive techniques. Northern analysis presents several advantages over the other techniques. The most compelling of these is that it is the easiest method for determining transcript size, and for identifying alternatively spliced transcripts and multigene family members. It can also be used to directly compare the relative abundance of a given message between all the samples on a blot.
The Northern blotting procedure is straightforward and provides opportunities to evaluate progress at various points (e.g., intactness of the RNA sample and how efficiently it has transferred to the membrane). RNA samples are first separated by size via electrophoresis in an agarose gel under denaturing conditions. The RNA is then transferred to a membrane, crosslinked and hybridized with a labeled probe. Nonisotopic or high specific activity radiolabeled probes can be used including random-primed, nick-translated, or PCR-generated DNA probes, in vitro transcribed RNA probes, and oligonucleotides. Additionally, sequences with only partial homology (e.g., cDNA from a different species or genomic DNA fragments that might contain an exon) may be used as probes.
Despite these advantages, there are limitations associated with Northern analysis. First, if RNA samples are even slightly degraded, the quality of the data and the ability to quantitate expression are severely compromised. Second, a standard Northern procedure is, in general, the least sensitive of the reviewed techniques. However, substantial improvements can be made to increase detection of most mRNA species (see below). Another limitation of Northern blotting has been the difficulty associated with multiple probe analysis. To detect more than one message, it is usually necessary to strip the initial probe before hybridizing with a second probe. This process can be time consuming and problematic.
Making a Good Technique Even Better
Although established Northern blotting procedures are up and working in most molecular biology laboratories, Ambion has found ways to considerably improve on standard protocols, resulting in greatly increased Northern sensitivity. We have developed RNase-free reagents optimized for each step of the procedure to provide complete, high-sensitivity Northern blotting systems. The NorthernMax™ Kits provide everything needed to perform Northern analysis except for the membrane and probe. Each of the reagents is also available separately including the NorthernMax Rapid Transfer Buffer, that facilitates complete transfer in as little as an hour and a half. Ambion's new ULTRAhyb™ Ultrasensitive Hybridization Buffer increases sensitivity up to 100 fold (Figure 2) by pushing hybridization to completion. As few as 10,000 molecules can be detected.
Figure 2. ULTRAhyb™ versus a Standard Hybridization Buffer Using DNA and RNA Probes. Identical Northern blots of indicated amounts of mouse thymus total RNA were hybridized overnight with radiolabeled StripAble™ DNA or RNA probe. The blots were incubated with probe in either ULTRAhyb or standard hybridization buffer as indicated. Standard low and high stringency SSC/SDS washes were used to remove non-specifically bound probe. Blots were exposed to film for 2.5 hours with a single intensifying screen.
Nuclease Protection Assays (NPAs)
The NPA (including both ribonuclease protection assays and S1 nuclease assays) is an extremely sensitive method for the detection and quantitation of specific mRNAs. The basis of the NPA is solution hybridization of an antisense probe (radiolabeled or nonisotopic) to an RNA sample. After hybridization, single-stranded, unhybridized probe and RNA are degraded by nucleases. The remaining protected fragments are separated on an acrylamide gel. Solution hybridization is typically more efficient than membrane-based hybridization, and it can accommodate up to 100 µg of sample RNA, compared with the 20-30 µg maximum of blot hybridizations. NPAs are also less sensitive to RNA sample degradation than Northern analysis since cleavage is only detected in the region of overlap with the probe (probes are usually about 100-400 bases in length).
NPAs are the method of choice for the simultaneous detection of several RNA species. Figure 1 shows how 6 different probes and an internal control can be used together for detection of mRNA transcripts each within 4 different RNA samples. During solution hybridization and subsequent analysis, individual probe/target interactions are completely independent of one another. Thus, several RNA targets and appropriate controls can be assayed simultaneously (up to twelve have been used in the same reaction), provided that the individual probes are of different lengths. NPAs are also commonly used to precisely map mRNA termini and intron/exon junctions.
The primary limitation of NPAs is the lack of information on transcript size. The portion of probe homologous to target RNA determines the size of the protected fragment. Another drawback to NPAs is the lack of probe flexibility. The most common type of NPA, the ribonuclease protection assay, requires the use of RNA probes. Oligonucleotides and other single-stranded DNA probes can only be used in assays containing S1 nuclease. The single-stranded, antisense probe must typically be completely homologous to target RNA to prevent cleavage of the probe:target hybrid by nuclease. This means that partially related sequences (i.e., cross species) usually cannot be used.
RNA Analysis in a Single Tube
Ambion released the first commercially available Ribonuclease Protection Assay Kit in 1990 and has since continued to refine and improve the procedure resulting in a number of kits with different advantages. The RPA III™ Kit is recommended for all new users and consists of a single-tube protocol. The HybSpeed™ RPA Kit utilizes a 10-minute hybridization step so that the entire assay can be completed in a single day.
In Situ Hybridization
In situ hybridization (ISH) is a powerful and versatile tool for the localization of specific mRNAs in cells or tissues. Unlike Northern analysis and nuclease protection assays, ISH does not require the isolation or electrophoretic separation of RNA. Hybridization of the probe takes place within the cell or tissue. Since cellular structure is maintained throughout the procedure, ISH provides information about the location of mRNA within the tissue sample.
The procedure begins by fixing samples in neutral-buffered formalin, and embedding the tissue in paraffin. The samples are then sliced into thin sections and mounted onto microscope slides. (Alternatively, tissue can be sectioned frozen and post-fixed in paraformaldehyde.) After a series of washes to dewax and rehydrate the sections, a Proteinase K digestion is performed to increase probe accessibility, and a labeled probe is then hybridized to the sample sections. Radiolabeled probes are visualized with liquid film dried onto the slides, while nonisotopically labeled probes are conveniently detected with colorimetric or fluorescent reagents.
The major drawback to ISH is the procedure itself. Standard protocols are time-consuming, laborious and may require specialized equipment for preparing samples and visualizing results of the experiment. Additionally, quantitation of gene expression is not as straightforward as with the other techniques.
RT-PCR
RT-PCR has revolutionized the study of gene expression. It is now theoretically possible to detect the RNA transcript of any gene, regardless of the scarcity of the starting material or relative abundance of the specific mRNA. In RT-PCR, an RNA template is copied into a complementary DNA (cDNA) using a retroviral reverse transcriptase. The cDNA is then amplified exponentially by PCR. As with NPAs, RT-PCR is somewhat tolerant of degraded RNA. As long as the RNA is intact within the region spanned by the primers, the target will be amplified.
Although RT-PCR is the most sensitive method of mRNA detection available, it does have drawbacks. It can be the most technically challenging method of detection and quantitation, often requiring substantial pre-experimental planning and design. Additionally, because of its extreme sensitivity, even minute amounts of contamination by genomic DNA or previously amplified PCR products can lead to aberrant results, so steps must be taken to avoid this pitfall.
Relative quantitative RT-PCR involves amplifying an internal control simultaneously with the gene of interest. The internal control is used to normalize the samples. Once normalized, direct comparisons of relative abundance of a specific mRNA can be made across the samples. It is crucial to choose an internal control with a constant level of expression across all experimental samples (i.e., not affected by experimental treatment). Commonly used internal controls (e.g., GAPDH, -actin, cyclophilin) often vary in expression and, therefore, may not be appropriate internal controls. Additionally, most common internal controls are expressed at much higher levels than the mRNA being studied. For relative RT-PCR results to be meaningful, all products of the PCR reaction must be analyzed in the linear range of amplification. This becomes difficult for transcripts of widely different levels of abundance.
Competitive RT-PCR is used for absolute quantitation. This technique involves designing, synthesizing, and accurately quantitating a competitor RNA that can be distinguished from the endogenous target by a small difference in size or sequence. Known amounts of the competitor RNA are added to experimental samples and RT-PCR is performed. Signals from the endogenous target are compared with signals from the competitor to determine the amount of target present in the sample.
Making RT-PCR Work for You
Ambion has a complete line of products to simplify RT-PCR. The RETROscript™ Kit provides a convenient and flexible system for synthesizing first-strand cDNA. The QuantumRNA™ 18S Internal Standards Kits utilize Ambion's Competimer™ technology (patent pending) to allow the use of the best internal control, 18S rRNA, for relative RT-PCR (Figure 3).
Figure 3. Ambion's QuantumRNA™ Technology in Multiplex Quantitative RT-PCR using 18S rRNA as an Internal Control. RT-PCR reactions on brain, embryo, liver, and spleen total RNA using A) primers for clathrin, B) primers for clathrin and 18S, or C) primers for clathrin, 18S rRNA primers and 18S rRNA Competimers. Note that without Competimers, 18S cannot be used as an internal control because of its high abundance (B). Addition of Competimers (C) makes multiplex PCR possible, providing sample-to-sample relative quantitation.
Summary
As detailed above, there are several methods for detecting and quantitating mRNA, each with advantages and disadvantages. Northern analysis provides information about the endogenous transcript size and is tolerant of many types of probe. The NPA is a simple format that is ideal for multi-target analysis and mapping studies. Gene expression can be localized within tissue or cells using in situ hybridization, and even the rarest messages can be detected with RT-PCR. Deciding which technique to use will depend on exactly what information is needed. Because no single method provides all of the information, a thorough analysis of gene expression often requires the use of multiple techniques.
Ordering Information
Catalog # Name Size Price (USD) Qty
AM10100 BrightStar™-Plus Positively Charged Nylon Membrane, 15 cm x 15 cm Each
181.65
Online Exclusive
187.00
Save 5.35 (3%)
AM10102 BrightStar™-Plus Positively Charged Nylon Membrane, 30 cm x 45 cm Each
189.00
AM10104 BrightStar™-Plus Positively Charged Nylon Membrane, 30 cm x 3 m Each
695.65
Online Exclusive
718.00
Save 22.35 (3%)
AM1716 QuantumRNA™ Classic 18S Internal Standard Each
300.99
Special offer
Online exclusive
Ends: 01-May-2025
320.00
Save 19.01 (6%)
AM1717 QuantumRNA™ Classic II 18S Internal Standard Each
287.98
Special offer
Online exclusive
Ends: 01-May-2025
306.00
Save 18.02 (6%)
AM1718 QuantumRNA™ Universal 18S Internal Standard Each
248.80
Special offer
Online exclusive
Ends: 01-May-2025
311.00
Save 62.20 (20%)
AM1940 NorthernMax™ Kit Each
739.65
Online Exclusive
762.00
Save 22.35 (3%)
AM1946 NorthernMax™-Gly Kit Each
852.00
AM5722G Random Decamers (50 μM) Each
147.68
Special offer
Online exclusive
Ends: 01-May-2025
165.00
Save 17.32 (10%)
AM5730G Oligo (dT) Primer (50 μM) Each
153.65
Special offer
Online exclusive
Ends: 01-May-2025
165.00
Save 11.35 (7%)
AM7118 Yeast RNA (10 mg/mL) Each
365.04
Special offer
Online exclusive
Ends: 01-May-2025
391.00
Save 25.96 (7%)
AM8551 NorthernMax™-Gly Sample Loading Dye Each
139.65
Online Exclusive
146.00
Save 6.35 (4%)
AM8552 NorthernMax™ Formaldehyde Load Dye Each
97.00
AM8669 ULTRAhyb™ Ultrasensitive Hybridization Buffer, 4 x 125 mL Each
672.65
Online Exclusive
700.00
Save 27.35 (4%)
AM8670 ULTRAhyb™ Ultrasensitive Hybridization Buffer, 125 mL Each
218.65
Online Exclusive
227.00
Save 8.35 (4%)
AM8671 NorthernMax™ 10X Running Buffer Each
313.65
Online Exclusive
329.00
Save 15.35 (5%)
AM8672 NorthernMax™ One-Hour Transfer Buffer Each
284.00
AM8673 NorthernMax™ Low Stringency Wash Buffer Each
246.00
AM8674 NorthernMax™ High Stringency Wash Buffer Each
241.00
AM8676 NorthernMax™ Denaturing Gel Buffer (10X) Each
290.00
AM8677 NorthernMax™ Prehybridization/Hybridization Buffer Each
376.00
AM8678 NorthernMax™-Gly Gel Prep/Running Buffer (10X) Each
479.65
Online Exclusive
500.00
Save 20.35 (4%)
AM9680 Salmon Sperm DNA, sheared (10 mg/mL) Each
329.64
Special offer
Online exclusive
Ends: 01-May-2025
386.00
Save 56.36 (15%)
AM9820 SDS, 20% Solution, RNase-free Each
67.65
Special offer
Online exclusive
Ends: 01-May-2025
75.00
Save 7.35 (10%)