Colorimetric ReadiLAMP kit for SARS-CoV-2 detection
Invitrogen Colorimetric ReadiLAMP™ kit, SARS-CoV-2
This kit uses a Loop-Mediated Isothermal Amplification (LAMP)- based solution for rapid SARS-CoV-2 detection and surveillance.
  • Detection is simple, intuitive and fast: colorimetric LAMP reaction changes color from purple to blue if SARS-target is present. No instrumentation is required for this assay, except for a 65℃ heat source.
  • Utilizes non-invasive saliva samples or swab samples
  • Ability to detect varying SARS-CoV-2 viral strains
  • It can be utilized either of two ways – direct assay using crude samples or assay based on RNA purified from saliva and swabs.
  • It allows input of up to 50% purified RNA per LAMP reaction to achieve maximal sensitivity in SARS-CoV-2 detection. See kit performance data.
  • Process 1-96 samples at a time: Customers can process more samples with the same number of technicians, using individual tubes, tubes strips, or 96 well plates.
Learn more from our application notesOrder Now
Video Player is loading.
Current Time 0:00
Duration 1:44
Loaded: 0.00%
Stream Type LIVE
Remaining Time 1:44
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Product details for the Invitrogen Colorimetric ReadiLAMP™ Kit, SARS-CoV-2
     Colorimetric ReadiLAMP kit, 100 reactionsColorimetric ReadiLAMP™ kit,
    100 reactions
    Colorimetric ReadiLAMP kit, 1000 reactions
    Colorimetric ReadiLAMP™ kit,
    1000 reactions
    Reactions100 reactions and 1000 reactions
    Test duration~30 mins – Direct LAMP assay
    ~1 hour – LAMP using purified RNA
    Sample typesSaliva and Swabs
    DetectionColorimetric readout
    SensitivityDirect: ≤250 copies/25 uL LAMP reaction
    Purified: ≤100 copies/25 uL LAMP reaction
    ControlsHuman Control Primer Mix and SARS-CoV-2 Control RNA
    Test format1-96 samples at a time
    Kit Contents
    100 Reactions
    Enzyme Mix (10X) - 250µL
    LAMP buffer (5X)- 500µL
    SARS-CoV-2 Primer Mix (5X)-500µL
    Human Control Primer Mix (5X) 250µL
    SARS-CoV-2 Control RNA-20µL
    1000 reactions
    Enzyme Mix (10X) – 2.5mL
    LAMP buffer (5X)- 5mL
    SARS-CoV-2 Primer Mix (5X)-5mL
    Human Control Primer Mix (5X) 2.5mL
    SARS-CoV-2 Control RNA-200µL
    Kit storage temperature–20℃
    Instrument required65℃ heating block with heated lid
    Regulatory classificationRUO
    Catalogue Number100 reactions (Cat. No. A52539) or 1,000 reactions (Cat. No. A52544)
    Why use the Invitrogen Colorimetric ReadiLAMP™ Kit, SARS-CoV-2?
    • Simplicity: Fast and simple protocol, easy to read results and robust system based on colorimetric indicators.
    • Range of Samples types:
      • Sample ranges from saliva, nasal swabs, nasopharyngeal swab, and other swabs.
    • Sensitivity and variant coverage:The kit allows input of up to 50% purified RNA per LAMP reaction enabling maximal sensitivity in SARS-CoV-2 detection.
      • Direct: ≤ 250 copies/25 µL LAMP reaction
      • Purified: ≤ 100 copies/25 µL LAMP reaction
      • Various SARS-CoV-2 variant coverage
    • Speed to result and protocol flexibility – choose from 2 workflows:
      • Direct LAMP protocol provides results in about 30 minutes using crude saliva or transport media samples.
      • Purified RNA-based LAMP assay provides results in about 60 minutes: 30 minutes purification, followed by 30 minutes LAMP assay. 
    • Throughput
      • Allows users to optimize their workflow for variable sample numbers (1-96 sample batches) and consumable types (individual tubes, strip tubes, 96-well plates) for efficient sample processing and rapid turn-around times. 
    Blue and violet colorimetric results from the Invitrogen Colorimetric ReadiLAMP™ kit, SARS-CoV-2 
    Protocols and workflows for using the Invitrogen Colorimetric ReadiLAMP™ Kit, SARS-CoV-2
    Use this kit to either perform direct assay or purified assay. Either method yields quality results that are easy to interpret with blue being positive Sars-CoV-2 reactions and violet being negative results.
    Illustration of pre-LAMP reaction, time and temperature for assay, and colored results 
    Direct Method: SARS-CoV-2 in crude saliva/NP samples
    Illustration of direct method assay protocol
    How to use Colorimetric ReadiLAMP™ kit for detecting SARS-CoV-2 from crude sample types
    Video Player is loading.
    Current Time 0:00
    Duration 5:41
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 5:41
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Indirect Method: SARS-CoV-2 detection workflow for purified RNA samples
      Illustration of RNA purification assay protocol
      How to use Colorimetric ReadiLAMP™ kit for detecting SARS-CoV-2 RNA
      Video Player is loading.
      Current Time 0:00
      Duration 4:39
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 4:39
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Performance data for Invitrogen Colorimetric ReadiLAMP™ Kit, Sars-CoV-2
        See the results of using our kit for various sample types and target infectious disease analytes. With color coded positive and negative results, interpreting the data is simple.
        Positive blue color change observed in the presence of SARS-CoV-2 when tested using nasopharyngeal samples
        Figure 1. The Colorimetric ReadiLAMP Kit enables highly specific SARS-CoV-2 detection.
        Samples from 12 unique nasopharyngeal donors (D1 – D12) were supplemented with γ-irradiated SARS-CoV-2 isolate USA-WA1/2020 (BEI Resources), Influenza A H1N1, or Influenza A H3N2 (ATCC) or 5.0 μL NATtrol™ Respiratory Pathogen Panel-1 or Panel-2 (RP1/RP2, Zeptometrix). RNA was purified using the MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit and the Thermo Scientific KingFisher Flex Magnetic Particle Processor. The 25 μL Colorimetric ReadiLAMP reactions were incubated at 65 °C for 30 min. In all samples supplemented with SARS-CoV-2, a positive blue color change was observed, while all other virally supplemented samples and controls remained a negative purple color. [RP1 contains the following viral particles and bacteria: Influenza A H3N2 (Brisbane/10/07), Influenza A H1N1 (NY/02/2009), Rhinovirus (Type 1A), Adenovirus (Type 3), Parainfluenza (Type 1), Parainfluenza (Type 4), Metapneumovirus (Peru 6-2003), Chlamydia pneumoniae (CWL-029), Mycoplasma pneumoniae (M129), and Coxsackievirus (Type A1). Whereas RP2 contains the following viral particles and bacteria: from Influenza A H1 (New Caledonia/20/99), Influenza B (Florida/02/06), RSV (Type A), Parainfluenza (Type 2), Parainfluenza (Type 3), Coronavirus (HKU-1 recombinant), Coronavirus (OC43), Coronavirus (NL63), Coronavirus (229E), Bordetella pertussis (A639).]
        Positive blue color change observed in the presence of 15 various strains of SARS-CoV-2 at a sensitivity of 25 copies/reaction.
        Figure 2. The Colorimetric ReadiLAMP Kit efficiently detects multiple SARS-CoV-2 strains.
        Fifteen strains of SARS-CoV-2 RNA were obtained and evaluated in the Colorimetric ReadiLAMP assay. Each 25 μL reaction included 2.5 x 101 – 1.0 x 104 copies of SAR-CoV-2 in a background of 50 ng human genomic DNA. After 65 °C incubation for 30 min, 80% of strains were detected at a sensitivity of 25 copies/ reaction with all strains being detected at 50 copies/ reaction. These detectable strains, obtained from ATCC, were Hong Kong (NR-52388), Italy (NR-52498), England (NR-52499), Singapore (NR-52501), Germany (NR-52502),  USA-IL (NR-52503), USA-CA1 (NR-52504), USA-AZ (NR-52505), USA-WI (NR-52506), USA-CA3 (NR-52507), USA-CA4 (NR-52508), USA-CA2 (NR-52509), Chile-Santiago (NR-52510), USA-New York (NR-53518), and USA-WA (NR-52285).
        Positive blue color change observed in the presence of as few as 20 copies of SARS-CoV-2 per reaction when tested using 12 unique saliva samples.
        Figure 3. Using saliva-derived samples, the Colorimetric ReadiLAMP kit detections as few as 20 copies SARS-CoV-2 per reaction.
        Samples from 12 unique saliva donors (D1 - D12) were supplemented with γ-irradiated SARS-CoV-2 isolate USA-WA1/2020 (BEI Resources). RNA was purified using the MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit and the Thermo Scientific KingFisher Flex Magnetic Particle Processor. The 25 μL Colorimetric ReadiLAMP reactions were assembled using 1.0 – 12.5 μL input sample containing 0 – 250 copies of SARS-CoV-2 per reaction. After a 65 °C incubation for 30 min, all samples containing SARS-CoV-2 RNA transitioned to blue, positive reactions, while non-templated reactions remained purple. To increase sensitivity, sample input volume may be increased to 50 % of the total reaction volume (12.5/ 25 μL) with the caveat that individual reaction sensitivity is dependent on sample type, donor, and RNA purity. For saliva, the minimum range that can be robustly detected varies from 20 – 100 copies/ reaction for uncomplicated to challenging samples, respectively.
        Product details for the Invitrogen Colorimetric ReadiLAMP™ Kit, SARS-CoV-2
         Colorimetric ReadiLAMP kit, 100 reactionsColorimetric ReadiLAMP™ kit,
        100 reactions
        Colorimetric ReadiLAMP kit, 1000 reactions
        Colorimetric ReadiLAMP™ kit,
        1000 reactions
        Reactions100 reactions and 1000 reactions
        Test duration~30 mins – Direct LAMP assay
        ~1 hour – LAMP using purified RNA
        Sample typesSaliva and Swabs
        DetectionColorimetric readout
        SensitivityDirect: ≤250 copies/25 uL LAMP reaction
        Purified: ≤100 copies/25 uL LAMP reaction
        ControlsHuman Control Primer Mix and SARS-CoV-2 Control RNA
        Test format1-96 samples at a time
        Kit Contents
        100 Reactions
        Enzyme Mix (10X) - 250µL
        LAMP buffer (5X)- 500µL
        SARS-CoV-2 Primer Mix (5X)-500µL
        Human Control Primer Mix (5X) 250µL
        SARS-CoV-2 Control RNA-20µL
        1000 reactions
        Enzyme Mix (10X) – 2.5mL
        LAMP buffer (5X)- 5mL
        SARS-CoV-2 Primer Mix (5X)-5mL
        Human Control Primer Mix (5X) 2.5mL
        SARS-CoV-2 Control RNA-200µL
        Kit storage temperature–20℃
        Instrument required65℃ heating block with heated lid
        Regulatory classificationRUO
        Catalogue Number100 reactions (Cat. No. A52539) or 1,000 reactions (Cat. No. A52544)
        Why use the Invitrogen Colorimetric ReadiLAMP™ Kit, SARS-CoV-2?
        • Simplicity: Fast and simple protocol, easy to read results and robust system based on colorimetric indicators.
        • Range of Samples types:
          • Sample ranges from saliva, nasal swabs, nasopharyngeal swab, and other swabs.
        • Sensitivity and variant coverage:The kit allows input of up to 50% purified RNA per LAMP reaction enabling maximal sensitivity in SARS-CoV-2 detection.
          • Direct: ≤ 250 copies/25 µL LAMP reaction
          • Purified: ≤ 100 copies/25 µL LAMP reaction
          • Various SARS-CoV-2 variant coverage
        • Speed to result and protocol flexibility – choose from 2 workflows:
          • Direct LAMP protocol provides results in about 30 minutes using crude saliva or transport media samples.
          • Purified RNA-based LAMP assay provides results in about 60 minutes: 30 minutes purification, followed by 30 minutes LAMP assay. 
        • Throughput
          • Allows users to optimize their workflow for variable sample numbers (1-96 sample batches) and consumable types (individual tubes, strip tubes, 96-well plates) for efficient sample processing and rapid turn-around times. 
        Blue and violet colorimetric results from the Invitrogen Colorimetric ReadiLAMP™ kit, SARS-CoV-2 
        Protocols and workflows for using the Invitrogen Colorimetric ReadiLAMP™ Kit, SARS-CoV-2
        Use this kit to either perform direct assay or purified assay. Either method yields quality results that are easy to interpret with blue being positive Sars-CoV-2 reactions and violet being negative results.
        Illustration of pre-LAMP reaction, time and temperature for assay, and colored results 
        Direct Method: SARS-CoV-2 in crude saliva/NP samples
        Illustration of direct method assay protocol
        How to use Colorimetric ReadiLAMP™ kit for detecting SARS-CoV-2 from crude sample types
        Video Player is loading.
        Current Time 0:00
        Duration 5:41
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 5:41
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Indirect Method: SARS-CoV-2 detection workflow for purified RNA samples
          Illustration of RNA purification assay protocol
          How to use Colorimetric ReadiLAMP™ kit for detecting SARS-CoV-2 RNA
          Video Player is loading.
          Current Time 0:00
          Duration 4:39
          Loaded: 0%
          Stream Type LIVE
          Remaining Time 4:39
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            Performance data for Invitrogen Colorimetric ReadiLAMP™ Kit, Sars-CoV-2
            See the results of using our kit for various sample types and target infectious disease analytes. With color coded positive and negative results, interpreting the data is simple.
            Positive blue color change observed in the presence of SARS-CoV-2 when tested using nasopharyngeal samples
            Figure 1. The Colorimetric ReadiLAMP Kit enables highly specific SARS-CoV-2 detection.
            Samples from 12 unique nasopharyngeal donors (D1 – D12) were supplemented with γ-irradiated SARS-CoV-2 isolate USA-WA1/2020 (BEI Resources), Influenza A H1N1, or Influenza A H3N2 (ATCC) or 5.0 μL NATtrol™ Respiratory Pathogen Panel-1 or Panel-2 (RP1/RP2, Zeptometrix). RNA was purified using the MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit and the Thermo Scientific KingFisher Flex Magnetic Particle Processor. The 25 μL Colorimetric ReadiLAMP reactions were incubated at 65 °C for 30 min. In all samples supplemented with SARS-CoV-2, a positive blue color change was observed, while all other virally supplemented samples and controls remained a negative purple color. [RP1 contains the following viral particles and bacteria: Influenza A H3N2 (Brisbane/10/07), Influenza A H1N1 (NY/02/2009), Rhinovirus (Type 1A), Adenovirus (Type 3), Parainfluenza (Type 1), Parainfluenza (Type 4), Metapneumovirus (Peru 6-2003), Chlamydia pneumoniae (CWL-029), Mycoplasma pneumoniae (M129), and Coxsackievirus (Type A1). Whereas RP2 contains the following viral particles and bacteria: from Influenza A H1 (New Caledonia/20/99), Influenza B (Florida/02/06), RSV (Type A), Parainfluenza (Type 2), Parainfluenza (Type 3), Coronavirus (HKU-1 recombinant), Coronavirus (OC43), Coronavirus (NL63), Coronavirus (229E), Bordetella pertussis (A639).]
            Positive blue color change observed in the presence of 15 various strains of SARS-CoV-2 at a sensitivity of 25 copies/reaction.
            Figure 2. The Colorimetric ReadiLAMP Kit efficiently detects multiple SARS-CoV-2 strains.
            Fifteen strains of SARS-CoV-2 RNA were obtained and evaluated in the Colorimetric ReadiLAMP assay. Each 25 μL reaction included 2.5 x 101 – 1.0 x 104 copies of SAR-CoV-2 in a background of 50 ng human genomic DNA. After 65 °C incubation for 30 min, 80% of strains were detected at a sensitivity of 25 copies/ reaction with all strains being detected at 50 copies/ reaction. These detectable strains, obtained from ATCC, were Hong Kong (NR-52388), Italy (NR-52498), England (NR-52499), Singapore (NR-52501), Germany (NR-52502),  USA-IL (NR-52503), USA-CA1 (NR-52504), USA-AZ (NR-52505), USA-WI (NR-52506), USA-CA3 (NR-52507), USA-CA4 (NR-52508), USA-CA2 (NR-52509), Chile-Santiago (NR-52510), USA-New York (NR-53518), and USA-WA (NR-52285).
            Positive blue color change observed in the presence of as few as 20 copies of SARS-CoV-2 per reaction when tested using 12 unique saliva samples.
            Figure 3. Using saliva-derived samples, the Colorimetric ReadiLAMP kit detections as few as 20 copies SARS-CoV-2 per reaction.
            Samples from 12 unique saliva donors (D1 - D12) were supplemented with γ-irradiated SARS-CoV-2 isolate USA-WA1/2020 (BEI Resources). RNA was purified using the MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit and the Thermo Scientific KingFisher Flex Magnetic Particle Processor. The 25 μL Colorimetric ReadiLAMP reactions were assembled using 1.0 – 12.5 μL input sample containing 0 – 250 copies of SARS-CoV-2 per reaction. After a 65 °C incubation for 30 min, all samples containing SARS-CoV-2 RNA transitioned to blue, positive reactions, while non-templated reactions remained purple. To increase sensitivity, sample input volume may be increased to 50 % of the total reaction volume (12.5/ 25 μL) with the caveat that individual reaction sensitivity is dependent on sample type, donor, and RNA purity. For saliva, the minimum range that can be robustly detected varies from 20 – 100 copies/ reaction for uncomplicated to challenging samples, respectively.