Search Thermo Fisher Scientific
Organism Type | Human | Prepared from low passage Master Cell Bank derived from parental 293 F cells (fast growing variant) that were recloned by limiting dilution. Maintained in serum-free conditions for only 30-35 total passages. Demonstrates high transfection efficiencies with 293fectin. Permits transfection of large volumes of cells. |
Tissues | Kidney | |
Phenotype | Suspension | |
Primary | no | |
Application | Cell Culture / Growth Conditions,Protein Expression,Transient Transfection |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||