Fluoromount-G™ Mounting Medium
Fluoromount-G™ Mounting Medium
Invitrogen™

Fluoromount-G™ Mounting Medium

Fluoromount-G Mounting Medium is a clear liquid medium designed for use in mounting slides following immunofluorescent staining. For convenience, thisRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
00-4958-02Promo Image25 mL
Catalog number 00-4958-02
Price (USD)
91.65
Online Exclusive
98.00
Save 6.35 (6%)
Each
In stock
Add to cart
Quantity:
25 mL
Recurring order eligible. Learn more »
Price (USD)
91.65
Online Exclusive
98.00
Save 6.35 (6%)
Each
Add to cart
Ask our AI about this Product
Fluoromount-G Mounting Medium is a clear liquid medium designed for use in mounting slides following immunofluorescent staining. For convenience, this medium is provided with and without DAPI, a blue nuclear counter stain. It is water soluble and used to mount slides in which the final step of staining is aqueous. It forms a semi-permanent seal for prolonged storage of slides at 2-8°C. Fluoromount-G Mounting Medium does not fluoresce and may reduce the amount of fluorochrome quenching and photobleaching during fluorescence microscopy. This medium is supplied as a 1X stock solution containing sodium azide. The refractive index of Fluoromount-G Mounting Medium is reported to be ∼1.4, and it is best used when the thickness of the specimen is less than 10 μm.

Select the antifade mounting or optical clearing reagent that matches your fluorescence imaging needs ›

For mountants with a 1.52 refractive index, optimized for oil immersion objective imaging, try curing mountant Prolong Glass Antifade Mountant or non-curing SlowFade Glass Soft-set Antifade Mountant.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity25 mL
Product LineFluoromount-G™
Product TypeMounting Medium
Reagent TypeMounting Solution
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
E143398Certificate of AnalysisJun 13, 202500-4958-02
E143350Certificate of AnalysisApr 09, 202500-4958-02
E143336Certificate of AnalysisApr 07, 202500-4958-02
E143291Certificate of AnalysisFeb 07, 202500-4958-02
E143292Certificate of AnalysisFeb 07, 202500-4958-02
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection