eBioscience™ Lipopolysaccharide (LPS) Solution (500X)
Product Image
Invitrogen™

eBioscience™ Lipopolysaccharide (LPS) Solution (500X)

The Lipoploysaccharide (LPS) Solution (500X) is a ready to use solution of LPS from Escherichia coli 026:B6 in aqueous solution.Read more
Promo ImagePromotion
Promo code:RFLOW25
Get more, spend less on every flow cytometry run
Fuel discovery without breaking your budget. Save up to 33% on flow cytometry essentials—compensation beads, buffers, assays & more. Use code RFLOW25 at checkout. The more you buy, the more you save. Limited time only!Learn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
00-4976-03Promo Image4 x 100 μL
00-4976-93Promo Image100 μL
Catalog number 00-4976-03
Price (USD)
195.00
Each
In stock
Add to cart
Quantity:
4 x 100 μL
Recurring order eligible. Learn more »
Price (USD)
195.00
Each
Add to cart
Ask our AI about this Product
The Lipoploysaccharide (LPS) Solution (500X) is a ready to use solution of LPS from Escherichia coli 026:B6 in aqueous solution. LPS is a major component of the cell wall of gram negative bacteria. It is highly immunogenic and strongly activates immune cells bearing the CD14/TLR4/MD2 receptor complex. This reagent is intended for use in in vitro culture of B cells, monocytes, macrophages and other cells that are responsive to LPS.

Reported Application
Flow Cytometric Analysis, Functional Assays

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeB cells, monocytes, macrophages and other cells that are responsive to LPS.
Concentration500 X
Product LineeBioscience™
Quantity4 x 100 μL
FormLiquid
Product TypeLPS Solution
SterilitySterile
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3125951Certificate of AnalysisJul 03, 202500-4976-93
3148741Certificate of AnalysisJul 02, 202500-4976-93
3047098Certificate of AnalysisJun 11, 202500-4976-93
3125952Certificate of AnalysisJun 02, 202500-4976-93
3108534Certificate of AnalysisApr 28, 202500-4976-93
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection