1x1 image pixel for data collection
eBioscience™ Cell Stimulation Cocktail (500X)
eBioscience™ Cell Stimulation Cocktail (500X)
Invitrogen™

eBioscience™ Cell Stimulation Cocktail (500X)

The Cell Stimulation Cocktail (500X) is a cocktail of phorbol 12-myristate 13-acetate (PMA) and ionomycin. Treatment with PMA and ionomycinRead more
Promo ImagePromotion
Promo code:RFLOW25
Get more, spend less on every flow cytometry run
Fuel discovery without breaking your budget. Save up to 33% on flow cytometry essentials—compensation beads, buffers, assays & more. Use code RFLOW25 at checkout. The more you buy, the more you save. Limited time only!Learn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
00-4970-93Promo Image100 μL
00-4970-03Promo Image4 x 100 μL
Catalog number 00-4970-93
Price (USD)
131.00
Each
In stock
Add to cart
Quantity:
100 μL
Recurring order eligible. Learn more »
Price (USD)
131.00
Each
Add to cart
Ask our AI about this Product
The Cell Stimulation Cocktail (500X) is a cocktail of phorbol 12-myristate 13-acetate (PMA) and ionomycin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines. This cocktail can be used to induce cytokine production in in vitro cell culture for subsequent detection in immunoassays. It can also be used in conjunction with the Protein Transport Inhibitor Cocktail (500X) (cat. 00-4980) for intracellular retention of the induced cytokine expression.

Reported Application
Flow Cytometric Analysis, Functional Assays, Cell Culture, ELISA

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity100 μL
Volume (Metric)100μL
Product LineeBioscience™
Product TypeCell Stimulation Cocktail
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3137772Certificate of AnalysisJun 12, 202500-4970-93
3051961Certificate of AnalysisMar 13, 202500-4970-93
3010925Certificate of AnalysisFeb 08, 202500-4970-93
2916128Certificate of AnalysisOct 21, 202400-4970-93
2891274Certificate of AnalysisApr 25, 202400-4970-93
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection