Terminal Deoxynucleotidyl Transferase, recombinant
Terminal Deoxynucleotidyl Transferase, recombinant
Invitrogen™

Terminal Deoxynucleotidyl Transferase, recombinant

Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is a DNA polymerase that catalyzes the addition of deoxynucleotides to the 3fi hydroxyl terminusRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
10533065Promo Image500 U
10533073Promo Image3 x 500 U
Catalog number 10533065
Price (USD)
291.65
Online Exclusive
312.00
Save 20.35 (7%)
Each
In stock
Add to cart
Quantity:
500 U
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
291.65
Online Exclusive
312.00
Save 20.35 (7%)
Each
Add to cart
Ask our AI about this Product
Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is a DNA polymerase that catalyzes the addition of deoxynucleotides to the 3fi hydroxyl terminus of DNA. A TdT Technical Bulletin is available.

Applications:
Homopolymer tailing of vector and insert for cloning. Labeling oligonucleotides with biotin (1,2), 32P- or 35S-label (3), or in apoptosis (TUNEL) (4,5).

Source:
Purified from E. coli clone of calf thymus TdT.

Performance and Quality Testing:
Endonuclease, 3´ and 5´ exodeoxyribonuclease, and levels of incorporation tested.

Unit Definition:
One unit incorporates 1 nmol dATP into acid-precipitable material in 1 h at 37°C, using d(pA)50 as a primer.

Hazard Warning:
Toxic; potassium cacodylate contained in reaction buffer. Also contains cobalt chloride, a highly toxic chemical.
See MSDS.

Unit Reaction Conditions:
0.2 M potassium cacodylate (pH 7.2), 10 mM MgO4 C4 H6 , 1 mM 2-mercaptoethanol, 0.5 mg/ml BSA,
100 flM d(pA)50, 1 mM [3H]dATP, and enzyme in 0.15 ml for 1 h at 37°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible Buffer5X Buffer, Reaction Buffer
Product TypeTdT
Quantity500 U
Shipping ConditionApproved for shipment on Wet or Dry Ice
EnzymeTerminal Deoxynucleotidyl Transferase
Unit SizeEach
Contents & Storage
Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is supplied with vial of 5X buffer [500 mM potassium cacodylate (pH 7.2), 10 mM CoCl2 , 1 mM DTT]. Store at -20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3165193Certificate of AnalysisJun 06, 202510533065
3107155Certificate of AnalysisMar 17, 202510533065
3086185Certificate of AnalysisDec 09, 202410533065
3103885Certificate of AnalysisNov 27, 202410533065
2954105Certificate of AnalysisNov 22, 202410533065
5 results displayed, search above for a specific certificate

Safety Data Sheets

Scientific Resources

Frequently asked questions (FAQs)

Yes, but not the specific one from that same kit. We have the following comparable Terminal Deoxynucleotidyl Transferases (Cat. No. 10533065, 10533073 or EP0161, EP0162).

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Gel Electrophoresis and Blotting Support Center.

Citations & References (9)

Citations & References
Abstract
Interaction of human nuclear topoisomerase I with guanosine quartet-forming and guanosine-rich single-stranded DNA and RNA oligonucleotides.
Authors: Marchand Christophe; Pourquier Philippe; Laco Gary S; Jing Naijie; Pommier Yves;
Journal:J Biol Chem
PubMed ID:11756434
'Human nuclear DNA topoisomerase I (top1) plays a crucial role in DNA replication, transcription, and chromosome condensation. In this study, we show that intra- and intermolecular guanosine quartets (G-quartets) can inhibit top1-mediated DNA cleavage at a high affinity site. Top1-mediated DNA cleavage was also inhibited by a 16-mer single-stranded oligodeoxynucleotide ... More
The novel WD-repeat protein MORG1 acts as a molecular scaffold for HIF prolyl-hydroxylase 3 (PHD3).
Authors:Hopfer U, Hopfer H, Jablonski K, Stahl RA, Wolf G,
Journal:J Biol Chem
PubMed ID:16407229
'Hypoxia-inducible factor-1 (HIF-1), a transcriptional complex composed of an oxygen-sensitive alpha- and a beta-subunit, plays a pivotal role in cellular adaptation to low oxygen availability. Under normoxia, the alpha-subunit of HIF-1 is hydroxylated by a family of prolyl hydroxylases (PHDs) and consequently targeted for proteasomal degradation. Three different prolyl hydroxylases ... More
Coordinated transcription of key pathways in the mouse by the circadian clock.
Authors: Panda Satchidananda; Antoch Marina P; Miller Brooke H; Su Andrew I; Schook Andrew B; Straume Marty; Schultz Peter G; Kay Steve A; Takahashi Joseph S; Hogenesch John B;
Journal:Cell
PubMed ID:12015981
'In mammals, circadian control of physiology and behavior is driven by a master pacemaker located in the suprachiasmatic nuclei (SCN) of the hypothalamus. We have used gene expression profiling to identify cycling transcripts in the SCN and in the liver. Our analysis revealed approximately 650 cycling transcripts and showed that ... More
DEMETER, a DNA glycosylase domain protein, is required for endosperm gene imprinting and seed viability in arabidopsis.
Authors: Choi Yeonhee; Gehring Mary; Johnson Lianna; Hannon Mike; Harada John J; Goldberg Robert B; Jacobsen Steven E; Fischer Robert L;
Journal:Cell
PubMed ID:12150995
'We isolated mutations in Arabidopsis to understand how the female gametophyte controls embryo and endosperm development. For the DEMETER (DME) gene, seed viability depends only on the maternal allele. DME encodes a large protein with DNA glycosylase and nuclear localization domains. DME is expressed primarily in the central cell of ... More
The orphan nuclear receptor REV-ERBalpha controls circadian transcription within the positive limb of the mammalian circadian oscillator.
Authors: Preitner Nicolas; Damiola Francesca; Lopez-Molina Luis; Zakany Joszef; Duboule Denis; Albrecht Urs; Schibler Ueli;
Journal:Cell
PubMed ID:12150932
Mammalian circadian rhythms are generated by a feedback loop in which BMAL1 and CLOCK, players of the positive limb, activate transcription of the cryptochrome and period genes, components of the negative limb. Bmal1 and Per transcription cycles display nearly opposite phases and are thus governed by different mechanisms. Here, we ... More
9 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection