Terminal Deoxynucleotidyl Transferase (TdT) Buffer
Product Image
Invitrogen™

Terminal Deoxynucleotidyl Transferase (TdT) Buffer

Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is a DNA polymerase that catalyzes the addition of deoxynucleotides to the 3fi hydroxyl terminusRead more
Have Questions?
Catalog NumberQuantity
163140152 x 0.8 mL
Catalog number 16314015
Price (USD)
88.75
Each
-
Add to cart
Quantity:
2 x 0.8 mL
Request bulk or custom format
Price (USD)
88.75
Each
Add to cart
Ask our AI about this Product
Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is a DNA polymerase that catalyzes the addition of deoxynucleotides to the 3fi hydroxyl terminus of DNA. A TdT Technical Bulletin is available.

Applications:
Homopolymer tailing of vector and insert for cloning. Labeling oligonucleotides with biotin (1,2), 32P- or 35S-label (3), or in apoptosis (TUNEL) (4,5).

Source:
Purified from a baculovirus clone of calf thymus TdT.

Performance and Quality Testing:
Endonuclease, 3´ and 5´ exodeoxyribonuclease, and levels of incorporation tested.

Unit Definition:
One unit incorporates 1 nmol dATP into acid-precipitable material in 1 h at 37°C, using d(pA)50 as a primer.

Hazard Warning:
Toxic; potassium cacodylate contained in reaction buffer. Also contains cobalt chloride, a highly toxic chemical.
See MSDS.

Unit Reaction Conditions:
0.2 M potassium cacodylate (pH 7.2), 10 mM MgO4 C4 H6 , 1 mM 2-mercaptoethanol, 0.5 mg/ml BSA,
100 flM d(pA)50, 1 mM [3H]dATP, and enzyme in 0.15 ml for 1 h at 37°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible Buffer5X Buffer, Reaction Buffer
Product TypeTdT
Quantity2 x 0.8 mL
Shipping ConditionApproved for shipment on Wet or Dry Ice
Unit SizeEach
Contents & Storage
Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is supplied with vial of 5X buffer [500 mM potassium cacodylate (pH 7.2), 10 mM CoCl2 , 1 mM DTT]. Store at -20°C.
Have questions about this product? Ask our AI assisted search.
What is the stability of Thermo Scientific restriction enzymes if they freeze-thaw during shipment?
Can I double digest my DNA using a Thermo Scientific conventional restriction enzyme and a FastDigest restriction enzyme?
Do Thermo Scientific restriction enzymes come with the buffer(s)?
Why do you recommend only 2 µL of 10X Reaction Buffer when digesting unpurified PCR product in a 30 µL reaction?
What are key factors promoting star activity?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Los clientes que vieron este artículo también vieron



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2947667Certificate of AnalysisJun 25, 202416314-015, 16314015
2748692Certificate of AnalysisOct 06, 202316314015
2563721Certificate of AnalysisApr 11, 202316314015
2525138Certificate of AnalysisSep 30, 202216314-015, 16314015
1652941Certificate of AnalysisJul 21, 201716314015
5 results displayed, search above for a specific certificate

Safety Data Sheets

Scientific Resources

Citations & References (1)

Citations & References
Abstract
Different effects on human topoisomerase I by minor groove and intercalated deoxyguanosine adducts derived from two polycyclic aromatic hydrocarbon diol epoxides at or near a normal cleavage site.
Authors: Pommier Yves; Kohlhagen Glenda; Laco Gary S; Kroth Heiko; Sayer Jane M; Jerina Donald M;
Journal:J Biol Chem
PubMed ID:11832494
Topoisomerase I (top1) relieves supercoiling in DNA by forming transient covalent cleavage complexes. These cleavage complexes can accumulate in the presence of damaged DNA or anticancer drugs that either intercalate or lie in the minor groove. Recently we reported that covalent diol epoxide (DE) adducts of benzo[a]pyrene (BaP) at the ... More
1 total citations

Share catalog number, name or link

1x1 image pixel for data collection