1x1 image pixel for data collection
ELISA Reagent Reservoir
ELISA Reagent Reservoir
Thermo Scientific™

ELISA Reagent Reservoir

Thermo Scientific Reagent Reservoirs are sterile, sturdy, disposable and molded from high-impact polystyrene. Reservoirs facilitate repeated multi-channel pipetting for reagentRead more
Promo ImagePromotion
Promo code:RBLOT25
Save up to 24% on Western Blot Reagents and Gels
From now until June 30th, get 16% off when you spend $400 or more, and get 24% off when you spend $750 or more on select western blot reagent and protein gels.Learn More
Have Questions?
Catalog NumberQuantity
15075Promo Image200 Units
Catalog number 15075
Price (USD)
200.65
Online Exclusive
227.00
Save 26.35 (12%)
Each
In stock
Add to cart
Quantity:
200 Units
Recurring order eligible. Learn more »
Price (USD)
200.65
Online Exclusive
227.00
Save 26.35 (12%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Reagent Reservoirs are sterile, sturdy, disposable and molded from high-impact polystyrene. Reservoirs facilitate repeated multi-channel pipetting for reagent delivery to microplates.

Features of the Reagent Reservoirs:

• Sterile and durable reservoirs molded from high-impact polystyrene
• Reservoirs facilitate repeated pick-up of liquid reagents by multi-channel pipettors for delivery to 96-well plates
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce ELISA Reagent Reservoirs
Plate Compatibility96-Well Plate, 384-Well Plate
Quantity200 Units
Bottom ShapeV-bottom
Capacity (Metric)50 mL
DisposableSingle-use
MaterialPolystyrene
SterilitySterile
TypeReagent Reservoir
Unit SizeEach
Contents & Storage
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3216681ACertificate of AnalysisJul 16, 202515075
3216681Certificate of AnalysisJul 16, 202515075
3212853CCertificate of AnalysisMay 01, 202515075
3212853BCertificate of AnalysisMay 01, 202515075
3212853ACertificate of AnalysisMay 01, 202515075
5 results displayed, search above for a specific certificate

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection