Informational:This product has been Gibco FBS Fingerprinted. We developed a technology in partnership with Oritain, a global leader in verifying product origins. Based on geographical differences, we can create "fingerprints" of our FBS that connect them to their origin. Find out more ›
Gibco™

Fetal Bovine Serum, certified, United States

Catalog number: 16000069
Have Questions?
Gibco™

Fetal Bovine Serum, certified, United States

Catalog number: 16000069
Have Questions?
Catalog Number
16000069
Unit Size
1,000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
160000691,000 mL
-
A316040150 mL
-
A316040210 x 50 mL
-
16000044500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

Gibco Sera: Trusted Quality for Consistent Performance

Sera Category: Premium (Performance Plus)

  • Use for a broad range of cell types, especially sensitive cell lines
  • Our most characterized sera with the lowest endotoxin and hemoglobin levels
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤15 mg/dL
  • Origin: United States

Gibco Serum Delivers

  • ISO 13485 certified, processed in FDA registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications

Age
Fetal
Species
Cattle/Bovine
Country of Origin
United States
Form
Liquid
Format
Bottle
Product Type
Fetal Bovine Serum
Quantity
1000 mL
Serum Treatment
Standard (Sterile-filtered)
Sterility
Sterile
Sterilization Method
Triple-filtered, 0.1 μm
Endotoxin Concentration
≤5 EU/mL
Green Features
Sustainable packaging
Hemoglobin Concentration
≤15 mg/dL
Purity or Quality Grade
Certified
Shipping Condition
Frozen

Contents & Storage

Storage conditions: ≤-10°C
Shipping conditions: Frozen

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text