Informational:

***ATTENTION***

*** NEW CATALOG NUMBER***



These products are being replaced with Premium Plus (FBS) performance-based products. The Premium product specifications are improved and product stability, performance, manufacturing, storage, and transport processes are unchanged.

New and improved replacement available, Buy now›
Gibco™

Fetal Bovine Serum, certified, One Shot™ format, United States

Catalog number: A3160402
Have Questions?
Gibco™

Fetal Bovine Serum, certified, One Shot™ format, United States

Catalog number: A3160402
Have Questions?
Catalog Number
A3160402
Unit Size
10 x 50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A316040210 x 50 mL
-
A316040150 mL
-
16000044500 mL
-
160000691,000 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

Gibco Sera: Trusted Quality for Consistent Performance

Sera Category: Premium (Performance Plus)

  • Use for a broad range of cell types, especially sensitive cell lines
  • Our most characterized sera with the lowest endotoxin and hemoglobin levels
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤15 mg/dL
  • Origin: United States

Gibco Serum Delivers

  • ISO 13485 certified, processed in FDA registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications

Description
Fetal Bovine Serum, certified, One Shot™ format, United States
Age
Fetal
Species
Bovine
Country of Origin
United States
Form
Liquid
Format
One Shot
Product Type
Fetal Bovine Serum
Quantity
10 x 50 mL
Serum Treatment
Standard (Sterile-filtered)
Sterility
Sterile
Sterilization Method
Triple-filtered, 0.1 μm
Endotoxin Concentration
≤5 EU/mL
Green Features
Fewer resources used and less waste
Hemoglobin Concentration
≤15 mg/dL
Purity or Quality Grade
Certified
Product Line
One Shot™
Shipping Condition
Frozen

Contents & Storage

Storage conditions: ≤-10°C
Shipping conditions: Frozen

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text