Potassium phosphate, monobasic, 99+%, extra pure
Potassium phosphate, monobasic, 99+%, extra pure
Potassium phosphate, monobasic, 99+%, extra pure
Potassium phosphate, monobasic, 99+%, extra pure
Thermo Scientific Chemicals

Potassium phosphate, monobasic, 99+%, extra pure

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
10 kg
Packaging:
View all
Plastic bottle
Catalog number 205925000
Price (USD)
94.65
Special offer
Online exclusive
111.00
Save 16.35 (15%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
94.65
Special offer
Online exclusive
111.00
Save 16.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7778-77-0
IUPAC Namepotassium dihydrogen phosphate
Molecular FormulaH2KO4P
InChI KeyGNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES[K+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=99.0 % (on dried substance)
pH4.2 to 4.7 (1 % at 20°C)
Appearance (Form)Crystalline powder or crystals
Loss on drying=<1.0 % (105°C)
Arsenic (As)=<3 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470214Certificate of AnalysisMay 17, 2025205925000, 205920025, 205920100
A0472239Certificate of AnalysisMay 17, 2025205925000, 205920025, 205920100
A0464403Certificate of AnalysisJul 25, 2024205925000, 205920025, 205920100
A0457830Certificate of AnalysisDec 08, 2023205925000, 205920025, 205920100
A0441958Certificate of AnalysisSep 27, 2023205925000, 205920025, 205920100
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection