Sodium molybdate(VI) dihydrate, 99+%
Sodium molybdate(VI) dihydrate, 99+%
Sodium molybdate(VI) dihydrate, 99+%
Sodium molybdate(VI) dihydrate, 99+%
Thermo Scientific Chemicals

Sodium molybdate(VI) dihydrate, 99+%

CAS: 10102-40-6 | MoNa2O4 | 205.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 206375000
Price (USD)
181.65
Online Exclusive
202.00
Save 20.35 (10%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
181.65
Online Exclusive
202.00
Save 20.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS10102-40-6
IUPAC Namedisodium dioxomolybdenumbis(olate) dihydrate
Molecular FormulaH4MoNa2O6
InChI KeyFDEIWTXVNPKYDL-UHFFFAOYSA-N
SMILESO.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=99.0 %
Appearance (Form)Crystalline powder
Appearance (Color)White
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0467027Certificate of AnalysisOct 23, 2024206371000, 206370025, 206375000, 206370050
A0467028Certificate of AnalysisOct 23, 2024206371000, 206370025, 206375000, 206370050
A0450390Certificate of AnalysisFeb 26, 2024206371000, 206370025, 206375000, 206370050
A0444522Certificate of AnalysisAug 25, 2023206371000, 206370025, 206375000, 206370050
A0434076Certificate of AnalysisSep 21, 2022206371000, 206370025, 206375000, 206370050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection