Sodium hypochlorite, 10-15% active chlorine
Sodium hypochlorite, 10-15% active chlorine
Sodium hypochlorite, 10-15% active chlorine
Sodium hypochlorite, 10-15% active chlorine
Thermo Scientific Chemicals

Sodium hypochlorite, 10-15% active chlorine

CAS: 7681-52-9 | ClNaO | 74.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
10 L
Packaging:
View all
Plastic bottle
Catalog number 219250025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7681-52-9
IUPAC Namesodium hypochlorite
Molecular FormulaClNaO
InChI KeySUKJFIGYRHOWBL-UHFFFAOYSA-N
SMILES[Na+].[O-]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Free alkali (NaOH)=<1.0 %
Appearance (Color)Clear yellow
Titration Iodometric10 to 15 %
Iron (Fe)=<5 ppm
Appearance (Form)Solution
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

해당 제품을 검색한 고객들이 함께 검색한 제품



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473043Certificate of AnalysisMay 29, 2025219250100, 219255000, 219250025
A0471522Certificate of AnalysisMay 17, 2025219250100, 219255000, 219250025
A0462985Certificate of AnalysisJan 31, 2025219250100, 219255000, 219250025
A0467172Certificate of AnalysisNov 01, 2024219250100, 219255000, 219250025
A0462985Certificate of AnalysisMay 20, 2024219250100, 219255000, 219250025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection