Titanium(IV) oxide, 98.0-100.5% TiO2
Titanium(IV) oxide, 98.0-100.5% TiO2
Titanium(IV) oxide, 98.0-100.5% TiO2
Titanium(IV) oxide, 98.0-100.5% TiO2
Titanium(IV) oxide, 98.0-100.5% TiO2
Thermo Scientific Chemicals

Titanium(IV) oxide, 98.0-100.5% TiO2

CAS: 13463-67-7 | O2Ti | 79.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
10 kg
Packaging:
View all
Glass Bottle
Catalog number 277370010
Price (USD)
66.65
Special offer
Online exclusive
Ends: 26-Sep-2025
82.20
Save 15.55 (19%)
Each
In stock
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
66.65
Special offer
Online exclusive
Ends: 26-Sep-2025
82.20
Save 15.55 (19%)
Each
Add to cart
Chemical Identifiers
CAS13463-67-7
IUPAC Namedioxotitanium
Molecular FormulaO2Ti
InChI KeyGWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILESO=[Ti]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration98.0 to 100.5 %
Soluble matter=<0.5 % (in water)
Arsenic (As)=<5 ppm
Acidity/alkalinityPasses test
Appearance (Form)Powder
View more
Ask our AI about this Product

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470571Certificate of AnalysisMay 17, 2025277370010, 277370100
A0464725Certificate of AnalysisJul 25, 2024277370010, 277370100
A0458515Certificate of AnalysisDec 04, 2023277370010, 277370100
A0458523Certificate of AnalysisDec 04, 2023277370010, 277370100
A0451876Certificate of AnalysisMay 02, 2023277370010, 277370100
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection