Nunc™ 96-Well Filter Plates
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ 96-Well Filter Plates
Actual product may vary
Thermo Scientific™

Nunc™ 96-Well Filter Plates

Efficiently perform plasmid DNA filtration, PCR filtration and vacuum filtration with these Thermo Scientific™ Nunc™ 96-well filter plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
278011Fritted 96 Well DeepWell
278010Fritted Deep Well Plate w/ Glass Fiber (Binding)
278012Unfritted Deep Well Plate
Catalog number 278011
Price (USD)
1,311.65
Online Exclusive
1,352.00
Save 40.35 (3%)
Case of 50
In stock
Add to cart
Product Type:
Fritted 96 Well DeepWell
Recurring order eligible. Learn more »
Price (USD)
1,311.65
Online Exclusive
1,352.00
Save 40.35 (3%)
Case of 50
Add to cart
Ask our AI about this Product
Efficiently perform purification of high quality plasmid DNA or PCR products with the Thermo Scientific™ Nunc™ 96 Deep Well Filter Plates. Made of chemical resistant polypropylene material with superior mechanical strength, these plates conform to standard 96 well footprint compatible with high throughput sample processing.
Key Features:
  • Designed specifically for high throughput sample processing
  • Shared wall configuration gives reduced plate height, wider wells for faster pocessing, higher yields and less clogging
  • Suitable for vacuum, filtration, centrifugatioin or positive pressure techniques
  • Polyethylene terapthalate (PET) frit in the fritted plates (278010, 278011) has 20μm pore size
  • Glass Fiber plate (278010) has the PET frit plus two layers of glass fiber membrane for efficient binding of DNA
  • Customized bar coding is available
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
LidWithout Lid
MembraneGlass Fiber
No. per Case50
No. per Pack5
Pore Size20 μm
Product TypeFritted 96 Well DeepWell
Quantity50/Cs.
Volume (Metric) Working1 mL
Array8 x 12
ColorNatural
FormatStandard
MaterialPolypropylene
No. of Wells96
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeFilter
Unit SizeCase of 50
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
1313933Certificate of ComplianceDec 11, 2024278011
1282376Certificate of ComplianceDec 11, 2024278011
1302491Certificate of ComplianceDec 11, 2024278010
1314529Certificate of ComplianceDec 11, 2024278010
1336390Certificate of ComplianceDec 11, 2024278010
5 results displayed, search above for a specific certificate

Frequently asked questions (FAQs)

29.936 in. HG equals 1.013 bars
So -5 to -10 in. Hg equals -170 to -340 mbar and -15 to -20 in. Hg equals -500 to -680 mbar

Any vacuum manifold (for 96-well microplates) with an inside depth of 4 cm or more should be fine.

Yes. The polypropylene plate material naturally resists chemicals and organic solvents, as does the polyethylene terephthalate (PET) frit and glass fiber filter. For very strong chemicals, prolonged exposure times, and/or higher temperatures, we recommend that you test the plates with the chemical or solution in question.

The protocol is designed to be flexible, depending on customer needs and instrumentation. For example, certain steps can be performed with vacuum filtration, centrifugation, or positive pressure, with similar results. We do recommend that customers use our formulas for buffers and solutions, since they were optimized to produce superior results.

We recommend using the following Nunc products:

- 96-Well Polypropylene DeepWell Storage Plate, 2 mL (Cat. No. 278743) (sterile growth plate)
- Microplate Lid for 96 microwell plate (Cat. No. 264122) (to cover plate during growth)
- Polyolefin Acrylate Sealing Tape (Cat. No. 232701) (to cover plate during mixing)
- Fritted 96-Well Deep Well Filter Plate (Cat. No. 278011) (for lysate clarification)
- Fritted 96-Well Deep Well Filter Plate with Glass Fiber Filter (Cat. No. 278010) (for DNA binding)
- 96-Well Polypropylene DeepWell Storage Plate, 1.3 mL (Cat. No. 260252) (reception plate)

Share catalog number, name or link

1x1 image pixel for data collection