Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Sterility | Well Shape | Fit for Purpose |
---|---|---|---|---|
3734 | 0.5 mL | Non-sterile | V-bottom | PCR Ready |
3719-11 | 1.4 mL | Non-sterile | Flat | PCR Ready |
3725TS also known as 3725 | 0.5 mL | Sterile | V-bottom | Sterile and PCR Ready |
3715-11 | 1.4 mL | Non-sterile | Flat | PCR Ready |
3710 | 1.4 mL | Non-sterile | Flat | PCR Ready |
3730-11 | 0.75 mL | Non-sterile | V-bottom | PCR Ready |
3731-11 | 0.75 mL | Non-sterile | V-bottom | PCR Ready |
3712-11 | 1.4 mL | Sterile | Flat | Sterile and PCR Ready |
3711 | 1.4 mL | Non-sterile | Flat | PCR Ready |
3712 | 1.4 mL | Sterile | Flat | Sterile and PCR Ready |
3730 | 0.75 mL | Non-sterile | V-bottom | Sterile and PCR Ready |
3732 | 0.75 mL | Sterile | V-bottom | Sterile and PCR Ready |
3735 | 500 μL | Sterile | V-bottom | Sterile and PCR Ready |
3750 | 500 μL | Non-sterile | V-bottom | PCR Ready |
3790 | 1.4 mL | Non-sterile | V-bottom | PCR Ready |
3791 | 1.4 mL | Non-sterile | V-bottom | PCR Ready |
3792 | 1.4 mL | Sterile | V-bottom | Sterile and PCR Ready |
Store and track samples in Thermo Scientific™ Matrix™ 2D Barcoded Clear Polypropylene Open-Top Storage Tubes. Laser etching produces a permanent, high-contrast, 2D barcode offering maximum reliability and traceability of compound, biological and genomic samples. Offered in three sizes, 0.5mL, 0.75mL, and 1.4mL, these tubes can be sealed with Thermo Scientific SepraSeal septum caps and stored down to -80°C. Item number 3792 is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Secure Tracking:
Robust Storage Rack Design:
Stringent Quality Control:
Storage Tracking Solutions for Any Laboratory:
Recommended for:
Individual sample retrieval, instant sample identification, secure identification of archived samples, secure shipping of samples, storage of samples and tracking of samples.
Recycling Information:
Matrix tubes, tube racks and covers are made from polypropylene copolymer and are recyclable in communities where polypropylene is recycled. Please recycle only clean, dry components that have been decontaminated of any hazardous materials or residues. The orange nylon rack latches must be removed from the tube rack base and discarded before recycling the rack base. Tip: Orange latches can be removed with relative ease with the aid of pliers.
Matrix tubes are delivered in high graphic paper cartons. Please recycle the paper cartons and product inserts.
Gain confidence that the products you buy are ideally suited to your application.
When you're looking for sustainable products, we believe it should be easy to make informed purchasing decisions. We're working to provide more information about our products' environmental impact by stepping up to participate in the ACT Label program with a selection of popular Thermo Scientific™ Nalgene™ and Matrix™ products.
Learn more ›
Yes, the Thermo Scientific 2D barcoded storage tubes can be stored in standard cryoboxes. However, we do not carry 2D barcode readers capable of reading a full box of 2D barcodes in this storage format.
The 2D barcode laser etching software includes algorithms that prevent the barcode generator from ever counting backwards, subsequently preventing the re-etching of any previously generated barcode. To verify the above procedure, several quality control checks are performed on the 2D barcodes including:
- Checking for light transmission through the barcode as light transmission will reduce readability
- Inspecting the barcode under magnification
- Reading the barcode on a full rack 2D barcode reader
- Checking the barcode against a database of previously etched 2D barcodes
As the above quality control steps are completed, defective storage tubes are discarded, replaced, and re-checked, ensuring that the 2D barcodes are 100% readable and have not been previously used. This quality control process is the reason why we cannot guarantee sequential 2D barcode printing within a single rack of 2D barcoded products.
The Thermo Scientific 2D barcoded storage tubes have been subjected in-house to consecutive liquid nitrogen freeze-thaws followed by submersion in boiling water. These tests did not cause any damage to the 2D barcode.
The 2D barcodes are made of the same material as the storage tubes and are resistant to standard lab chemicals.
The 2D barcode has a built-in ECC200 error barcode correction, which ensures that a damaged barcode is still readable as long as less than 25% of the barcode is missing.
Share catalog number, name or link